Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2860999-2861224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS16
Entrez Gene ID
64601
Gene Name
VPS16, CORVET/HOPS core subunit
Gene Aliases
hVPS16
UniGene
Hs.269577
Gene Chromosome Location
Chr.20: 2840697-2866732 on Build GRCh38
Transcripts
NM_022575.3 NM_080413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6055107 c.729C>G p.I243M Chr.20: 2861068-2861068 on Build GRCh38
366380 c.672G>T p.Q224H Chr.20: 2861011-2861011 on Build GRCh38
316477 c.731G>T p.W244L Chr.20: 2861070-2861070 on Build GRCh38
1025494 c.699C>A p.H233Q Chr.20: 2861038-2861038 on Build GRCh38
264660 c.695G>A p.R232Q Chr.20: 2861034-2861034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200746035 Chr.20: 2861109-2861109 on Build GRCh38
rs768164425 Chr.20: 2861115-2861115 on Build GRCh38
rs1031816625 Chr.20: 2861147-2861147 on Build GRCh38
rs117270313 Chr.20: 2861171-2861171 on Build GRCh38
rs6076436 Chr.20: 2861126-2861126 on Build GRCh38
rs192393227 Chr.20: 2861116-2861116 on Build GRCh38
rs767329493 Chr.20: 2861026-2861026 on Build GRCh38
rs376038159 Chr.20: 2861158-2861158 on Build GRCh38
rs746714433 Chr.20: 2861053-2861053 on Build GRCh38
rs770157144 Chr.20: 2861103-2861103 on Build GRCh38
rs374585828 Chr.20: 2861170-2861170 on Build GRCh38
rs747158866 Chr.20: 2861077-2861077 on Build GRCh38
rs750215291 Chr.20: 2861031-2861031 on Build GRCh38
rs889916747 Chr.20: 2861036-2861036 on Build GRCh38
rs778947851 Chr.20: 2861189-2861189 on Build GRCh38
rs759689816 Chr.20: 2861134-2861134 on Build GRCh38
rs369133033 Chr.20: 2861092-2861092 on Build GRCh38
rs201669561 Chr.20: 2861059-2861059 on Build GRCh38
rs746218240 Chr.20: 2861100-2861100 on Build GRCh38
rs532737090 Chr.20: 2861139-2861139 on Build GRCh38
rs551279022 Chr.20: 2861157-2861157 on Build GRCh38
rs756043022 Chr.20: 2861034-2861034 on Build GRCh38
rs185353565 Chr.20: 2861138-2861138 on Build GRCh38
rs771109557 Chr.20: 2861080-2861080 on Build GRCh38
rs543158583 Chr.20: 2861052-2861052 on Build GRCh38
rs376025545 Chr.20: 2861038-2861038 on Build GRCh38
rs759921592 Chr.20: 2861108-2861108 on Build GRCh38
rs749201066 Chr.20: 2861054-2861054 on Build GRCh38
rs750162214 Chr.20: 2861136-2861136 on Build GRCh38
rs754845363 Chr.20: 2861144-2861144 on Build GRCh38
rs777734312 Chr.20: 2861058-2861058 on Build GRCh38
rs760493091 Chr.20: 2861137-2861137 on Build GRCh38
rs775999383 Chr.20: 2861106-2861106 on Build GRCh38
rs758460846 Chr.20: 2861057-2861057 on Build GRCh38

View Full Product Details