Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209763253-209763508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF3IP3
Entrez Gene ID
80342
Gene Name
TRAF3 interacting protein 3
Gene Aliases
T3JAM
UniGene
Hs.147434
Gene Chromosome Location
Chr.1: 209756032-209782323 on Build GRCh38
Transcripts
NM_001320143.1 NM_001320144.1 NM_025228.3 NR_109871.1 XM_011510018.2 XM_011510019.1 XM_017002399.1 XM_017002400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5102126 c.607-10G>A p.? Chr.1: 209763482-209763482 on Build GRCh38
4027757 c.611C>T p.A204V Chr.1: 209763496-209763496 on Build GRCh38
4912589 c.554T>A p.L185Q Chr.1: 209763499-209763499 on Build GRCh38
6807249 c.517G>T p.E173* Chr.1: 209763363-209763363 on Build GRCh38
5102127 c.547-10G>A p.? Chr.1: 209763482-209763482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765298927 Chr.1: 209763479-209763479 on Build GRCh38
rs759589265 Chr.1: 209763464-209763464 on Build GRCh38
rs143123645 Chr.1: 209763437-209763437 on Build GRCh38
rs946845726 Chr.1: 209763283-209763283 on Build GRCh38
rs1005856443 Chr.1: 209763431-209763431 on Build GRCh38
rs759536263 Chr.1: 209763332-209763332 on Build GRCh38
rs887700896 Chr.1: 209763477-209763477 on Build GRCh38
rs912954027 Chr.1: 209763345-209763345 on Build GRCh38
rs750677077 Chr.1: 209763487-209763487 on Build GRCh38
rs769614934 Chr.1: 209763484-209763485 on Build GRCh38
rs376499875 Chr.1: 209763451-209763451 on Build GRCh38
rs765176027 Chr.1: 209763338-209763338 on Build GRCh38
rs781292720 Chr.1: 209763370-209763370 on Build GRCh38
rs772878168 Chr.1: 209763319-209763319 on Build GRCh38
rs780103712 Chr.1: 209763407-209763407 on Build GRCh38
rs772798933 Chr.1: 209763442-209763442 on Build GRCh38
rs896435955 Chr.1: 209763386-209763386 on Build GRCh38
rs756315411 Chr.1: 209763488-209763488 on Build GRCh38
rs372288997 Chr.1: 209763489-209763489 on Build GRCh38
rs749852050 Chr.1: 209763408-209763408 on Build GRCh38
rs770627085 Chr.1: 209763444-209763444 on Build GRCh38
rs570651167 Chr.1: 209763320-209763320 on Build GRCh38
rs772429358 Chr.1: 209763441-209763441 on Build GRCh38
rs756190745 Chr.1: 209763405-209763405 on Build GRCh38
rs553362520 Chr.1: 209763481-209763481 on Build GRCh38
rs762671547 Chr.1: 209763482-209763482 on Build GRCh38
rs752405734 Chr.1: 209763347-209763347 on Build GRCh38
rs763883521 Chr.1: 209763485-209763485 on Build GRCh38
rs944476022 Chr.1: 209763421-209763421 on Build GRCh38
rs758174264 Chr.1: 209763366-209763366 on Build GRCh38
rs769128511 Chr.1: 209763428-209763428 on Build GRCh38
rs753898183 Chr.1: 209763331-209763331 on Build GRCh38
rs552370575 Chr.1: 209763307-209763307 on Build GRCh38
rs371695095 Chr.1: 209763433-209763433 on Build GRCh38
rs7536882 Chr.1: 209763328-209763328 on Build GRCh38
rs887426596 Chr.1: 209763411-209763411 on Build GRCh38
rs771590550 Chr.1: 209763313-209763313 on Build GRCh38
rs1056274802 Chr.1: 209763368-209763368 on Build GRCh38
rs6540556 Chr.1: 209763286-209763286 on Build GRCh38
rs750601439 Chr.1: 209763390-209763390 on Build GRCh38
rs748489993 Chr.1: 209763438-209763438 on Build GRCh38
rs368909821 Chr.1: 209763452-209763452 on Build GRCh38
rs535040314 Chr.1: 209763440-209763440 on Build GRCh38

View Full Product Details