Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156378281-156378518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBG
Entrez Gene ID
57127
Gene Name
Rh family B glycoprotein (gene/pseudogene)
Gene Aliases
SLC42A2
UniGene
Hs.131835
Gene Chromosome Location
Chr.1: 156366044-156385221 on Build GRCh38
Transcripts
NM_001256395.1 NM_001256396.1 NM_020407.4 NR_046115.1 XM_011509799.2 XM_011509800.2 XM_017001857.1 XM_017001858.1 XM_017001859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1983439 c.546C>T p.S182S Chr.1: 156378368-156378368 on Build GRCh38
5341601 c.465C>G p.T155T Chr.1: 156378287-156378287 on Build GRCh38
7416766 c.494C>A p.S165* Chr.1: 156378316-156378316 on Build GRCh38
6766147 c.547G>A p.V183I Chr.1: 156378369-156378369 on Build GRCh38
3475224 c.464C>T p.T155I Chr.1: 156378286-156378286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534576776 Chr.1: 156378335-156378335 on Build GRCh38
rs768986701 Chr.1: 156378332-156378332 on Build GRCh38
rs765489907 Chr.1: 156378369-156378369 on Build GRCh38
rs267598074 Chr.1: 156378316-156378316 on Build GRCh38
rs114322922 Chr.1: 156378476-156378476 on Build GRCh38
rs894845388 Chr.1: 156378469-156378469 on Build GRCh38
rs538971671 Chr.1: 156378368-156378368 on Build GRCh38
rs750445883 Chr.1: 156378451-156378451 on Build GRCh38
rs921282012 Chr.1: 156378360-156378360 on Build GRCh38
rs1042951819 Chr.1: 156378420-156378420 on Build GRCh38
rs775383585 Chr.1: 156378439-156378439 on Build GRCh38
rs113186314 Chr.1: 156378413-156378413 on Build GRCh38
rs892609782 Chr.1: 156378471-156378472 on Build GRCh38
rs932618607 Chr.1: 156378373-156378373 on Build GRCh38
rs201683131 Chr.1: 156378438-156378438 on Build GRCh38
rs753835039 Chr.1: 156378397-156378397 on Build GRCh38
rs893874874 Chr.1: 156378406-156378406 on Build GRCh38
rs370053677 Chr.1: 156378308-156378308 on Build GRCh38
rs185277732 Chr.1: 156378302-156378302 on Build GRCh38
rs375155417 Chr.1: 156378356-156378356 on Build GRCh38
rs368582633 Chr.1: 156378318-156378318 on Build GRCh38
rs1022342546 Chr.1: 156378477-156378477 on Build GRCh38
rs747713024 Chr.1: 156378401-156378401 on Build GRCh38
rs751504976 Chr.1: 156378303-156378303 on Build GRCh38
rs748956574 Chr.1: 156378417-156378417 on Build GRCh38
rs780949174 Chr.1: 156378365-156378365 on Build GRCh38
rs751822473 Chr.1: 156378389-156378389 on Build GRCh38
rs371596072 Chr.1: 156378474-156378474 on Build GRCh38
rs765295234 Chr.1: 156378446-156378446 on Build GRCh38
rs537105074 Chr.1: 156378315-156378315 on Build GRCh38
rs780482312 Chr.1: 156378385-156378385 on Build GRCh38
rs767305932 Chr.1: 156378361-156378361 on Build GRCh38
rs769418262 Chr.1: 156378410-156378410 on Build GRCh38
rs116575041 Chr.1: 156378443-156378443 on Build GRCh38
rs758827981 Chr.1: 156378383-156378383 on Build GRCh38
rs770429534 Chr.1: 156378418-156378418 on Build GRCh38
rs564983120 Chr.1: 156378491-156378491 on Build GRCh38
rs969917474 Chr.1: 156378486-156378486 on Build GRCh38
rs759526546 Chr.1: 156378358-156378358 on Build GRCh38
rs372135339 Chr.1: 156378393-156378393 on Build GRCh38
rs370851160 Chr.1: 156378319-156378319 on Build GRCh38
rs950524902 Chr.1: 156378477-156378477 on Build GRCh38
rs190623502 Chr.1: 156378357-156378357 on Build GRCh38
rs371780489 Chr.1: 156378317-156378317 on Build GRCh38
rs764234493 Chr.1: 156378309-156378309 on Build GRCh38
rs1033874519 Chr.1: 156378467-156378467 on Build GRCh38
rs998421245 Chr.1: 156378458-156378458 on Build GRCh38
rs750631213 Chr.1: 156378376-156378376 on Build GRCh38
rs755542961 Chr.1: 156378390-156378390 on Build GRCh38
rs575770340 Chr.1: 156378434-156378434 on Build GRCh38
rs553048664 Chr.1: 156378336-156378336 on Build GRCh38
rs762903550 Chr.1: 156378339-156378339 on Build GRCh38

View Full Product Details