Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100400885-100401146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
UniGene
Hs.105191
Gene Chromosome Location
Chr.7: 100400826-100429239 on Build GRCh38
Transcripts
NM_001258008.1 NM_017984.4 XM_005250479.2 XM_005250480.2 XM_006716035.3 XM_006716036.2 XM_006716037.3 XM_006716038.3 XM_006716040.3 XM_017012379.1 XM_017012380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778755 c.1868G>C p.R623T Chr.7: 100401093-100401093 on Build GRCh38
6826015 c.1932G>A p.A644A Chr.7: 100401029-100401029 on Build GRCh38
6826014 c.1819G>A p.E607K Chr.7: 100401142-100401142 on Build GRCh38
1453414 c.1878G>A p.G626G Chr.7: 100401083-100401083 on Build GRCh38
227040 c.1858G>A p.D620N Chr.7: 100401103-100401103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375055540 Chr.7: 100401029-100401029 on Build GRCh38
rs760537716 Chr.7: 100401087-100401087 on Build GRCh38
rs753104858 Chr.7: 100401124-100401124 on Build GRCh38
rs766572733 Chr.7: 100401098-100401098 on Build GRCh38
rs763753444 Chr.7: 100401094-100401094 on Build GRCh38
rs748935435 Chr.7: 100401035-100401035 on Build GRCh38
rs562682793 Chr.7: 100401007-100401007 on Build GRCh38
rs543019705 Chr.7: 100400939-100400939 on Build GRCh38
rs772145336 Chr.7: 100401081-100401081 on Build GRCh38
rs11769886 Chr.7: 100400984-100400984 on Build GRCh38
rs374316095 Chr.7: 100400972-100400972 on Build GRCh38
rs775228992 Chr.7: 100401086-100401086 on Build GRCh38
rs372286297 Chr.7: 100401077-100401077 on Build GRCh38
rs751703224 Chr.7: 100401102-100401102 on Build GRCh38
rs145308367 Chr.7: 100401127-100401127 on Build GRCh38
rs369980465 Chr.7: 100401034-100401034 on Build GRCh38
rs199622831 Chr.7: 100401046-100401046 on Build GRCh38
rs757071349 Chr.7: 100401110-100401120 on Build GRCh38
rs202051814 Chr.7: 100401097-100401097 on Build GRCh38
rs375057602 Chr.7: 100401096-100401096 on Build GRCh38
rs758502357 Chr.7: 100401001-100401001 on Build GRCh38
rs919379773 Chr.7: 100400935-100400935 on Build GRCh38
rs530221721 Chr.7: 100401071-100401071 on Build GRCh38
rs778050195 Chr.7: 100401128-100401128 on Build GRCh38
rs754982870 Chr.7: 100401108-100401108 on Build GRCh38
rs774065913 Chr.7: 100401075-100401075 on Build GRCh38
rs771400582 Chr.7: 100401113-100401113 on Build GRCh38
rs767552122 Chr.7: 100400973-100400973 on Build GRCh38
rs943635185 Chr.7: 100400987-100400987 on Build GRCh38
rs750559439 Chr.7: 100401000-100401000 on Build GRCh38
rs576037279 Chr.7: 100400924-100400924 on Build GRCh38
rs770526648 Chr.7: 100400965-100400965 on Build GRCh38
rs1029119324 Chr.7: 100400948-100400948 on Build GRCh38
rs751615182 Chr.7: 100401016-100401016 on Build GRCh38
rs746973587 Chr.7: 100401072-100401073 on Build GRCh38
rs200988509 Chr.7: 100401030-100401030 on Build GRCh38
rs768091022 Chr.7: 100401121-100401121 on Build GRCh38
rs777732910 Chr.7: 100401027-100401028 on Build GRCh38
rs1024884510 Chr.7: 100401104-100401104 on Build GRCh38
rs773970146 Chr.7: 100400971-100400971 on Build GRCh38
rs755062603 Chr.7: 100401019-100401020 on Build GRCh38
rs752608290 Chr.7: 100400983-100400983 on Build GRCh38

View Full Product Details