Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133351212-133351485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRAP1
Entrez Gene ID
118471
Gene Name
proline rich acidic protein 1
Gene Aliases
PRO1195, UPA
UniGene
Hs.15951
Gene Chromosome Location
Chr.10: 133347340-133352683 on Build GRCh38
Transcripts
NM_001145201.1 NM_145202.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916497 c.126G>T p.E42D Chr.10: 133351431-133351431 on Build GRCh38
7136111 c.81T>C p.P27P Chr.10: 133351386-133351386 on Build GRCh38
4169278 c.110C>A p.S37* Chr.10: 133351415-133351415 on Build GRCh38
2024423 c.118G>A p.D40N Chr.10: 133351423-133351423 on Build GRCh38
1603213 c.128+7C>A p.? Chr.10: 133351440-133351440 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774053299 Chr.10: 133351450-133351450 on Build GRCh38
rs763444008 Chr.10: 133351344-133351344 on Build GRCh38
rs372784990 Chr.10: 133351441-133351441 on Build GRCh38
rs1024568292 Chr.10: 133351436-133351436 on Build GRCh38
rs901837585 Chr.10: 133351271-133351271 on Build GRCh38
rs767435227 Chr.10: 133351375-133351375 on Build GRCh38
rs534371826 Chr.10: 133351339-133351339 on Build GRCh38
rs1040745493 Chr.10: 133351257-133351257 on Build GRCh38
rs897957794 Chr.10: 133351308-133351308 on Build GRCh38
rs374533877 Chr.10: 133351398-133351398 on Build GRCh38
rs894401419 Chr.10: 133351381-133351381 on Build GRCh38
rs1046837954 Chr.10: 133351322-133351322 on Build GRCh38
rs202026951 Chr.10: 133351343-133351343 on Build GRCh38
rs567777046 Chr.10: 133351440-133351440 on Build GRCh38
rs183041551 Chr.10: 133351340-133351340 on Build GRCh38
rs763786594 Chr.10: 133351449-133351449 on Build GRCh38
rs79130099 Chr.10: 133351295-133351295 on Build GRCh38
rs756815704 Chr.10: 133351366-133351366 on Build GRCh38
rs755877755 Chr.10: 133351394-133351394 on Build GRCh38
rs771999563 Chr.10: 133351333-133351333 on Build GRCh38
rs145678335 Chr.10: 133351426-133351426 on Build GRCh38
rs944946273 Chr.10: 133351277-133351277 on Build GRCh38
rs777658930 Chr.10: 133351408-133351408 on Build GRCh38
rs748455974 Chr.10: 133351331-133351331 on Build GRCh38
rs747455124 Chr.10: 133351338-133351338 on Build GRCh38
rs751092007 Chr.10: 133351360-133351360 on Build GRCh38
rs758653852 Chr.10: 133351406-133351406 on Build GRCh38
rs571306661 Chr.10: 133351341-133351341 on Build GRCh38
rs943389105 Chr.10: 133351251-133351251 on Build GRCh38
rs549525566 Chr.10: 133351283-133351283 on Build GRCh38
rs1050431335 Chr.10: 133351232-133351232 on Build GRCh38
rs144079438 Chr.10: 133351235-133351235 on Build GRCh38
rs201333706 Chr.10: 133351419-133351419 on Build GRCh38
rs750396982 Chr.10: 133351386-133351386 on Build GRCh38
rs777247064 Chr.10: 133351434-133351434 on Build GRCh38
rs748281551 Chr.10: 133351405-133351405 on Build GRCh38
rs773374744 Chr.10: 133351334-133351334 on Build GRCh38
rs762453398 Chr.10: 133351444-133351444 on Build GRCh38
rs879115689 Chr.10: 133351263-133351263 on Build GRCh38
rs751586458 Chr.10: 133351321-133351321 on Build GRCh38
rs1042598892 Chr.10: 133351307-133351307 on Build GRCh38
rs561071829 Chr.10: 133351231-133351231 on Build GRCh38

View Full Product Details