Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196561292-196561517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR53
Entrez Gene ID
348793
Gene Name
WD repeat domain 53
Gene Aliases
-
UniGene
Hs.385865
Gene Chromosome Location
Chr.3: 196554188-196568542 on Build GRCh38
Transcripts
NM_182627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5355525 c.103G>T p.A35S Chr.3: 196561373-196561373 on Build GRCh38
2947398 c.138G>A p.T46T Chr.3: 196561338-196561338 on Build GRCh38
332922 c.124C>G p.P42A Chr.3: 196561352-196561352 on Build GRCh38
5398960 c.61G>A p.E21K Chr.3: 196561415-196561415 on Build GRCh38
4116409 c.35C>A p.P12H Chr.3: 196561441-196561441 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025272593 Chr.3: 196561324-196561324 on Build GRCh38
rs772480758 Chr.3: 196561350-196561350 on Build GRCh38
rs756604044 Chr.3: 196561336-196561336 on Build GRCh38
rs780187714 Chr.3: 196561485-196561485 on Build GRCh38
rs191731254 Chr.3: 196561327-196561327 on Build GRCh38
rs762568994 Chr.3: 196561411-196561411 on Build GRCh38
rs145241634 Chr.3: 196561449-196561449 on Build GRCh38
rs778419372 Chr.3: 196561337-196561337 on Build GRCh38
rs779524232 Chr.3: 196561477-196561477 on Build GRCh38
rs556124469 Chr.3: 196561458-196561458 on Build GRCh38
rs145559692 Chr.3: 196561386-196561386 on Build GRCh38
rs943286659 Chr.3: 196561420-196561420 on Build GRCh38
rs148755612 Chr.3: 196561459-196561459 on Build GRCh38
rs537400299 Chr.3: 196561410-196561410 on Build GRCh38
rs764760779 Chr.3: 196561394-196561394 on Build GRCh38
rs371685054 Chr.3: 196561367-196561367 on Build GRCh38
rs928374185 Chr.3: 196561385-196561385 on Build GRCh38
rs745923160 Chr.3: 196561478-196561478 on Build GRCh38
rs765930417 Chr.3: 196561412-196561412 on Build GRCh38
rs201680617 Chr.3: 196561372-196561372 on Build GRCh38
rs919938404 Chr.3: 196561465-196561465 on Build GRCh38
rs140055649 Chr.3: 196561375-196561375 on Build GRCh38
rs749815051 Chr.3: 196561338-196561338 on Build GRCh38
rs775840166 Chr.3: 196561374-196561374 on Build GRCh38
rs570101400 Chr.3: 196561335-196561335 on Build GRCh38
rs758573986 Chr.3: 196561484-196561484 on Build GRCh38
rs367598741 Chr.3: 196561353-196561353 on Build GRCh38
rs768449279 Chr.3: 196561377-196561377 on Build GRCh38
rs536161283 Chr.3: 196561494-196561494 on Build GRCh38
rs761763742 Chr.3: 196561390-196561390 on Build GRCh38
rs199721313 Chr.3: 196561354-196561354 on Build GRCh38
rs751322067 Chr.3: 196561442-196561442 on Build GRCh38
rs757303024 Chr.3: 196561355-196561355 on Build GRCh38
rs898756194 Chr.3: 196561475-196561475 on Build GRCh38
rs764564912 Chr.3: 196561452-196561452 on Build GRCh38
rs183093679 Chr.3: 196561486-196561486 on Build GRCh38
rs778892250 Chr.3: 196561365-196561365 on Build GRCh38
rs957705352 Chr.3: 196561342-196561342 on Build GRCh38

View Full Product Details