Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218402120-218402371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_011511569.2 XM_011511570.2 XM_017004615.1 XM_017004616.1 XM_017004617.1 XM_017004618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3577692 c.241C>T p.P81S Chr.2: 218402135-218402135 on Build GRCh38
7363196 c.342C>A p.F114L Chr.2: 218402369-218402369 on Build GRCh38
7267013 c.277G>A p.V93M Chr.2: 218402171-218402171 on Build GRCh38
4632870 c.233A>G p.Y78C Chr.2: 218402127-218402127 on Build GRCh38
5631296 c.328A>G p.N110D Chr.2: 218402355-218402355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748115993 Chr.2: 218402171-218402171 on Build GRCh38
rs138242622 Chr.2: 218402308-218402308 on Build GRCh38
rs554678491 Chr.2: 218402277-218402277 on Build GRCh38
rs199791299 Chr.2: 218402218-218402218 on Build GRCh38
rs760684567 Chr.2: 218402267-218402267 on Build GRCh38
rs774977260 Chr.2: 218402340-218402340 on Build GRCh38
rs200929399 Chr.2: 218402301-218402301 on Build GRCh38
rs375845906 Chr.2: 218402250-218402250 on Build GRCh38
rs750477899 Chr.2: 218402179-218402179 on Build GRCh38
rs755635864 Chr.2: 218402319-218402319 on Build GRCh38
rs764954295 Chr.2: 218402334-218402335 on Build GRCh38
rs745358670 Chr.2: 218402235-218402235 on Build GRCh38
rs376907785 Chr.2: 218402206-218402206 on Build GRCh38
rs756810983 Chr.2: 218402250-218402251 on Build GRCh38
rs937519476 Chr.2: 218402170-218402170 on Build GRCh38
rs367622179 Chr.2: 218402341-218402341 on Build GRCh38
rs773485560 Chr.2: 218402256-218402256 on Build GRCh38
rs758302524 Chr.2: 218402302-218402302 on Build GRCh38
rs948467115 Chr.2: 218402195-218402195 on Build GRCh38
rs781041746 Chr.2: 218402230-218402230 on Build GRCh38
rs769019081 Chr.2: 218402338-218402338 on Build GRCh38
rs975485291 Chr.2: 218402329-218402329 on Build GRCh38
rs547688629 Chr.2: 218402228-218402228 on Build GRCh38
rs759158940 Chr.2: 218402149-218402149 on Build GRCh38
rs745529377 Chr.2: 218402316-218402316 on Build GRCh38
rs779459596 Chr.2: 218402320-218402320 on Build GRCh38
rs746658218 Chr.2: 218402143-218402143 on Build GRCh38
rs758158061 Chr.2: 218402209-218402209 on Build GRCh38
rs766444859 Chr.2: 218402345-218402345 on Build GRCh38
rs895586157 Chr.2: 218402188-218402188 on Build GRCh38
rs766465594 Chr.2: 218402298-218402298 on Build GRCh38
rs199738523 Chr.2: 218402244-218402244 on Build GRCh38
rs370760593 Chr.2: 218402343-218402343 on Build GRCh38
rs751754736 Chr.2: 218402168-218402168 on Build GRCh38
rs1008614748 Chr.2: 218402265-218402265 on Build GRCh38
rs756241505 Chr.2: 218402190-218402190 on Build GRCh38
rs746635096 Chr.2: 218402222-218402222 on Build GRCh38
rs776313857 Chr.2: 218402148-218402148 on Build GRCh38
rs769496049 Chr.2: 218402236-218402236 on Build GRCh38
rs750118204 Chr.2: 218402339-218402339 on Build GRCh38
rs374939013 Chr.2: 218402157-218402157 on Build GRCh38
rs902184043 Chr.2: 218402264-218402264 on Build GRCh38
rs370798418 Chr.2: 218402342-218402342 on Build GRCh38
rs778814009 Chr.2: 218402194-218402194 on Build GRCh38
rs539181892 Chr.2: 218402147-218402147 on Build GRCh38
rs780988397 Chr.2: 218402313-218402313 on Build GRCh38
rs764379742 Chr.2: 218402161-218402161 on Build GRCh38
rs776007664 Chr.2: 218402243-218402243 on Build GRCh38
rs372400662 Chr.2: 218402303-218402303 on Build GRCh38
rs763310373 Chr.2: 218402159-218402159 on Build GRCh38
rs776544845 Chr.2: 218402351-218402351 on Build GRCh38
rs370091958 Chr.2: 218402251-218402251 on Build GRCh38
rs372797160 Chr.2: 218402330-218402330 on Build GRCh38
rs770402295 Chr.2: 218402151-218402151 on Build GRCh38

View Full Product Details