Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34404750-34405023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD2L
Entrez Gene ID
84306
Gene Name
programmed cell death 2 like
Gene Aliases
-
UniGene
Hs.515344
Gene Chromosome Location
Chr.19: 34404398-34426168 on Build GRCh38
Transcripts
NM_032346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4713007 c.291C>T p.R97R Chr.19: 34404945-34404945 on Build GRCh38
1612044 c.279G>A p.W93* Chr.19: 34404933-34404933 on Build GRCh38
212154 c.280A>G p.K94E Chr.19: 34404934-34404934 on Build GRCh38
6150550 c.293C>T p.S98F Chr.19: 34404947-34404947 on Build GRCh38
4407511 c.276-9A>C p.? Chr.19: 34404921-34404921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765526510 Chr.19: 34404907-34404907 on Build GRCh38
rs764709004 Chr.19: 34404904-34404904 on Build GRCh38
rs755300332 Chr.19: 34404975-34404975 on Build GRCh38
rs777111058 Chr.19: 34404788-34404799 on Build GRCh38
rs752265644 Chr.19: 34404906-34404906 on Build GRCh38
rs181216678 Chr.19: 34404984-34404984 on Build GRCh38
rs760361738 Chr.19: 34404800-34404800 on Build GRCh38
rs199618298 Chr.19: 34404861-34404861 on Build GRCh38
rs376443396 Chr.19: 34404804-34404804 on Build GRCh38
rs904971179 Chr.19: 34404990-34404990 on Build GRCh38
rs775855722 Chr.19: 34404942-34404942 on Build GRCh38
rs766010307 Chr.19: 34404895-34404895 on Build GRCh38
rs773617043 Chr.19: 34404986-34404986 on Build GRCh38
rs374501702 Chr.19: 34404844-34404844 on Build GRCh38
rs756295798 Chr.19: 34404842-34404842 on Build GRCh38
rs111818375 Chr.19: 34404847-34404847 on Build GRCh38
rs896828036 Chr.19: 34404845-34404845 on Build GRCh38
rs1007038684 Chr.19: 34404870-34404870 on Build GRCh38
rs759226991 Chr.19: 34404903-34404903 on Build GRCh38
rs776298242 Chr.19: 34404901-34404901 on Build GRCh38
rs763945883 Chr.19: 34404949-34404949 on Build GRCh38
rs746848700 Chr.19: 34404825-34404825 on Build GRCh38
rs757021656 Chr.19: 34404966-34404966 on Build GRCh38
rs747360925 Chr.19: 34404989-34404989 on Build GRCh38
rs754507022 Chr.19: 34404803-34404803 on Build GRCh38
rs769224441 Chr.19: 34404777-34404777 on Build GRCh38
rs758354370 Chr.19: 34404923-34404923 on Build GRCh38
rs752600661 Chr.19: 34404805-34404805 on Build GRCh38
rs767164204 Chr.19: 34404966-34404967 on Build GRCh38
rs375361877 Chr.19: 34404971-34404971 on Build GRCh38
rs941213455 Chr.19: 34404978-34404978 on Build GRCh38
rs775016998 Chr.19: 34404783-34404783 on Build GRCh38
rs761331362 Chr.19: 34404941-34404942 on Build GRCh38
rs968251555 Chr.19: 34404806-34404806 on Build GRCh38
rs760950524 Chr.19: 34404892-34404892 on Build GRCh38
rs758320892 Chr.19: 34404813-34404813 on Build GRCh38
rs978874100 Chr.19: 34404793-34404793 on Build GRCh38
rs771255044 Chr.19: 34404995-34404995 on Build GRCh38
rs767892516 Chr.19: 34404789-34404789 on Build GRCh38
rs374816557 Chr.19: 34404864-34404864 on Build GRCh38
rs114822664 Chr.19: 34404843-34404843 on Build GRCh38
rs202226528 Chr.19: 34404769-34404769 on Build GRCh38
rs753289642 Chr.19: 34404802-34404802 on Build GRCh38
rs201906686 Chr.19: 34404981-34404981 on Build GRCh38
rs73926774 Chr.19: 34404853-34404853 on Build GRCh38
rs749329025 Chr.19: 34404846-34404846 on Build GRCh38
rs372294731 Chr.19: 34404963-34404963 on Build GRCh38
rs200414433 Chr.19: 34404982-34404982 on Build GRCh38
rs201461270 Chr.19: 34404820-34404820 on Build GRCh38
rs916481401 Chr.19: 34404837-34404837 on Build GRCh38
rs372410686 Chr.19: 34404801-34404801 on Build GRCh38
rs750801666 Chr.19: 34404796-34404796 on Build GRCh38
rs780379373 Chr.19: 34404969-34404969 on Build GRCh38
rs367925417 Chr.19: 34404884-34404884 on Build GRCh38
rs760040299 Chr.19: 34404844-34404844 on Build GRCh38

View Full Product Details