Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141400472-141400697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141330685-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGA2
Entrez Gene ID
56113
Gene Name
protocadherin gamma subfamily A, 2
Gene Aliases
PCDH-GAMMA-A2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141338760-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGA3
Entrez Gene ID
56112
Gene Name
protocadherin gamma subfamily A, 3
Gene Aliases
PCDH-GAMMA-A3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141344034-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGA4
Entrez Gene ID
56111
Gene Name
protocadherin gamma subfamily A, 4
Gene Aliases
PCDH-GAMMA-A4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141355025-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGA5
Entrez Gene ID
56110
Gene Name
protocadherin gamma subfamily A, 5
Gene Aliases
CDH-GAMMA-A5, ME3, PCDH-GAMMA-A5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141364331-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGA6
Entrez Gene ID
56109
Gene Name
protocadherin gamma subfamily A, 6
Gene Aliases
PCDH-GAMMA-A6
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141374084-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGA7
Entrez Gene ID
56108
Gene Name
protocadherin gamma subfamily A, 7
Gene Aliases
PCDH-GAMMA-A7
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141382739-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGA8
Entrez Gene ID
9708
Gene Name
protocadherin gamma subfamily A, 8
Gene Aliases
PCDH-GAMMA-A8
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141391916-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141350261-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141360136-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141370264-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGB4
Entrez Gene ID
8641
Gene Name
protocadherin gamma subfamily B, 4
Gene Aliases
CDH20, FIB2, PCDH-GAMMA-B4
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141387885-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

Gene Symbol
PCDHGB5
Entrez Gene ID
56101
Gene Name
protocadherin gamma subfamily B, 5
Gene Aliases
PCDH-GAMMA-B5
UniGene
Hs.368160
Gene Chromosome Location
Chr.5: 141398128-141512979 on Build GRCh38
Transcripts
NM_018912.2 NM_018915.3 NM_018916.3 NM_018917.3 NM_018918.2 NM_018919.2 NM_018920.3 NM_032088.1 NM_018922.2 NM_018923.2 NM_018924.4 NM_003736.2 NM_018925.2 NM_032099.1

SNP Detail

SNP ID Chromosome Location
rs534226736 Chr.5: 141400547-141400547 on Build GRCh38
rs72790034 Chr.5: 141400656-141400656 on Build GRCh38
rs373097307 Chr.5: 141400542-141400542 on Build GRCh38
rs1025270322 Chr.5: 141400500-141400500 on Build GRCh38
rs373703769 Chr.5: 141400581-141400581 on Build GRCh38
rs748573702 Chr.5: 141400579-141400579 on Build GRCh38
rs753705723 Chr.5: 141400535-141400535 on Build GRCh38
rs994052728 Chr.5: 141400538-141400538 on Build GRCh38
rs958686867 Chr.5: 141400524-141400524 on Build GRCh38
rs753425146 Chr.5: 141400602-141400603 on Build GRCh38
rs781701893 Chr.5: 141400571-141400571 on Build GRCh38
rs765907405 Chr.5: 141400548-141400548 on Build GRCh38
rs746471054 Chr.5: 141400591-141400591 on Build GRCh38
rs376731583 Chr.5: 141400609-141400609 on Build GRCh38
rs532914635 Chr.5: 141400564-141400564 on Build GRCh38
rs745551734 Chr.5: 141400584-141400584 on Build GRCh38
rs963844263 Chr.5: 141400586-141400586 on Build GRCh38
rs367864769 Chr.5: 141400515-141400515 on Build GRCh38
rs977303120 Chr.5: 141400575-141400575 on Build GRCh38
rs754437274 Chr.5: 141400566-141400566 on Build GRCh38
rs775174347 Chr.5: 141400590-141400590 on Build GRCh38
rs555600694 Chr.5: 141400611-141400611 on Build GRCh38
rs551157559 Chr.5: 141400548-141400548 on Build GRCh38
rs771674964 Chr.5: 141400506-141400506 on Build GRCh38
rs771561940 Chr.5: 141400589-141400589 on Build GRCh38
rs756569671 Chr.5: 141400580-141400580 on Build GRCh38
rs759107285 Chr.5: 141400505-141400505 on Build GRCh38
rs766459739 Chr.5: 141400565-141400565 on Build GRCh38
rs376764580 Chr.5: 141400568-141400568 on Build GRCh38
rs182297545 Chr.5: 141400627-141400627 on Build GRCh38
rs768255831 Chr.5: 141400593-141400593 on Build GRCh38
rs775086396 Chr.5: 141400512-141400512 on Build GRCh38
rs764084750 Chr.5: 141400528-141400528 on Build GRCh38
rs574905149 Chr.5: 141400501-141400501 on Build GRCh38

View Full Product Details