Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13182500-13182731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHACTR1
Entrez Gene ID
221692
Gene Name
phosphatase and actin regulator 1
Gene Aliases
RPEL, RPEL1, dJ257A7.2
UniGene
Hs.436996
Gene Chromosome Location
Chr.6: 12716635-13290484 on Build GRCh38
Transcripts
NM_001242648.2 NM_001322308.1 NM_001322309.1 NM_001322310.1 NM_001322311.1 NM_001322312.1 NM_001322313.1 NM_001322314.1 NM_030948.3 XM_005248934.2 XM_017010452.1 XM_017010454.1 XM_017010455.1 XM_017010456.1 XM_017010457.1 XM_017010458.1 XM_017010459.1 XM_017010460.1 XM_017010461.1 XM_017010462.1 XM_017010463.1 XM_017010464.1 XM_017010465.1 XM_017010466.1 XM_017010467.1 XM_017010469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3920730 c.630G>A p.E210E Chr.6: 13182652-13182652 on Build GRCh38
740153 c.647A>T p.D216V Chr.6: 13182669-13182669 on Build GRCh38
1620904 c.168+10C>T p.? Chr.6: 13182696-13182696 on Build GRCh38
4554688 c.628G>A p.E210K Chr.6: 13182650-13182650 on Build GRCh38
3429998 c.736G>C p.E246Q Chr.6: 13182652-13182652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368581704 Chr.6: 13182705-13182705 on Build GRCh38
rs376467653 Chr.6: 13182526-13182526 on Build GRCh38
rs765090294 Chr.6: 13182697-13182697 on Build GRCh38
rs373983813 Chr.6: 13182655-13182655 on Build GRCh38
rs756458588 Chr.6: 13182645-13182645 on Build GRCh38
rs867058077 Chr.6: 13182599-13182599 on Build GRCh38
rs771578859 Chr.6: 13182564-13182564 on Build GRCh38
rs772556170 Chr.6: 13182567-13182567 on Build GRCh38
rs931491066 Chr.6: 13182687-13182687 on Build GRCh38
rs745812169 Chr.6: 13182693-13182693 on Build GRCh38
rs749820467 Chr.6: 13182537-13182537 on Build GRCh38
rs766071210 Chr.6: 13182590-13182590 on Build GRCh38
rs372653527 Chr.6: 13182707-13182707 on Build GRCh38
rs780926379 Chr.6: 13182684-13182685 on Build GRCh38
rs776404997 Chr.6: 13182681-13182681 on Build GRCh38
rs758310838 Chr.6: 13182612-13182612 on Build GRCh38
rs533330849 Chr.6: 13182609-13182609 on Build GRCh38
rs781326499 Chr.6: 13182619-13182619 on Build GRCh38
rs1000968652 Chr.6: 13182552-13182552 on Build GRCh38
rs373815868 Chr.6: 13182607-13182607 on Build GRCh38
rs115101968 Chr.6: 13182711-13182711 on Build GRCh38
rs746155299 Chr.6: 13182638-13182638 on Build GRCh38
rs749693216 Chr.6: 13182652-13182652 on Build GRCh38
rs370905078 Chr.6: 13182618-13182618 on Build GRCh38
rs143168559 Chr.6: 13182696-13182696 on Build GRCh38
rs991796097 Chr.6: 13182543-13182543 on Build GRCh38
rs889162542 Chr.6: 13182529-13182529 on Build GRCh38
rs376126977 Chr.6: 13182614-13182614 on Build GRCh38
rs1032482403 Chr.6: 13182569-13182569 on Build GRCh38
rs370949552 Chr.6: 13182600-13182600 on Build GRCh38
rs751548287 Chr.6: 13182617-13182617 on Build GRCh38
rs775110460 Chr.6: 13182698-13182698 on Build GRCh38
rs760303560 Chr.6: 13182675-13182675 on Build GRCh38
rs760255382 Chr.6: 13182577-13182577 on Build GRCh38
rs970962705 Chr.6: 13182677-13182677 on Build GRCh38
rs780332849 Chr.6: 13182648-13182648 on Build GRCh38
rs765179968 Chr.6: 13182608-13182608 on Build GRCh38
rs372411363 Chr.6: 13182664-13182664 on Build GRCh38
rs367868310 Chr.6: 13182663-13182663 on Build GRCh38

View Full Product Details