Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33761835-33762082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF341
Entrez Gene ID
84905
Gene Name
zinc finger protein 341
Gene Aliases
-
UniGene
Hs.516989
Gene Chromosome Location
Chr.20: 33731760-33792269 on Build GRCh38
Transcripts
NM_001282933.1 NM_001282935.1 NM_032819.4 NR_104259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6004567 c.1149C>T p.T383T Chr.20: 33761982-33761982 on Build GRCh38
7235495 c.1086G>C p.K362N Chr.20: 33761919-33761919 on Build GRCh38
3545501 c.1142C>T p.S381F Chr.20: 33761996-33761996 on Build GRCh38
5958235 c.1070G>C p.R357P Chr.20: 33761903-33761903 on Build GRCh38
3963494 c.1065G>C p.K355N Chr.20: 33761919-33761919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752743710 Chr.20: 33761901-33761901 on Build GRCh38
rs769528494 Chr.20: 33761942-33761942 on Build GRCh38
rs189857381 Chr.20: 33762016-33762016 on Build GRCh38
rs866641266 Chr.20: 33762026-33762026 on Build GRCh38
rs757168357 Chr.20: 33762030-33762030 on Build GRCh38
rs765475558 Chr.20: 33761869-33761869 on Build GRCh38
rs761189575 Chr.20: 33761985-33761985 on Build GRCh38
rs779837021 Chr.20: 33762047-33762047 on Build GRCh38
rs759934373 Chr.20: 33761993-33761993 on Build GRCh38
rs756167334 Chr.20: 33761931-33761931 on Build GRCh38
rs764166660 Chr.20: 33762009-33762009 on Build GRCh38
rs372933175 Chr.20: 33761943-33761943 on Build GRCh38
rs376521884 Chr.20: 33761911-33761911 on Build GRCh38
rs369875482 Chr.20: 33761925-33761925 on Build GRCh38
rs1055659546 Chr.20: 33762063-33762063 on Build GRCh38
rs6120367 Chr.20: 33762055-33762055 on Build GRCh38
rs376477111 Chr.20: 33761948-33761948 on Build GRCh38
rs192880441 Chr.20: 33761868-33761868 on Build GRCh38
rs750207536 Chr.20: 33762042-33762042 on Build GRCh38
rs758556957 Chr.20: 33761903-33761903 on Build GRCh38
rs558442228 Chr.20: 33761930-33761930 on Build GRCh38
rs758248799 Chr.20: 33762027-33762027 on Build GRCh38
rs6088297 Chr.20: 33762003-33762003 on Build GRCh38
rs765423752 Chr.20: 33761994-33761994 on Build GRCh38
rs151234184 Chr.20: 33761907-33761907 on Build GRCh38
rs745679012 Chr.20: 33761937-33761937 on Build GRCh38
rs766702364 Chr.20: 33761858-33761858 on Build GRCh38
rs982121798 Chr.20: 33761989-33761989 on Build GRCh38
rs771367918 Chr.20: 33761987-33761987 on Build GRCh38
rs752976862 Chr.20: 33761996-33761996 on Build GRCh38
rs760896105 Chr.20: 33761902-33761902 on Build GRCh38
rs780151851 Chr.20: 33761982-33761982 on Build GRCh38
rs6120366 Chr.20: 33762029-33762029 on Build GRCh38
rs767590915 Chr.20: 33762032-33762032 on Build GRCh38
rs184936741 Chr.20: 33761983-33761983 on Build GRCh38
rs1055782676 Chr.20: 33762007-33762007 on Build GRCh38
rs201202530 Chr.20: 33762043-33762043 on Build GRCh38
rs541415583 Chr.20: 33762019-33762019 on Build GRCh38
rs2747553 Chr.20: 33761990-33761990 on Build GRCh38

View Full Product Details