Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50342242-50342515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYND10
Entrez Gene ID
51364
Gene Name
zinc finger MYND-type containing 10
Gene Aliases
BLU, CILD22, FLU
UniGene
Hs.526735
Gene Chromosome Location
Chr.3: 50341106-50346028 on Build GRCh38
Transcripts
NM_001308379.1 NM_015896.3 XM_005265216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4963465 c.862C>A p.R288R Chr.3: 50342408-50342408 on Build GRCh38
6827601 c.764T>C p.L255P Chr.3: 50342506-50342506 on Build GRCh38
5471767 c.830C>T p.A277V Chr.3: 50342440-50342440 on Build GRCh38
4742943 c.778G>A p.A260T Chr.3: 50342477-50342477 on Build GRCh38
5994579 c.792C>T p.I264I Chr.3: 50342478-50342478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746513031 Chr.3: 50342364-50342364 on Build GRCh38
rs768120521 Chr.3: 50342453-50342453 on Build GRCh38
rs966939985 Chr.3: 50342297-50342297 on Build GRCh38
rs774727836 Chr.3: 50342385-50342385 on Build GRCh38
rs753798328 Chr.3: 50342432-50342432 on Build GRCh38
rs760826410 Chr.3: 50342418-50342418 on Build GRCh38
rs372975420 Chr.3: 50342438-50342438 on Build GRCh38
rs922773961 Chr.3: 50342332-50342332 on Build GRCh38
rs149071790 Chr.3: 50342476-50342476 on Build GRCh38
rs752891754 Chr.3: 50342411-50342411 on Build GRCh38
rs990796568 Chr.3: 50342477-50342477 on Build GRCh38
rs1009176270 Chr.3: 50342271-50342271 on Build GRCh38
rs771411301 Chr.3: 50342478-50342478 on Build GRCh38
rs767713937 Chr.3: 50342407-50342407 on Build GRCh38
rs955648602 Chr.3: 50342337-50342337 on Build GRCh38
rs202147494 Chr.3: 50342493-50342493 on Build GRCh38
rs374838122 Chr.3: 50342356-50342356 on Build GRCh38
rs985594994 Chr.3: 50342339-50342339 on Build GRCh38
rs368913303 Chr.3: 50342365-50342365 on Build GRCh38
rs746568217 Chr.3: 50342443-50342443 on Build GRCh38
rs779663116 Chr.3: 50342442-50342442 on Build GRCh38
rs181274355 Chr.3: 50342382-50342382 on Build GRCh38
rs772481555 Chr.3: 50342495-50342495 on Build GRCh38
rs927833122 Chr.3: 50342454-50342454 on Build GRCh38
rs755676243 Chr.3: 50342392-50342392 on Build GRCh38
rs771709845 Chr.3: 50342357-50342357 on Build GRCh38
rs937998049 Chr.3: 50342470-50342470 on Build GRCh38
rs587682566 Chr.3: 50342422-50342422 on Build GRCh38
rs771228949 Chr.3: 50342383-50342383 on Build GRCh38
rs185474627 Chr.3: 50342393-50342393 on Build GRCh38
rs773683727 Chr.3: 50342368-50342368 on Build GRCh38
rs778670018 Chr.3: 50342348-50342348 on Build GRCh38
rs1010606559 Chr.3: 50342480-50342480 on Build GRCh38
rs587594668 Chr.3: 50342439-50342439 on Build GRCh38
rs755072957 Chr.3: 50342263-50342263 on Build GRCh38
rs141234138 Chr.3: 50342331-50342331 on Build GRCh38
rs1022577948 Chr.3: 50342295-50342295 on Build GRCh38
rs774583071 Chr.3: 50342492-50342492 on Build GRCh38
rs200913791 Chr.3: 50342473-50342473 on Build GRCh38
rs746014252 Chr.3: 50342437-50342437 on Build GRCh38
rs989145292 Chr.3: 50342291-50342291 on Build GRCh38
rs941387815 Chr.3: 50342362-50342362 on Build GRCh38

View Full Product Details