Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32995634-32995859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPACA3
Entrez Gene ID
124912
Gene Name
sperm acrosome associated 3
Gene Aliases
ALLP17, CT54, LYC3, LYZC, LYZL3, SLLP1
UniGene
Hs.434112
Gene Chromosome Location
Chr.17: 32991864-32997877 on Build GRCh38
Transcripts
NM_001317225.1 NM_001317226.1 NM_173847.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3516018 c.317G>A p.G106E Chr.17: 32995691-32995691 on Build GRCh38
177115 c.275G>A p.R92H Chr.17: 32995649-32995649 on Build GRCh38
6146214 c.265C>T p.L89F Chr.17: 32995639-32995639 on Build GRCh38
5386045 c.316G>A p.G106R Chr.17: 32995690-32995690 on Build GRCh38
3516017 c.272G>A p.G91D Chr.17: 32995646-32995646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769214421 Chr.17: 32995688-32995688 on Build GRCh38
rs529990942 Chr.17: 32995756-32995756 on Build GRCh38
rs372148348 Chr.17: 32995741-32995741 on Build GRCh38
rs756949250 Chr.17: 32995660-32995660 on Build GRCh38
rs539810077 Chr.17: 32995810-32995810 on Build GRCh38
rs145497386 Chr.17: 32995715-32995715 on Build GRCh38
rs1040106541 Chr.17: 32995778-32995778 on Build GRCh38
rs201373267 Chr.17: 32995738-32995738 on Build GRCh38
rs762952782 Chr.17: 32995714-32995714 on Build GRCh38
rs773401025 Chr.17: 32995766-32995766 on Build GRCh38
rs200269504 Chr.17: 32995681-32995681 on Build GRCh38
rs745568120 Chr.17: 32995670-32995670 on Build GRCh38
rs1050168281 Chr.17: 32995803-32995803 on Build GRCh38
rs780760504 Chr.17: 32995733-32995733 on Build GRCh38
rs566759555 Chr.17: 32995814-32995814 on Build GRCh38
rs149695417 Chr.17: 32995691-32995691 on Build GRCh38
rs773265437 Chr.17: 32995697-32995697 on Build GRCh38
rs981322277 Chr.17: 32995703-32995703 on Build GRCh38
rs764929193 Chr.17: 32995698-32995698 on Build GRCh38
rs756790181 Chr.17: 32995724-32995724 on Build GRCh38
rs930388170 Chr.17: 32995786-32995788 on Build GRCh38
rs780661706 Chr.17: 32995661-32995661 on Build GRCh38
rs749983803 Chr.17: 32995736-32995736 on Build GRCh38
rs909093536 Chr.17: 32995765-32995765 on Build GRCh38
rs371858703 Chr.17: 32995758-32995758 on Build GRCh38
rs1010627364 Chr.17: 32995668-32995668 on Build GRCh38
rs919105235 Chr.17: 32995711-32995711 on Build GRCh38
rs147867748 Chr.17: 32995739-32995739 on Build GRCh38
rs1051161843 Chr.17: 32995825-32995825 on Build GRCh38
rs751573579 Chr.17: 32995720-32995720 on Build GRCh38
rs775934021 Chr.17: 32995680-32995680 on Build GRCh38
rs760803624 Chr.17: 32995767-32995767 on Build GRCh38
rs868612830 Chr.17: 32995700-32995700 on Build GRCh38
rs12450506 Chr.17: 32995744-32995744 on Build GRCh38
rs28963 Chr.17: 32995673-32995673 on Build GRCh38
rs530643498 Chr.17: 32995696-32995696 on Build GRCh38
rs752604604 Chr.17: 32995704-32995704 on Build GRCh38
rs199736765 Chr.17: 32995689-32995689 on Build GRCh38
rs991529280 Chr.17: 32995665-32995665 on Build GRCh38
rs201729342 Chr.17: 32995658-32995658 on Build GRCh38
rs748775401 Chr.17: 32995832-32995832 on Build GRCh38
rs752861274 Chr.17: 32995760-32995771 on Build GRCh38
rs920298055 Chr.17: 32995777-32995777 on Build GRCh38
rs761929137 Chr.17: 32995690-32995690 on Build GRCh38

View Full Product Details