Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30647352-30647597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf136
Entrez Gene ID
221545
Gene Name
chromosome 6 open reading frame 136
Gene Aliases
-
UniGene
Hs.591787
Gene Chromosome Location
Chr.6: 30647039-30653210 on Build GRCh38
Transcripts
NM_001109938.2 NM_001161376.1 NM_145029.3 XM_011514384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5967933 c.190C>A p.L64I Chr.6: 30647421-30647421 on Build GRCh38
4830655 c.239C>A p.A80E Chr.6: 30647470-30647470 on Build GRCh38
6397669 c.254G>A p.C85Y Chr.6: 30647485-30647485 on Build GRCh38
6387542 c.268G>A p.A90T Chr.6: 30647499-30647499 on Build GRCh38
6500389 c.342T>G p.V114V Chr.6: 30647573-30647573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051593990 Chr.6: 30647399-30647399 on Build GRCh38
rs925765586 Chr.6: 30647453-30647453 on Build GRCh38
rs563563696 Chr.6: 30647567-30647567 on Build GRCh38
rs775316855 Chr.6: 30647417-30647417 on Build GRCh38
rs899121199 Chr.6: 30647413-30647414 on Build GRCh38
rs773509215 Chr.6: 30647520-30647520 on Build GRCh38
rs28360037 Chr.6: 30647467-30647467 on Build GRCh38
rs747516318 Chr.6: 30647374-30647374 on Build GRCh38
rs1015016209 Chr.6: 30647407-30647407 on Build GRCh38
rs764140056 Chr.6: 30647421-30647421 on Build GRCh38
rs769601702 Chr.6: 30647416-30647416 on Build GRCh38
rs933129123 Chr.6: 30647397-30647397 on Build GRCh38
rs747264656 Chr.6: 30647521-30647521 on Build GRCh38
rs748320980 Chr.6: 30647375-30647375 on Build GRCh38
rs975077908 Chr.6: 30647480-30647480 on Build GRCh38
rs762622744 Chr.6: 30647420-30647420 on Build GRCh38
rs774701606 Chr.6: 30647371-30647371 on Build GRCh38
rs926043544 Chr.6: 30647508-30647508 on Build GRCh38
rs888985195 Chr.6: 30647418-30647418 on Build GRCh38
rs539750251 Chr.6: 30647380-30647380 on Build GRCh38
rs995231343 Chr.6: 30647532-30647532 on Build GRCh38
rs894495085 Chr.6: 30647565-30647565 on Build GRCh38
rs116816200 Chr.6: 30647507-30647507 on Build GRCh38
rs866892600 Chr.6: 30647541-30647541 on Build GRCh38
rs747822653 Chr.6: 30647456-30647457 on Build GRCh38
rs555516210 Chr.6: 30647433-30647433 on Build GRCh38
rs535688451 Chr.6: 30647429-30647429 on Build GRCh38
rs1051979589 Chr.6: 30647497-30647497 on Build GRCh38
rs771514519 Chr.6: 30647559-30647560 on Build GRCh38
rs372110822 Chr.6: 30647393-30647393 on Build GRCh38
rs115572115 Chr.6: 30647436-30647436 on Build GRCh38
rs1034372439 Chr.6: 30647538-30647538 on Build GRCh38
rs1026530312 Chr.6: 30647465-30647465 on Build GRCh38
rs755287603 Chr.6: 30647445-30647445 on Build GRCh38
rs899279402 Chr.6: 30647531-30647531 on Build GRCh38
rs550423101 Chr.6: 30647381-30647381 on Build GRCh38
rs755776732 Chr.6: 30647441-30647441 on Build GRCh38
rs762903321 Chr.6: 30647555-30647555 on Build GRCh38
rs932212946 Chr.6: 30647488-30647488 on Build GRCh38
rs754282301 Chr.6: 30647442-30647442 on Build GRCh38
rs970224908 Chr.6: 30647492-30647492 on Build GRCh38
rs977537787 Chr.6: 30647449-30647449 on Build GRCh38
rs61606469 Chr.6: 30647485-30647485 on Build GRCh38
rs3132594 Chr.6: 30647483-30647483 on Build GRCh38
rs186287743 Chr.6: 30647580-30647580 on Build GRCh38
rs890590022 Chr.6: 30647506-30647506 on Build GRCh38

View Full Product Details