Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 2977837-2978110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFY
Entrez Gene ID
7544
Gene Name
zinc finger protein, Y-linked
Gene Aliases
ZNF911
UniGene
Hs.522845
Gene Chromosome Location
Chr.Y: 2934402-2982508 on Build GRCh38
Transcripts
NM_001145275.1 NM_001145276.1 NM_003411.3 XM_005262570.2 XM_011531473.2 XM_011531474.2 XM_011531475.2 XM_017030075.1 XM_017030076.1 XM_017030077.1

SNP Detail

SNP ID Chromosome Location
rs779744913 Chr.Y: 2977998-2977998 on Build GRCh38
rs748871040 Chr.Y: 2978040-2978040 on Build GRCh38
rs113476308 Chr.Y: 2977936-2977936 on Build GRCh38
rs755778894 Chr.Y: 2977987-2977987 on Build GRCh38
rs377204801 Chr.Y: 2977967-2977967 on Build GRCh38
rs889926716 Chr.Y: 2977968-2977968 on Build GRCh38
rs763979419 Chr.Y: 2977904-2977906 on Build GRCh38
rs918815724 Chr.Y: 2977916-2977916 on Build GRCh38
rs377413046 Chr.Y: 2977976-2977976 on Build GRCh38
rs774407226 Chr.Y: 2978082-2978082 on Build GRCh38
rs764701101 Chr.Y: 2977905-2977905 on Build GRCh38
rs745529739 Chr.Y: 2977984-2977984 on Build GRCh38
rs931608329 Chr.Y: 2977920-2977920 on Build GRCh38
rs781686736 Chr.Y: 2977980-2977980 on Build GRCh38
rs199819030 Chr.Y: 2978053-2978053 on Build GRCh38
rs758858369 Chr.Y: 2977892-2977892 on Build GRCh38

View Full Product Details