Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23364726-23364979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GZF1
Entrez Gene ID
64412
Gene Name
GDNF inducible zinc finger protein 1
Gene Aliases
ZBTB23, ZNF336
UniGene
Hs.709462
Gene Chromosome Location
Chr.20: 23361585-23373062 on Build GRCh38
Transcripts
NM_001317012.1 NM_001317019.1 NM_022482.4 XM_005260800.2 XM_005260802.3 XM_011529321.2 XM_011529322.1 XM_017028017.1 XM_017028018.1 XM_017028019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1209239 c.508G>A p.D170N Chr.20: 23364891-23364891 on Build GRCh38
7260294 c.359A>T p.D120V Chr.20: 23364742-23364742 on Build GRCh38
1025199 c.429C>A p.F143L Chr.20: 23364812-23364812 on Build GRCh38
4097139 c.572G>A p.G191D Chr.20: 23364955-23364955 on Build GRCh38
723356 c.365C>G p.S122* Chr.20: 23364748-23364748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374194682 Chr.20: 23364939-23364939 on Build GRCh38
rs760991420 Chr.20: 23364922-23364922 on Build GRCh38
rs767598898 Chr.20: 23364916-23364916 on Build GRCh38
rs375809914 Chr.20: 23364907-23364907 on Build GRCh38
rs755810794 Chr.20: 23364838-23364838 on Build GRCh38
rs767723250 Chr.20: 23364834-23364834 on Build GRCh38
rs181976274 Chr.20: 23364871-23364871 on Build GRCh38
rs761939271 Chr.20: 23364833-23364833 on Build GRCh38
rs368817546 Chr.20: 23364890-23364890 on Build GRCh38
rs774382390 Chr.20: 23364829-23364829 on Build GRCh38
rs753682536 Chr.20: 23364856-23364856 on Build GRCh38
rs141727593 Chr.20: 23364917-23364917 on Build GRCh38
rs896575394 Chr.20: 23364897-23364897 on Build GRCh38
rs946967452 Chr.20: 23364825-23364825 on Build GRCh38
rs754917634 Chr.20: 23364868-23364868 on Build GRCh38
rs1050099914 Chr.20: 23364814-23364815 on Build GRCh38
rs372556804 Chr.20: 23364891-23364891 on Build GRCh38
rs759458320 Chr.20: 23364941-23364941 on Build GRCh38
rs548202788 Chr.20: 23364796-23364796 on Build GRCh38
rs777363362 Chr.20: 23364954-23364954 on Build GRCh38
rs1047740895 Chr.20: 23364932-23364932 on Build GRCh38
rs766222115 Chr.20: 23364852-23364852 on Build GRCh38
rs369155120 Chr.20: 23364915-23364915 on Build GRCh38
rs764118632 Chr.20: 23364822-23364822 on Build GRCh38
rs866817750 Chr.20: 23364789-23364789 on Build GRCh38
rs189316929 Chr.20: 23364760-23364760 on Build GRCh38
rs376790566 Chr.20: 23364884-23364884 on Build GRCh38
rs546744247 Chr.20: 23364943-23364943 on Build GRCh38
rs988499404 Chr.20: 23364765-23364765 on Build GRCh38
rs758857169 Chr.20: 23364871-23364872 on Build GRCh38
rs150596910 Chr.20: 23364931-23364931 on Build GRCh38
rs868043734 Chr.20: 23364946-23364946 on Build GRCh38
rs770197032 Chr.20: 23364776-23364776 on Build GRCh38
rs773053982 Chr.20: 23364757-23364757 on Build GRCh38
rs778302954 Chr.20: 23364949-23364954 on Build GRCh38
rs3810574 Chr.20: 23364952-23364952 on Build GRCh38
rs774906859 Chr.20: 23364909-23364909 on Build GRCh38
rs887749407 Chr.20: 23364942-23364942 on Build GRCh38
rs139704196 Chr.20: 23364874-23364874 on Build GRCh38
rs746210854 Chr.20: 23364768-23364768 on Build GRCh38
rs747644259 Chr.20: 23364872-23364872 on Build GRCh38
rs752683173 Chr.20: 23364949-23364949 on Build GRCh38
rs756084011 Chr.20: 23364895-23364895 on Build GRCh38
rs766552834 Chr.20: 23364923-23364923 on Build GRCh38
rs753648965 Chr.20: 23364927-23364927 on Build GRCh38
rs748548388 Chr.20: 23364767-23364770 on Build GRCh38
rs777498440 Chr.20: 23364881-23364881 on Build GRCh38
rs747979077 Chr.20: 23364806-23364806 on Build GRCh38
rs763537182 Chr.20: 23364818-23364818 on Build GRCh38

View Full Product Details