Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128820704-128820953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENDOG
Entrez Gene ID
2021
Gene Name
endonuclease G
Gene Aliases
-
UniGene
Hs.745161
Gene Chromosome Location
Chr.9: 128818402-128822676 on Build GRCh38
Transcripts
NM_004435.2 XM_011518347.2 NM_016390.3 XM_017014804.1

Gene Symbol
SPOUT1
Entrez Gene ID
51490
Gene Name
SPOUT domain containing methyltransferase 1
Gene Aliases
C9orf114, CENP-32, HSPC109
UniGene
Hs.224137
Gene Chromosome Location
Chr.9: 128819651-128833309 on Build GRCh38
Transcripts
NM_004435.2 XM_011518347.2 NM_016390.3 XM_017014804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4007066 c.555C>A p.S185R Chr.9: 128820792-128820792 on Build GRCh38
5643623 c.555C>T p.S185S Chr.9: 128820792-128820792 on Build GRCh38
1460335 c.551G>A p.R184H Chr.9: 128820788-128820788 on Build GRCh38
5794401 c.513C>T p.L171L Chr.9: 128820750-128820750 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs546113970 Chr.9: 128820928-128820928 on Build GRCh38
rs771057670 Chr.9: 128820842-128820842 on Build GRCh38
rs888204657 Chr.9: 128820726-128820726 on Build GRCh38
rs749266301 Chr.9: 128820809-128820809 on Build GRCh38
rs993096822 Chr.9: 128820729-128820729 on Build GRCh38
rs192190401 Chr.9: 128820892-128820892 on Build GRCh38
rs936701993 Chr.9: 128820887-128820887 on Build GRCh38
rs952015394 Chr.9: 128820753-128820753 on Build GRCh38
rs114639346 Chr.9: 128820792-128820792 on Build GRCh38
rs944199141 Chr.9: 128820905-128820905 on Build GRCh38
rs1040084678 Chr.9: 128820723-128820723 on Build GRCh38
rs547914635 Chr.9: 128820787-128820787 on Build GRCh38
rs539285547 Chr.9: 128820838-128820838 on Build GRCh38
rs535536814 Chr.9: 128820738-128820738 on Build GRCh38
rs375007966 Chr.9: 128820870-128820870 on Build GRCh38
rs2280845 Chr.9: 128820891-128820891 on Build GRCh38
rs758275298 Chr.9: 128820881-128820881 on Build GRCh38
rs759763346 Chr.9: 128820783-128820783 on Build GRCh38
rs966797376 Chr.9: 128820819-128820819 on Build GRCh38
rs372072772 Chr.9: 128820799-128820799 on Build GRCh38
rs532717496 Chr.9: 128820922-128820922 on Build GRCh38
rs145739062 Chr.9: 128820788-128820788 on Build GRCh38
rs779795207 Chr.9: 128820802-128820802 on Build GRCh38
rs755527226 Chr.9: 128820866-128820866 on Build GRCh38
rs201201300 Chr.9: 128820862-128820862 on Build GRCh38
rs773548246 Chr.9: 128820826-128820826 on Build GRCh38
rs762157621 Chr.9: 128820727-128820727 on Build GRCh38
rs748004074 Chr.9: 128820818-128820818 on Build GRCh38
rs757590731 Chr.9: 128820918-128820918 on Build GRCh38
rs747950682 Chr.9: 128820878-128820878 on Build GRCh38
rs189147687 Chr.9: 128820865-128820865 on Build GRCh38
rs776955894 Chr.9: 128820848-128820848 on Build GRCh38
rs1041211773 Chr.9: 128820914-128820914 on Build GRCh38
rs753325227 Chr.9: 128820868-128820868 on Build GRCh38
rs375132327 Chr.9: 128820804-128820804 on Build GRCh38
rs768580470 Chr.9: 128820817-128820818 on Build GRCh38
rs777801827 Chr.9: 128820883-128820883 on Build GRCh38
rs773183767 Chr.9: 128820813-128820813 on Build GRCh38
rs556705072 Chr.9: 128820814-128820814 on Build GRCh38
rs763369147 Chr.9: 128820864-128820864 on Build GRCh38
rs563842895 Chr.9: 128820916-128820916 on Build GRCh38
rs746001081 Chr.9: 128820900-128820900 on Build GRCh38
rs766793888 Chr.9: 128820765-128820765 on Build GRCh38
rs760067135 Chr.9: 128820849-128820849 on Build GRCh38
rs528071859 Chr.9: 128820773-128820773 on Build GRCh38
rs41275918 Chr.9: 128820851-128820851 on Build GRCh38
rs755651224 Chr.9: 128820800-128820800 on Build GRCh38
rs749202156 Chr.9: 128820764-128820764 on Build GRCh38
rs765941137 Chr.9: 128820867-128820867 on Build GRCh38
rs767210119 Chr.9: 128820796-128820796 on Build GRCh38
rs754942992 Chr.9: 128820871-128820871 on Build GRCh38
rs116714293 Chr.9: 128820820-128820820 on Build GRCh38
rs376004424 Chr.9: 128820731-128820731 on Build GRCh38
rs779050708 Chr.9: 128820873-128820873 on Build GRCh38
rs565666069 Chr.9: 128820725-128820725 on Build GRCh38

View Full Product Details