Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136846937-136847164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf172
Entrez Gene ID
389813
Gene Name
chromosome 9 open reading frame 172
Gene Aliases
-
UniGene
Hs.495485
Gene Chromosome Location
Chr.9: 136844415-136847345 on Build GRCh38
Transcripts
NM_001080482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6218919 c.2665G>T p.E889* Chr.9: 136847079-136847079 on Build GRCh38
3395679 c.2560G>A p.A854T Chr.9: 136846974-136846974 on Build GRCh38
5497176 c.2720G>T p.R907L Chr.9: 136847134-136847134 on Build GRCh38
3675292 c.2664C>A p.G888G Chr.9: 136847078-136847078 on Build GRCh38
7400373 c.2605G>T p.D869Y Chr.9: 136847019-136847019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867760870 Chr.9: 136847098-136847098 on Build GRCh38
rs1013058186 Chr.9: 136846961-136846961 on Build GRCh38
rs1030666710 Chr.9: 136847054-136847054 on Build GRCh38
rs768781018 Chr.9: 136847132-136847132 on Build GRCh38
rs1022008966 Chr.9: 136847073-136847073 on Build GRCh38
rs746839398 Chr.9: 136847051-136847051 on Build GRCh38
rs964145649 Chr.9: 136847084-136847084 on Build GRCh38
rs749364488 Chr.9: 136847117-136847117 on Build GRCh38
rs766028366 Chr.9: 136847089-136847090 on Build GRCh38
rs374792288 Chr.9: 136847139-136847139 on Build GRCh38
rs922461536 Chr.9: 136847143-136847143 on Build GRCh38
rs770750717 Chr.9: 136847088-136847088 on Build GRCh38
rs999138824 Chr.9: 136847049-136847049 on Build GRCh38
rs956401505 Chr.9: 136847057-136847057 on Build GRCh38
rs760383017 Chr.9: 136847070-136847082 on Build GRCh38
rs907436648 Chr.9: 136847018-136847018 on Build GRCh38
rs777387423 Chr.9: 136847016-136847016 on Build GRCh38
rs776670031 Chr.9: 136847096-136847096 on Build GRCh38
rs1024913245 Chr.9: 136846999-136846999 on Build GRCh38
rs761984563 Chr.9: 136847142-136847142 on Build GRCh38

View Full Product Details