Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33799064-33799337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLN
Entrez Gene ID
4295
Gene Name
motilin
Gene Aliases
-
UniGene
Hs.2813
Gene Chromosome Location
Chr.6: 33794672-33804016 on Build GRCh38
Transcripts
NM_001040109.1 NM_001184698.1 NM_002418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7150703 c.211G>A p.E71K Chr.6: 33799128-33799128 on Build GRCh38
1078176 c.199G>A p.E67K Chr.6: 33799140-33799140 on Build GRCh38
6713506 c.152T>C p.L51P Chr.6: 33799187-33799187 on Build GRCh38
1697022 c.173G>A p.G58E Chr.6: 33799166-33799166 on Build GRCh38
5568473 c.150C>T p.S50S Chr.6: 33799189-33799189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188344611 Chr.6: 33799189-33799189 on Build GRCh38
rs760576772 Chr.6: 33799133-33799133 on Build GRCh38
rs747262986 Chr.6: 33799212-33799212 on Build GRCh38
rs974692071 Chr.6: 33799246-33799246 on Build GRCh38
rs933604639 Chr.6: 33799269-33799269 on Build GRCh38
rs770255520 Chr.6: 33799222-33799222 on Build GRCh38
rs753837143 Chr.6: 33799273-33799273 on Build GRCh38
rs749913537 Chr.6: 33799256-33799256 on Build GRCh38
rs778626354 Chr.6: 33799138-33799138 on Build GRCh38
rs199860474 Chr.6: 33799216-33799216 on Build GRCh38
rs780783209 Chr.6: 33799172-33799172 on Build GRCh38
rs528168479 Chr.6: 33799137-33799137 on Build GRCh38
rs368261744 Chr.6: 33799116-33799116 on Build GRCh38
rs745984051 Chr.6: 33799220-33799220 on Build GRCh38
rs79561907 Chr.6: 33799186-33799186 on Build GRCh38
rs886647773 Chr.6: 33799281-33799281 on Build GRCh38
rs776045835 Chr.6: 33799228-33799228 on Build GRCh38
rs776799196 Chr.6: 33799219-33799219 on Build GRCh38
rs753038378 Chr.6: 33799136-33799136 on Build GRCh38
rs745629444 Chr.6: 33799316-33799316 on Build GRCh38
rs150413005 Chr.6: 33799117-33799117 on Build GRCh38
rs756653724 Chr.6: 33799152-33799152 on Build GRCh38
rs750279668 Chr.6: 33799179-33799179 on Build GRCh38
rs763778072 Chr.6: 33799183-33799183 on Build GRCh38
rs760655213 Chr.6: 33799115-33799115 on Build GRCh38
rs768627229 Chr.6: 33799210-33799210 on Build GRCh38
rs369819518 Chr.6: 33799252-33799252 on Build GRCh38
rs760124773 Chr.6: 33799129-33799129 on Build GRCh38
rs377738233 Chr.6: 33799141-33799141 on Build GRCh38
rs764445650 Chr.6: 33799243-33799243 on Build GRCh38
rs534729096 Chr.6: 33799265-33799265 on Build GRCh38
rs142640840 Chr.6: 33799211-33799211 on Build GRCh38
rs772388459 Chr.6: 33799111-33799111 on Build GRCh38
rs767124087 Chr.6: 33799253-33799253 on Build GRCh38
rs751285877 Chr.6: 33799142-33799142 on Build GRCh38
rs763292177 Chr.6: 33799240-33799240 on Build GRCh38
rs992004598 Chr.6: 33799089-33799089 on Build GRCh38
rs780202334 Chr.6: 33799270-33799270 on Build GRCh38
rs942205350 Chr.6: 33799310-33799310 on Build GRCh38

View Full Product Details