Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130699224-130699497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC2
Entrez Gene ID
23404
Gene Name
exosome component 2
Gene Aliases
RRP4, Rrp4p, hRrp4p, p7
UniGene
Hs.654643
Gene Chromosome Location
Chr.9: 130693760-130704894 on Build GRCh38
Transcripts
NM_001282708.1 NM_001282709.1 NM_014285.6 NR_104230.1 XM_005272176.2 XM_006717023.2 XM_017014558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366776 c.384G>T p.E128D Chr.9: 130699352-130699352 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773224500 Chr.9: 130699439-130699439 on Build GRCh38
rs376407286 Chr.9: 130699459-130699459 on Build GRCh38
rs768699876 Chr.9: 130699262-130699262 on Build GRCh38
rs760806358 Chr.9: 130699441-130699441 on Build GRCh38
rs1031920226 Chr.9: 130699420-130699420 on Build GRCh38
rs149268432 Chr.9: 130699350-130699350 on Build GRCh38
rs894884681 Chr.9: 130699258-130699258 on Build GRCh38
rs767528539 Chr.9: 130699281-130699281 on Build GRCh38
rs780578336 Chr.9: 130699396-130699396 on Build GRCh38
rs768158074 Chr.9: 130699427-130699428 on Build GRCh38
rs1017642500 Chr.9: 130699474-130699474 on Build GRCh38
rs369854065 Chr.9: 130699403-130699403 on Build GRCh38
rs745368024 Chr.9: 130699399-130699399 on Build GRCh38
rs562874644 Chr.9: 130699422-130699422 on Build GRCh38
rs748326368 Chr.9: 130699412-130699412 on Build GRCh38
rs756865286 Chr.9: 130699383-130699383 on Build GRCh38
rs779816695 Chr.9: 130699311-130699311 on Build GRCh38
rs766898401 Chr.9: 130699314-130699314 on Build GRCh38
rs990137658 Chr.9: 130699458-130699458 on Build GRCh38
rs373191394 Chr.9: 130699442-130699442 on Build GRCh38
rs768598729 Chr.9: 130699405-130699405 on Build GRCh38
rs113416483 Chr.9: 130699382-130699382 on Build GRCh38
rs750873098 Chr.9: 130699410-130699410 on Build GRCh38
rs200061464 Chr.9: 130699304-130699304 on Build GRCh38
rs754267970 Chr.9: 130699318-130699318 on Build GRCh38
rs774252290 Chr.9: 130699411-130699411 on Build GRCh38
rs762081942 Chr.9: 130699280-130699280 on Build GRCh38
rs549126631 Chr.9: 130699309-130699309 on Build GRCh38
rs774696225 Chr.9: 130699278-130699278 on Build GRCh38
rs771515440 Chr.9: 130699419-130699420 on Build GRCh38
rs751242634 Chr.9: 130699363-130699363 on Build GRCh38
rs773278906 Chr.9: 130699307-130699307 on Build GRCh38
rs780105414 Chr.9: 130699404-130699404 on Build GRCh38
rs778566059 Chr.9: 130699329-130699329 on Build GRCh38
rs560974430 Chr.9: 130699256-130699256 on Build GRCh38
rs759002672 Chr.9: 130699475-130699475 on Build GRCh38
rs755305419 Chr.9: 130699342-130699342 on Build GRCh38

View Full Product Details