Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52032889-52033118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KTI12
Entrez Gene ID
112970
Gene Name
KTI12 chromatin associated homolog
Gene Aliases
SBBI81, TOT4
UniGene
Hs.476033
Gene Chromosome Location
Chr.1: 52032105-52033800 on Build GRCh38
Transcripts
NM_138417.2 NM_015913.3 NR_046405.1 NR_046406.1

Gene Symbol
TXNDC12
Entrez Gene ID
51060
Gene Name
thioredoxin domain containing 12
Gene Aliases
AG1, AGR1, ERP16, ERP18, ERP19, PDIA16, TLP19, hAG-1, hTLP19
UniGene
Hs.476033
Gene Chromosome Location
Chr.1: 52020131-52056171 on Build GRCh38
Transcripts
NM_138417.2 NM_015913.3 NR_046405.1 NR_046406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5938697 c.719C>T p.P240L Chr.1: 52033043-52033043 on Build GRCh38
119887 c.862G>T p.A288S Chr.1: 52032900-52032900 on Build GRCh38
5846943 c.697G>A p.V233M Chr.1: 52033065-52033065 on Build GRCh38
354130 c.664C>A p.R222R Chr.1: 52033098-52033098 on Build GRCh38
4697098 c.779C>T p.T260M Chr.1: 52032983-52032983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781474674 Chr.1: 52032919-52032919 on Build GRCh38
rs548825981 Chr.1: 52032924-52032924 on Build GRCh38
rs776174698 Chr.1: 52032973-52032973 on Build GRCh38
rs752499921 Chr.1: 52032911-52032911 on Build GRCh38
rs199935747 Chr.1: 52032995-52032995 on Build GRCh38
rs762348795 Chr.1: 52032956-52032956 on Build GRCh38
rs79423354 Chr.1: 52033049-52033049 on Build GRCh38
rs759082165 Chr.1: 52032975-52032975 on Build GRCh38
rs749629638 Chr.1: 52032941-52032941 on Build GRCh38
rs138411665 Chr.1: 52033061-52033061 on Build GRCh38
rs757324544 Chr.1: 52032986-52032986 on Build GRCh38
rs371352386 Chr.1: 52033076-52033076 on Build GRCh38
rs3170843 Chr.1: 52032984-52032984 on Build GRCh38
rs762703904 Chr.1: 52033042-52033042 on Build GRCh38
rs148367224 Chr.1: 52033027-52033027 on Build GRCh38
rs747505851 Chr.1: 52033090-52033090 on Build GRCh38
rs574449882 Chr.1: 52033081-52033081 on Build GRCh38
rs756161230 Chr.1: 52033005-52033005 on Build GRCh38
rs764800535 Chr.1: 52032976-52032976 on Build GRCh38
rs763828945 Chr.1: 52033045-52033045 on Build GRCh38
rs968235383 Chr.1: 52033094-52033094 on Build GRCh38
rs768181978 Chr.1: 52032942-52032942 on Build GRCh38
rs372433236 Chr.1: 52033014-52033014 on Build GRCh38
rs375793609 Chr.1: 52032982-52032982 on Build GRCh38
rs779297184 Chr.1: 52033020-52033020 on Build GRCh38
rs750477587 Chr.1: 52033001-52033001 on Build GRCh38
rs374491748 Chr.1: 52033082-52033082 on Build GRCh38
rs749774837 Chr.1: 52033062-52033062 on Build GRCh38
rs772543782 Chr.1: 52032961-52032961 on Build GRCh38
rs748497468 Chr.1: 52033021-52033021 on Build GRCh38
rs777862178 Chr.1: 52032914-52032914 on Build GRCh38
rs202157272 Chr.1: 52033056-52033056 on Build GRCh38
rs556254591 Chr.1: 52033067-52033067 on Build GRCh38
rs749487996 Chr.1: 52033007-52033007 on Build GRCh38
rs145860172 Chr.1: 52032918-52032918 on Build GRCh38
rs995431199 Chr.1: 52032933-52032933 on Build GRCh38
rs753840698 Chr.1: 52033065-52033065 on Build GRCh38
rs763939293 Chr.1: 52032981-52032981 on Build GRCh38
rs758141732 Chr.1: 52032913-52032913 on Build GRCh38
rs374476013 Chr.1: 52032951-52032951 on Build GRCh38
rs534547542 Chr.1: 52033033-52033033 on Build GRCh38
rs368155864 Chr.1: 52032909-52032909 on Build GRCh38
rs768731017 Chr.1: 52033008-52033008 on Build GRCh38
rs775034676 Chr.1: 52033037-52033037 on Build GRCh38
rs1019435829 Chr.1: 52033029-52033029 on Build GRCh38
rs773641016 Chr.1: 52033025-52033025 on Build GRCh38
rs774872195 Chr.1: 52032943-52032943 on Build GRCh38
rs752332612 Chr.1: 52032979-52032979 on Build GRCh38
rs1051216606 Chr.1: 52033010-52033010 on Build GRCh38
rs1038812747 Chr.1: 52032936-52032936 on Build GRCh38
rs148811457 Chr.1: 52032929-52032929 on Build GRCh38
rs772633658 Chr.1: 52033023-52033023 on Build GRCh38
rs750326461 Chr.1: 52033050-52033050 on Build GRCh38

View Full Product Details