Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 175463290-175463515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAALADL2
Entrez Gene ID
254827
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
-
UniGene
Hs.565848
Gene Chromosome Location
Chr.3: 174439669-175809510 on Build GRCh38
Transcripts
NM_207015.2 XM_006713560.2 XM_011512612.2 XM_011512613.1 XM_011512615.2 XM_011512616.2 XM_011512617.2 XM_017006070.1 XM_017006071.1 XM_017006072.1 XM_017006073.1 XM_017006074.1 XM_017006075.1 XM_017006076.1 XM_017006077.1 XM_017006078.1 XM_017006079.1 XM_017006080.1 XM_017006081.1 XM_017006082.1 XM_017006083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
339805 c.1283C>A p.T428K Chr.3: 175463449-175463449 on Build GRCh38
1041402 c.1246G>A p.V416I Chr.3: 175463412-175463412 on Build GRCh38
5895129 c.1324C>T p.P442S Chr.3: 175463490-175463490 on Build GRCh38
1041401 c.1245C>T p.V415V Chr.3: 175463411-175463411 on Build GRCh38
5863963 c.1301G>A p.G434E Chr.3: 175463467-175463467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs578253029 Chr.3: 175463365-175463365 on Build GRCh38
rs938475140 Chr.3: 175463331-175463331 on Build GRCh38
rs755886615 Chr.3: 175463437-175463437 on Build GRCh38
rs767854854 Chr.3: 175463371-175463371 on Build GRCh38
rs774812790 Chr.3: 175463363-175463363 on Build GRCh38
rs1035581231 Chr.3: 175463318-175463318 on Build GRCh38
rs563896810 Chr.3: 175463403-175463403 on Build GRCh38
rs140774236 Chr.3: 175463476-175463476 on Build GRCh38
rs969239950 Chr.3: 175463384-175463384 on Build GRCh38
rs543188786 Chr.3: 175463421-175463421 on Build GRCh38
rs527375173 Chr.3: 175463442-175463442 on Build GRCh38
rs754178583 Chr.3: 175463337-175463337 on Build GRCh38
rs747922150 Chr.3: 175463425-175463425 on Build GRCh38
rs1021696946 Chr.3: 175463348-175463348 on Build GRCh38
rs868027292 Chr.3: 175463400-175463400 on Build GRCh38
rs147685357 Chr.3: 175463327-175463327 on Build GRCh38
rs368256642 Chr.3: 175463378-175463378 on Build GRCh38
rs764317937 Chr.3: 175463411-175463411 on Build GRCh38
rs181389157 Chr.3: 175463462-175463462 on Build GRCh38
rs144915385 Chr.3: 175463412-175463412 on Build GRCh38
rs189064809 Chr.3: 175463460-175463460 on Build GRCh38
rs956038817 Chr.3: 175463320-175463320 on Build GRCh38
rs748809910 Chr.3: 175463386-175463386 on Build GRCh38
rs765443531 Chr.3: 175463416-175463416 on Build GRCh38
rs771050085 Chr.3: 175463391-175463392 on Build GRCh38
rs544085272 Chr.3: 175463372-175463372 on Build GRCh38
rs757319395 Chr.3: 175463414-175463414 on Build GRCh38
rs183674783 Chr.3: 175463424-175463424 on Build GRCh38
rs928428681 Chr.3: 175463324-175463324 on Build GRCh38

View Full Product Details