Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120607564-120607837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEGF9
Entrez Gene ID
1955
Gene Name
multiple EGF like domains 9
Gene Aliases
EGFL5
UniGene
Hs.744903
Gene Chromosome Location
Chr.9: 120600918-120714487 on Build GRCh38
Transcripts
NM_001080497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3847511 c.1441G>A p.D481N Chr.9: 120607768-120607768 on Build GRCh38
7142997 c.1438C>A p.H480N Chr.9: 120607771-120607771 on Build GRCh38
6453894 c.1447G>A p.E483K Chr.9: 120607762-120607762 on Build GRCh38
7448996 c.1436T>G p.V479G Chr.9: 120607773-120607773 on Build GRCh38
5982999 c.1376G>A p.C459Y Chr.9: 120607833-120607833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757791120 Chr.9: 120607735-120607735 on Build GRCh38
rs1011844867 Chr.9: 120607652-120607652 on Build GRCh38
rs986837787 Chr.9: 120607615-120607615 on Build GRCh38
rs1047202800 Chr.9: 120607762-120607762 on Build GRCh38
rs752286487 Chr.9: 120607730-120607730 on Build GRCh38
rs1035474997 Chr.9: 120607751-120607751 on Build GRCh38
rs117904448 Chr.9: 120607660-120607660 on Build GRCh38
rs765853987 Chr.9: 120607745-120607745 on Build GRCh38
rs974523310 Chr.9: 120607655-120607655 on Build GRCh38
rs377425410 Chr.9: 120607786-120607786 on Build GRCh38
rs1002607257 Chr.9: 120607703-120607703 on Build GRCh38
rs958714659 Chr.9: 120607769-120607769 on Build GRCh38
rs767104634 Chr.9: 120607729-120607729 on Build GRCh38
rs762808714 Chr.9: 120607691-120607691 on Build GRCh38
rs923413250 Chr.9: 120607607-120607607 on Build GRCh38
rs759158527 Chr.9: 120607709-120607709 on Build GRCh38
rs781196686 Chr.9: 120607790-120607790 on Build GRCh38
rs758788598 Chr.9: 120607763-120607763 on Build GRCh38
rs750751150 Chr.9: 120607755-120607755 on Build GRCh38
rs183973988 Chr.9: 120607636-120607636 on Build GRCh38
rs371165156 Chr.9: 120607627-120607627 on Build GRCh38
rs748815705 Chr.9: 120607700-120607702 on Build GRCh38
rs774038445 Chr.9: 120607700-120607700 on Build GRCh38
rs374735546 Chr.9: 120607765-120607765 on Build GRCh38

View Full Product Details