Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6891500-6891773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2D2
Entrez Gene ID
120776
Gene Name
olfactory receptor family 2 subfamily D member 2
Gene Aliases
OR11-610, OR2D1, hg27
UniGene
Hs.690210
Gene Chromosome Location
Chr.11: 6891574-6892500 on Build GRCh38
Transcripts
NM_003700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1356595 c.907G>T p.A303S Chr.11: 6891594-6891594 on Build GRCh38
1356596 c.765A>T p.A255A Chr.11: 6891736-6891736 on Build GRCh38
458415 c.899G>A p.R300K Chr.11: 6891602-6891602 on Build GRCh38
4509022 c.794C>T p.S265F Chr.11: 6891707-6891707 on Build GRCh38
4511005 c.854C>T p.P285L Chr.11: 6891647-6891647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181589622 Chr.11: 6891688-6891688 on Build GRCh38
rs759782618 Chr.11: 6891646-6891646 on Build GRCh38
rs773005913 Chr.11: 6891543-6891543 on Build GRCh38
rs759888002 Chr.11: 6891550-6891550 on Build GRCh38
rs769748639 Chr.11: 6891540-6891540 on Build GRCh38
rs202199275 Chr.11: 6891597-6891597 on Build GRCh38
rs779107210 Chr.11: 6891735-6891735 on Build GRCh38
rs945113683 Chr.11: 6891696-6891696 on Build GRCh38
rs752846546 Chr.11: 6891555-6891555 on Build GRCh38
rs145606839 Chr.11: 6891672-6891672 on Build GRCh38
rs200086508 Chr.11: 6891663-6891663 on Build GRCh38
rs767118531 Chr.11: 6891744-6891744 on Build GRCh38
rs762181675 Chr.11: 6891730-6891730 on Build GRCh38
rs779102958 Chr.11: 6891679-6891679 on Build GRCh38
rs372237140 Chr.11: 6891729-6891729 on Build GRCh38
rs374844681 Chr.11: 6891564-6891564 on Build GRCh38
rs532310296 Chr.11: 6891706-6891706 on Build GRCh38
rs201106689 Chr.11: 6891612-6891612 on Build GRCh38
rs779380332 Chr.11: 6891587-6891587 on Build GRCh38
rs912856205 Chr.11: 6891658-6891658 on Build GRCh38
rs142545777 Chr.11: 6891655-6891655 on Build GRCh38
rs10566026 Chr.11: 6891691-6891692 on Build GRCh38
rs766789323 Chr.11: 6891738-6891738 on Build GRCh38
rs748844309 Chr.11: 6891750-6891750 on Build GRCh38
rs754433687 Chr.11: 6891579-6891579 on Build GRCh38
rs1006641478 Chr.11: 6891647-6891647 on Build GRCh38
rs150538963 Chr.11: 6891662-6891662 on Build GRCh38
rs761025081 Chr.11: 6891724-6891724 on Build GRCh38
rs745662413 Chr.11: 6891718-6891718 on Build GRCh38
rs769176545 Chr.11: 6891692-6891692 on Build GRCh38
rs769655037 Chr.11: 6891598-6891598 on Build GRCh38
rs546209667 Chr.11: 6891531-6891531 on Build GRCh38
rs770932912 Chr.11: 6891708-6891708 on Build GRCh38
rs117454717 Chr.11: 6891596-6891596 on Build GRCh38
rs773021712 Chr.11: 6891601-6891601 on Build GRCh38
rs997621860 Chr.11: 6891604-6891604 on Build GRCh38
rs775839284 Chr.11: 6891722-6891722 on Build GRCh38
rs139334477 Chr.11: 6891670-6891670 on Build GRCh38
rs113111396 Chr.11: 6891689-6891689 on Build GRCh38
rs1043242772 Chr.11: 6891701-6891701 on Build GRCh38
rs368013351 Chr.11: 6891591-6891591 on Build GRCh38
rs761702444 Chr.11: 6891539-6891539 on Build GRCh38
rs773958831 Chr.11: 6891712-6891712 on Build GRCh38
rs768128582 Chr.11: 6891536-6891536 on Build GRCh38
rs189179469 Chr.11: 6891681-6891681 on Build GRCh38
rs760830707 Chr.11: 6891666-6891666 on Build GRCh38
rs757818885 Chr.11: 6891674-6891674 on Build GRCh38
rs778146121 Chr.11: 6891571-6891571 on Build GRCh38
rs200826841 Chr.11: 6891653-6891653 on Build GRCh38
rs765791139 Chr.11: 6891731-6891731 on Build GRCh38
rs886492172 Chr.11: 6891623-6891623 on Build GRCh38
rs766265093 Chr.11: 6891548-6891548 on Build GRCh38
rs778233230 Chr.11: 6891530-6891530 on Build GRCh38
rs758760726 Chr.11: 6891736-6891736 on Build GRCh38
rs774998170 Chr.11: 6891535-6891535 on Build GRCh38
rs1035849729 Chr.11: 6891673-6891673 on Build GRCh38
rs767674678 Chr.11: 6891554-6891554 on Build GRCh38
rs772057797 Chr.11: 6891719-6891719 on Build GRCh38
rs762663377 Chr.11: 6891545-6891545 on Build GRCh38
rs144459508 Chr.11: 6891586-6891586 on Build GRCh38
rs563369489 Chr.11: 6891693-6891693 on Build GRCh38
rs774241928 Chr.11: 6891622-6891622 on Build GRCh38
rs71472615 Chr.11: 6891613-6891613 on Build GRCh38
rs752521490 Chr.11: 6891694-6891695 on Build GRCh38
rs750812955 Chr.11: 6891685-6891685 on Build GRCh38
rs771687046 Chr.11: 6891532-6891532 on Build GRCh38
rs755831124 Chr.11: 6891745-6891745 on Build GRCh38
rs754570829 Chr.11: 6891687-6891687 on Build GRCh38
rs763006889 Chr.11: 6891611-6891611 on Build GRCh38
rs375941397 Chr.11: 6891556-6891556 on Build GRCh38
rs375501732 Chr.11: 6891740-6891740 on Build GRCh38

View Full Product Details