Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41784026-41784299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRICKLE4
Entrez Gene ID
29964
Gene Name
prickle planar cell polarity protein 4
Gene Aliases
C6orf49, OBTP, OEBT, TOMM6
UniGene
Hs.715653
Gene Chromosome Location
Chr.6: 41780762-41787372 on Build GRCh38
Transcripts
NM_013397.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874006 c.227C>T p.P76L Chr.6: 41784225-41784225 on Build GRCh38
6442809 c.140C>A p.A47E Chr.6: 41784138-41784138 on Build GRCh38
1221981 c.164G>A p.C55Y Chr.6: 41784162-41784162 on Build GRCh38
1221982 c.164G>A p.C55Y Chr.6: 41784162-41784162 on Build GRCh38
1187030 c.133G>T p.G45C Chr.6: 41784131-41784131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746968534 Chr.6: 41784242-41784242 on Build GRCh38
rs750996996 Chr.6: 41784168-41784168 on Build GRCh38
rs368822026 Chr.6: 41784244-41784244 on Build GRCh38
rs1020289189 Chr.6: 41784187-41784187 on Build GRCh38
rs148499787 Chr.6: 41784215-41784215 on Build GRCh38
rs200945561 Chr.6: 41784140-41784140 on Build GRCh38
rs746903653 Chr.6: 41784113-41784113 on Build GRCh38
rs374944656 Chr.6: 41784226-41784226 on Build GRCh38
rs199995549 Chr.6: 41784238-41784238 on Build GRCh38
rs923960625 Chr.6: 41784221-41784221 on Build GRCh38
rs757876233 Chr.6: 41784119-41784119 on Build GRCh38
rs147190616 Chr.6: 41784154-41784154 on Build GRCh38
rs1015249509 Chr.6: 41784105-41784105 on Build GRCh38
rs752836843 Chr.6: 41784225-41784225 on Build GRCh38
rs745335706 Chr.6: 41784259-41784259 on Build GRCh38
rs149365193 Chr.6: 41784176-41784176 on Build GRCh38
rs778699841 Chr.6: 41784100-41784100 on Build GRCh38
rs755133927 Chr.6: 41784083-41784083 on Build GRCh38
rs767751683 Chr.6: 41784163-41784163 on Build GRCh38
rs1002741924 Chr.6: 41784087-41784087 on Build GRCh38
rs775257234 Chr.6: 41784264-41784264 on Build GRCh38
rs144685136 Chr.6: 41784199-41784199 on Build GRCh38
rs776211595 Chr.6: 41784126-41784126 on Build GRCh38
rs769401933 Chr.6: 41784261-41784261 on Build GRCh38
rs771374051 Chr.6: 41784175-41784182 on Build GRCh38
rs201242551 Chr.6: 41784073-41784073 on Build GRCh38
rs372003331 Chr.6: 41784204-41784204 on Build GRCh38
rs754128485 Chr.6: 41784076-41784076 on Build GRCh38
rs777812378 Chr.6: 41784108-41784108 on Build GRCh38
rs552176299 Chr.6: 41784070-41784072 on Build GRCh38
rs770630729 Chr.6: 41784125-41784125 on Build GRCh38
rs532404803 Chr.6: 41784255-41784255 on Build GRCh38
rs771731901 Chr.6: 41784106-41784106 on Build GRCh38
rs769524733 Chr.6: 41784145-41784145 on Build GRCh38
rs574774836 Chr.6: 41784101-41784101 on Build GRCh38
rs139191921 Chr.6: 41784162-41784162 on Build GRCh38
rs766459458 Chr.6: 41784195-41784195 on Build GRCh38

View Full Product Details