Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182902880-182903149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHCBP1L
Entrez Gene ID
81626
Gene Name
SHC binding and spindle associated 1 like
Gene Aliases
C1orf14, GE36
UniGene
Hs.497034
Gene Chromosome Location
Chr.1: 182899865-182953418 on Build GRCh38
Transcripts
NM_030933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6508363 c.1694G>T p.G565V Chr.1: 182903055-182903055 on Build GRCh38
5010667 c.1652G>A p.C551Y Chr.1: 182903097-182903097 on Build GRCh38
7256812 c.1645C>T p.H549Y Chr.1: 182903104-182903104 on Build GRCh38
4725848 c.1605G>A p.L535L Chr.1: 182903144-182903144 on Build GRCh38
900734 c.1645C>A p.H549N Chr.1: 182903104-182903104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898374004 Chr.1: 182902912-182902912 on Build GRCh38
rs887450879 Chr.1: 182903013-182903013 on Build GRCh38
rs368447502 Chr.1: 182903039-182903039 on Build GRCh38
rs1000782957 Chr.1: 182903087-182903087 on Build GRCh38
rs772583049 Chr.1: 182902998-182902998 on Build GRCh38
rs777379621 Chr.1: 182903118-182903118 on Build GRCh38
rs367611863 Chr.1: 182903114-182903114 on Build GRCh38
rs768044843 Chr.1: 182903091-182903091 on Build GRCh38
rs780391136 Chr.1: 182903104-182903104 on Build GRCh38
rs775788864 Chr.1: 182903064-182903064 on Build GRCh38
rs769657823 Chr.1: 182903050-182903050 on Build GRCh38
rs905199853 Chr.1: 182903035-182903035 on Build GRCh38
rs1006521335 Chr.1: 182903021-182903021 on Build GRCh38
rs757961209 Chr.1: 182903116-182903116 on Build GRCh38
rs528405720 Chr.1: 182902953-182902953 on Build GRCh38
rs143142615 Chr.1: 182903090-182903090 on Build GRCh38
rs1047305871 Chr.1: 182902978-182902978 on Build GRCh38
rs759423888 Chr.1: 182903041-182903041 on Build GRCh38
rs370479425 Chr.1: 182903094-182903094 on Build GRCh38
rs370979084 Chr.1: 182902996-182902996 on Build GRCh38
rs750882457 Chr.1: 182903093-182903093 on Build GRCh38
rs546883043 Chr.1: 182902976-182902976 on Build GRCh38
rs528204098 Chr.1: 182903073-182903073 on Build GRCh38
rs763085527 Chr.1: 182903066-182903066 on Build GRCh38
rs747011667 Chr.1: 182903008-182903008 on Build GRCh38
rs367881166 Chr.1: 182902918-182902918 on Build GRCh38
rs375280566 Chr.1: 182903038-182903038 on Build GRCh38
rs764297420 Chr.1: 182903089-182903089 on Build GRCh38
rs112777627 Chr.1: 182902921-182902921 on Build GRCh38
rs896345134 Chr.1: 182902987-182902987 on Build GRCh38

View Full Product Details