Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117099711-117099984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST7
Entrez Gene ID
7982
Gene Name
suppression of tumorigenicity 7
Gene Aliases
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7
UniGene
Hs.368131
Gene Chromosome Location
Chr.7: 116953327-117230023 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2

Gene Symbol
ST7-AS2
Entrez Gene ID
93654
Gene Name
ST7 antisense RNA 2
Gene Aliases
ST7AS2, ST7OT2
UniGene
Hs.628891
Gene Chromosome Location
Chr.7: 117072072-117145592 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7093221 c.28A>C p.M10L Chr.7: 117099767-117099767 on Build GRCh38
5796057 c.50C>T p.P17L Chr.7: 117099789-117099789 on Build GRCh38
7093220 c.157A>C p.M53L Chr.7: 117099767-117099767 on Build GRCh38
1647969 c.219A>T p.I73I Chr.7: 117099829-117099829 on Build GRCh38
7447644 c.95G>A p.G32E Chr.7: 117099834-117099834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773310860 Chr.7: 117099860-117099862 on Build GRCh38
rs759010682 Chr.7: 117099835-117099835 on Build GRCh38
rs200109461 Chr.7: 117099863-117099863 on Build GRCh38
rs940179441 Chr.7: 117099892-117099892 on Build GRCh38
rs543040003 Chr.7: 117099893-117099893 on Build GRCh38
rs113571541 Chr.7: 117099925-117099925 on Build GRCh38
rs772281894 Chr.7: 117099749-117099750 on Build GRCh38
rs768636943 Chr.7: 117099766-117099766 on Build GRCh38
rs751227866 Chr.7: 117099874-117099874 on Build GRCh38
rs901385088 Chr.7: 117099899-117099899 on Build GRCh38
rs969330792 Chr.7: 117099790-117099790 on Build GRCh38
rs773622593 Chr.7: 117099819-117099819 on Build GRCh38
rs368520848 Chr.7: 117099781-117099781 on Build GRCh38
rs780249073 Chr.7: 117099910-117099910 on Build GRCh38
rs961283677 Chr.7: 117099759-117099760 on Build GRCh38
rs553422266 Chr.7: 117099901-117099901 on Build GRCh38
rs749327257 Chr.7: 117099746-117099746 on Build GRCh38
rs992351149 Chr.7: 117099764-117099764 on Build GRCh38
rs1027971191 Chr.7: 117099924-117099924 on Build GRCh38
rs772419572 Chr.7: 117099799-117099799 on Build GRCh38
rs999893660 Chr.7: 117099930-117099930 on Build GRCh38
rs755030716 Chr.7: 117099853-117099853 on Build GRCh38
rs530179945 Chr.7: 117099747-117099751 on Build GRCh38
rs984848240 Chr.7: 117099841-117099841 on Build GRCh38
rs76146699 Chr.7: 117099905-117099905 on Build GRCh38
rs567991907 Chr.7: 117099823-117099823 on Build GRCh38
rs762476993 Chr.7: 117099856-117099856 on Build GRCh38
rs774606952 Chr.7: 117099778-117099778 on Build GRCh38

View Full Product Details