Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42907515-42907788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSG6
Entrez Gene ID
5675
Gene Name
pregnancy specific beta-1-glycoprotein 6
Gene Aliases
PSBG-10, PSBG-12, PSBG-6, PSG10, PSGGB
UniGene
Hs.646353
Gene Chromosome Location
Chr.19: 42902082-42917924 on Build GRCh38
Transcripts
NM_001031850.3 NM_002782.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3288393 c.940C>T p.R314* Chr.19: 42907621-42907621 on Build GRCh38
4078902 c.952A>G p.I318V Chr.19: 42907609-42907609 on Build GRCh38
3535078 c.798G>A p.R266R Chr.19: 42907763-42907763 on Build GRCh38
3535067 c.959G>A p.S320N Chr.19: 42907602-42907602 on Build GRCh38
3288410 c.848C>T p.P283L Chr.19: 42907713-42907713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758111461 Chr.19: 42907560-42907560 on Build GRCh38
rs150108851 Chr.19: 42907682-42907682 on Build GRCh38
rs370174981 Chr.19: 42907561-42907561 on Build GRCh38
rs778909429 Chr.19: 42907661-42907661 on Build GRCh38
rs746708210 Chr.19: 42907751-42907751 on Build GRCh38
rs146029289 Chr.19: 42907611-42907611 on Build GRCh38
rs768752410 Chr.19: 42907672-42907672 on Build GRCh38
rs765317566 Chr.19: 42907570-42907573 on Build GRCh38
rs747201776 Chr.19: 42907667-42907667 on Build GRCh38
rs771232910 Chr.19: 42907745-42907745 on Build GRCh38
rs763088500 Chr.19: 42907696-42907697 on Build GRCh38
rs767074930 Chr.19: 42907643-42907643 on Build GRCh38
rs192933037 Chr.19: 42907670-42907670 on Build GRCh38
rs151146504 Chr.19: 42907626-42907626 on Build GRCh38
rs780151842 Chr.19: 42907666-42907666 on Build GRCh38
rs148299784 Chr.19: 42907762-42907762 on Build GRCh38
rs763650711 Chr.19: 42907649-42907649 on Build GRCh38
rs774640978 Chr.19: 42907538-42907538 on Build GRCh38
rs149382921 Chr.19: 42907586-42907586 on Build GRCh38
rs776087393 Chr.19: 42907552-42907552 on Build GRCh38
rs757230889 Chr.19: 42907656-42907656 on Build GRCh38
rs368095253 Chr.19: 42907564-42907564 on Build GRCh38
rs144117726 Chr.19: 42907609-42907609 on Build GRCh38
rs752709303 Chr.19: 42907642-42907642 on Build GRCh38
rs567551600 Chr.19: 42907640-42907640 on Build GRCh38
rs200845699 Chr.19: 42907589-42907589 on Build GRCh38
rs138530848 Chr.19: 42907721-42907721 on Build GRCh38
rs76740241 Chr.19: 42907615-42907615 on Build GRCh38
rs558899002 Chr.19: 42907709-42907709 on Build GRCh38
rs775409558 Chr.19: 42907598-42907598 on Build GRCh38
rs773546902 Chr.19: 42907639-42907639 on Build GRCh38
rs531317587 Chr.19: 42907754-42907754 on Build GRCh38
rs200244036 Chr.19: 42907606-42907606 on Build GRCh38
rs199858569 Chr.19: 42907590-42907590 on Build GRCh38
rs917722601 Chr.19: 42907614-42907614 on Build GRCh38
rs778158670 Chr.19: 42907736-42907736 on Build GRCh38
rs373883813 Chr.19: 42907562-42907562 on Build GRCh38
rs564955936 Chr.19: 42907764-42907764 on Build GRCh38
rs767438804 Chr.19: 42907550-42907550 on Build GRCh38
rs764754374 Chr.19: 42907760-42907760 on Build GRCh38
rs144522626 Chr.19: 42907600-42907600 on Build GRCh38
rs772224955 Chr.19: 42907593-42907593 on Build GRCh38
rs755892290 Chr.19: 42907539-42907539 on Build GRCh38
rs756122986 Chr.19: 42907735-42907735 on Build GRCh38
rs375686772 Chr.19: 42907535-42907535 on Build GRCh38
rs753857556 Chr.19: 42907654-42907654 on Build GRCh38
rs111350515 Chr.19: 42907737-42907737 on Build GRCh38
rs188893610 Chr.19: 42907608-42907608 on Build GRCh38
rs771270387 Chr.19: 42907708-42907708 on Build GRCh38
rs527513587 Chr.19: 42907765-42907765 on Build GRCh38
rs200005028 Chr.19: 42907569-42907569 on Build GRCh38
rs770179988 Chr.19: 42907637-42907637 on Build GRCh38
rs141749854 Chr.19: 42907695-42907695 on Build GRCh38
rs202068387 Chr.19: 42907566-42907566 on Build GRCh38
rs1058725 Chr.19: 42907698-42907698 on Build GRCh38
rs201339573 Chr.19: 42907713-42907713 on Build GRCh38
rs138104787 Chr.19: 42907617-42907617 on Build GRCh38
rs533861405 Chr.19: 42907620-42907620 on Build GRCh38
rs777053898 Chr.19: 42907601-42907601 on Build GRCh38
rs779072398 Chr.19: 42907548-42907548 on Build GRCh38
rs768447561 Chr.19: 42907618-42907618 on Build GRCh38
rs140150194 Chr.19: 42907627-42907627 on Build GRCh38
rs752137945 Chr.19: 42907607-42907607 on Build GRCh38
rs763738520 Chr.19: 42907711-42907711 on Build GRCh38
rs143223284 Chr.19: 42907730-42907730 on Build GRCh38
rs762176964 Chr.19: 42907604-42907604 on Build GRCh38
rs374357654 Chr.19: 42907613-42907613 on Build GRCh38
rs750179815 Chr.19: 42907662-42907662 on Build GRCh38
rs34507332 Chr.19: 42907626-42907627 on Build GRCh38
rs761420127 Chr.19: 42907757-42907757 on Build GRCh38
rs1065516 Chr.19: 42907712-42907712 on Build GRCh38
rs750316162 Chr.19: 42907718-42907718 on Build GRCh38
rs372774102 Chr.19: 42907702-42907702 on Build GRCh38
rs774746293 Chr.19: 42907749-42907749 on Build GRCh38
rs755987161 Chr.19: 42907571-42907571 on Build GRCh38
rs748225819 Chr.19: 42907733-42907733 on Build GRCh38
rs1054341423 Chr.19: 42907567-42907567 on Build GRCh38
rs148457672 Chr.19: 42907605-42907605 on Build GRCh38
rs765268203 Chr.19: 42907714-42907714 on Build GRCh38
rs142652144 Chr.19: 42907722-42907722 on Build GRCh38
rs760117714 Chr.19: 42907644-42907644 on Build GRCh38
rs754187767 Chr.19: 42907556-42907556 on Build GRCh38
rs142100740 Chr.19: 42907663-42907663 on Build GRCh38
rs761055261 Chr.19: 42907554-42907554 on Build GRCh38
rs748706251 Chr.19: 42907690-42907690 on Build GRCh38
rs574785075 Chr.19: 42907723-42907723 on Build GRCh38
rs542265010 Chr.19: 42907727-42907727 on Build GRCh38
rs749754812 Chr.19: 42907744-42907744 on Build GRCh38

View Full Product Details