Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87441720-87441993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDT9
Entrez Gene ID
53343
Gene Name
nudix hydrolase 9
Gene Aliases
NUDT10
UniGene
Hs.149500
Gene Chromosome Location
Chr.4: 87420002-87459454 on Build GRCh38
Transcripts
NM_001248011.1 NM_024047.4 NM_198038.2 XM_011532031.2 XM_011532032.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
335670 c.496G>T p.G166C Chr.4: 87441881-87441881 on Build GRCh38
3409602 c.447T>C p.N149N Chr.4: 87441832-87441832 on Build GRCh38
343682 c.476G>A p.R159Q Chr.4: 87441861-87441861 on Build GRCh38
1486141 c.482T>G p.L161R Chr.4: 87441867-87441867 on Build GRCh38
3606828 c.449C>T p.P150L Chr.4: 87441834-87441834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910552037 Chr.4: 87441761-87441761 on Build GRCh38
rs770333037 Chr.4: 87441801-87441801 on Build GRCh38
rs898387728 Chr.4: 87441825-87441825 on Build GRCh38
rs764045825 Chr.4: 87441836-87441836 on Build GRCh38
rs768781660 Chr.4: 87441820-87441820 on Build GRCh38
rs781664110 Chr.4: 87441798-87441798 on Build GRCh38
rs980361109 Chr.4: 87441959-87441959 on Build GRCh38
rs115143660 Chr.4: 87441892-87441892 on Build GRCh38
rs1013338555 Chr.4: 87441893-87441893 on Build GRCh38
rs751160902 Chr.4: 87441904-87441904 on Build GRCh38
rs570706759 Chr.4: 87441759-87441759 on Build GRCh38
rs957829380 Chr.4: 87441912-87441912 on Build GRCh38
rs757402281 Chr.4: 87441811-87441812 on Build GRCh38
rs746228835 Chr.4: 87441799-87441799 on Build GRCh38
rs144581877 Chr.4: 87441871-87441871 on Build GRCh38
rs780560547 Chr.4: 87441817-87441817 on Build GRCh38
rs571942291 Chr.4: 87441875-87441875 on Build GRCh38
rs76684788 Chr.4: 87441955-87441955 on Build GRCh38
rs749815702 Chr.4: 87441951-87441951 on Build GRCh38
rs149319613 Chr.4: 87441842-87441842 on Build GRCh38
rs13104303 Chr.4: 87441752-87441752 on Build GRCh38
rs759324712 Chr.4: 87441867-87441867 on Build GRCh38
rs746239066 Chr.4: 87441861-87441861 on Build GRCh38
rs773183102 Chr.4: 87441854-87441854 on Build GRCh38
rs772177208 Chr.4: 87441843-87441843 on Build GRCh38
rs774234366 Chr.4: 87441822-87441822 on Build GRCh38
rs555381101 Chr.4: 87441780-87441780 on Build GRCh38
rs546726597 Chr.4: 87441874-87441874 on Build GRCh38
rs766231632 Chr.4: 87441860-87441860 on Build GRCh38
rs1050410750 Chr.4: 87441760-87441760 on Build GRCh38
rs79569855 Chr.4: 87441935-87441935 on Build GRCh38
rs535024591 Chr.4: 87441819-87441819 on Build GRCh38
rs376734452 Chr.4: 87441880-87441880 on Build GRCh38
rs772387372 Chr.4: 87441963-87441963 on Build GRCh38
rs756499072 Chr.4: 87441906-87441906 on Build GRCh38
rs763711551 Chr.4: 87441895-87441895 on Build GRCh38
rs780564558 Chr.4: 87441960-87441960 on Build GRCh38
rs201203653 Chr.4: 87441865-87441865 on Build GRCh38
rs760527779 Chr.4: 87441855-87441855 on Build GRCh38
rs1023287434 Chr.4: 87441945-87441945 on Build GRCh38
rs752399861 Chr.4: 87441778-87441778 on Build GRCh38

View Full Product Details