Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138145694-138145967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
UniGene
Hs.519337
Gene Chromosome Location
Chr.5: 138139766-138178669 on Build GRCh38
Transcripts
NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_011543109.1 XM_011543110.1 XM_017008969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357723 c.3316T>G p.L1106V Chr.5: 138145841-138145841 on Build GRCh38
4914183 c.3331A>G p.T1111A Chr.5: 138145826-138145826 on Build GRCh38
1486316 c.3302C>G p.P1101R Chr.5: 138145855-138145855 on Build GRCh38
1061230 c.3313C>A p.H1105N Chr.5: 138145844-138145844 on Build GRCh38
3608875 c.3351G>A p.K1117K Chr.5: 138145806-138145806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771499012 Chr.5: 138145824-138145824 on Build GRCh38
rs773926575 Chr.5: 138145819-138145819 on Build GRCh38
rs963005246 Chr.5: 138145768-138145768 on Build GRCh38
rs781238276 Chr.5: 138145781-138145781 on Build GRCh38
rs755480249 Chr.5: 138145745-138145745 on Build GRCh38
rs562443530 Chr.5: 138145844-138145844 on Build GRCh38
rs769765121 Chr.5: 138145784-138145784 on Build GRCh38
rs745772814 Chr.5: 138145782-138145782 on Build GRCh38
rs761182013 Chr.5: 138145822-138145822 on Build GRCh38
rs951435765 Chr.5: 138145735-138145735 on Build GRCh38
rs779581166 Chr.5: 138145889-138145889 on Build GRCh38
rs753050725 Chr.5: 138145850-138145850 on Build GRCh38
rs186114115 Chr.5: 138145863-138145863 on Build GRCh38
rs751523747 Chr.5: 138145745-138145745 on Build GRCh38
rs751679200 Chr.5: 138145879-138145879 on Build GRCh38
rs191930218 Chr.5: 138145933-138145933 on Build GRCh38
rs757229561 Chr.5: 138145766-138145766 on Build GRCh38
rs768128052 Chr.5: 138145809-138145809 on Build GRCh38
rs1042028077 Chr.5: 138145793-138145793 on Build GRCh38
rs572241884 Chr.5: 138145801-138145801 on Build GRCh38
rs777161857 Chr.5: 138145828-138145828 on Build GRCh38
rs552477382 Chr.5: 138145734-138145734 on Build GRCh38
rs748978579 Chr.5: 138145900-138145900 on Build GRCh38
rs982829938 Chr.5: 138145736-138145736 on Build GRCh38
rs747803564 Chr.5: 138145914-138145914 on Build GRCh38
rs369011132 Chr.5: 138145806-138145806 on Build GRCh38
rs375869120 Chr.5: 138145899-138145899 on Build GRCh38
rs114857951 Chr.5: 138145928-138145928 on Build GRCh38
rs765647270 Chr.5: 138145847-138145847 on Build GRCh38
rs180674606 Chr.5: 138145858-138145858 on Build GRCh38
rs756121412 Chr.5: 138145888-138145888 on Build GRCh38
rs757323117 Chr.5: 138145881-138145881 on Build GRCh38
rs1018588825 Chr.5: 138145846-138145846 on Build GRCh38
rs888844650 Chr.5: 138145839-138145839 on Build GRCh38
rs768378975 Chr.5: 138145908-138145908 on Build GRCh38
rs778391642 Chr.5: 138145912-138145912 on Build GRCh38
rs1029540148 Chr.5: 138145725-138145725 on Build GRCh38
rs750448260 Chr.5: 138145885-138145885 on Build GRCh38
rs79921495 Chr.5: 138145841-138145841 on Build GRCh38
rs373600592 Chr.5: 138145932-138145932 on Build GRCh38

View Full Product Details