Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14655480-14655753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC174
Entrez Gene ID
51244
Gene Name
coiled-coil domain containing 174
Gene Aliases
C3orf19, HSPC212, IHPM, IHPMR
UniGene
Hs.517820
Gene Chromosome Location
Chr.3: 14651683-14673186 on Build GRCh38
Transcripts
NM_016474.4 NR_135523.1 XM_017006554.1 XM_017006555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400361 c.148-4C>G p.? Chr.3: 14655525-14655525 on Build GRCh38
1246820 c.148-2A>T p.? Chr.3: 14655527-14655527 on Build GRCh38
6325077 c.165C>G p.S55R Chr.3: 14655546-14655546 on Build GRCh38
5465737 c.221A>G p.E74G Chr.3: 14655602-14655602 on Build GRCh38
355173 c.178G>T p.G60C Chr.3: 14655559-14655559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780581663 Chr.3: 14655572-14655572 on Build GRCh38
rs752011792 Chr.3: 14655647-14655647 on Build GRCh38
rs982987248 Chr.3: 14655668-14655668 on Build GRCh38
rs775199128 Chr.3: 14655644-14655645 on Build GRCh38
rs779334217 Chr.3: 14655547-14655547 on Build GRCh38
rs757325307 Chr.3: 14655541-14655541 on Build GRCh38
rs931645425 Chr.3: 14655717-14655717 on Build GRCh38
rs750958713 Chr.3: 14655555-14655555 on Build GRCh38
rs745353581 Chr.3: 14655679-14655679 on Build GRCh38
rs536096131 Chr.3: 14655654-14655654 on Build GRCh38
rs905451662 Chr.3: 14655557-14655557 on Build GRCh38
rs937682368 Chr.3: 14655552-14655553 on Build GRCh38
rs747368104 Chr.3: 14655586-14655586 on Build GRCh38
rs760457580 Chr.3: 14655624-14655624 on Build GRCh38
rs921636402 Chr.3: 14655697-14655697 on Build GRCh38
rs116166197 Chr.3: 14655674-14655674 on Build GRCh38
rs764339605 Chr.3: 14655625-14655625 on Build GRCh38
rs866474770 Chr.3: 14655626-14655626 on Build GRCh38
rs911830886 Chr.3: 14655727-14655727 on Build GRCh38
rs368397148 Chr.3: 14655520-14655520 on Build GRCh38
rs369953596 Chr.3: 14655608-14655608 on Build GRCh38
rs550199620 Chr.3: 14655603-14655603 on Build GRCh38
rs533165669 Chr.3: 14655710-14655710 on Build GRCh38
rs755372094 Chr.3: 14655650-14655650 on Build GRCh38
rs374806057 Chr.3: 14655637-14655637 on Build GRCh38
rs760898920 Chr.3: 14655508-14655508 on Build GRCh38
rs1023672586 Chr.3: 14655509-14655509 on Build GRCh38
rs372654465 Chr.3: 14655621-14655621 on Build GRCh38
rs954707271 Chr.3: 14655689-14655689 on Build GRCh38
rs376339243 Chr.3: 14655599-14655599 on Build GRCh38
rs777541132 Chr.3: 14655597-14655597 on Build GRCh38
rs765388143 Chr.3: 14655638-14655638 on Build GRCh38
rs753985103 Chr.3: 14655521-14655521 on Build GRCh38
rs987358659 Chr.3: 14655720-14655720 on Build GRCh38
rs771844299 Chr.3: 14655614-14655614 on Build GRCh38
rs775320473 Chr.3: 14655617-14655617 on Build GRCh38
rs776807893 Chr.3: 14655633-14655633 on Build GRCh38
rs201431396 Chr.3: 14655587-14655587 on Build GRCh38
rs1042243422 Chr.3: 14655644-14655644 on Build GRCh38
rs778500591 Chr.3: 14655676-14655676 on Build GRCh38
rs777907513 Chr.3: 14655612-14655612 on Build GRCh38
rs766990124 Chr.3: 14655642-14655642 on Build GRCh38
rs1015070381 Chr.3: 14655669-14655669 on Build GRCh38
rs1030858168 Chr.3: 14655681-14655681 on Build GRCh38
rs781651738 Chr.3: 14655655-14655655 on Build GRCh38
rs371975190 Chr.3: 14655562-14655562 on Build GRCh38
rs752425007 Chr.3: 14655506-14655506 on Build GRCh38
rs759004491 Chr.3: 14655639-14655639 on Build GRCh38
rs749004125 Chr.3: 14655659-14655659 on Build GRCh38

View Full Product Details