Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119274785-119275056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBL
Entrez Gene ID
867
Gene Name
Cbl proto-oncogene
Gene Aliases
C-CBL, CBL2, FRA11B, NSLL, RNF55
UniGene
Hs.504096
Gene Chromosome Location
Chr.11: 119206276-119308149 on Build GRCh38
Transcripts
NM_005188.3 XM_011543057.2 XM_017018511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6931898 c.749C>T p.P250L Chr.11: 119274833-119274833 on Build GRCh38
6978948 c.772T>C p.W258R Chr.11: 119274856-119274856 on Build GRCh38
4560222 c.835G>A p.A279T Chr.11: 119274919-119274919 on Build GRCh38
6848330 c.791_792delCT p.H265fs*10 Chr.11: 119274875-119274876 on Build GRCh38
4384633 c.793C>T p.H265Y Chr.11: 119274877-119274877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745855639 Chr.11: 119274936-119274936 on Build GRCh38
rs746902057 Chr.11: 119274942-119274942 on Build GRCh38
rs766797364 Chr.11: 119274906-119274906 on Build GRCh38
rs763429368 Chr.11: 119274973-119274973 on Build GRCh38
rs980197604 Chr.11: 119275005-119275005 on Build GRCh38
rs751768529 Chr.11: 119274909-119274909 on Build GRCh38
rs397751135 Chr.11: 119274996-119274997 on Build GRCh38
rs771883413 Chr.11: 119274945-119274945 on Build GRCh38
rs372452974 Chr.11: 119274889-119274889 on Build GRCh38
rs750907780 Chr.11: 119274903-119274903 on Build GRCh38
rs145155035 Chr.11: 119274923-119274923 on Build GRCh38
rs768380727 Chr.11: 119274963-119274963 on Build GRCh38
rs761328610 Chr.11: 119274968-119274968 on Build GRCh38
rs181589369 Chr.11: 119274972-119274972 on Build GRCh38
rs751962708 Chr.11: 119274977-119274977 on Build GRCh38
rs780039741 Chr.11: 119274952-119274952 on Build GRCh38
rs776443679 Chr.11: 119274967-119274967 on Build GRCh38
rs3842642 Chr.11: 119274995-119274996 on Build GRCh38
rs927136981 Chr.11: 119275023-119275023 on Build GRCh38
rs759887134 Chr.11: 119275004-119275004 on Build GRCh38
rs771295995 Chr.11: 119274994-119274995 on Build GRCh38
rs730880432 Chr.11: 119274922-119274922 on Build GRCh38
rs200533967 Chr.11: 119274816-119274816 on Build GRCh38
rs77284821 Chr.11: 119274957-119274957 on Build GRCh38
rs756156834 Chr.11: 119274995-119274995 on Build GRCh38
rs763112996 Chr.11: 119274975-119274975 on Build GRCh38
rs373395283 Chr.11: 119274975-119274975 on Build GRCh38
rs755244263 Chr.11: 119274910-119274910 on Build GRCh38
rs140250592 Chr.11: 119274921-119274921 on Build GRCh38
rs759928015 Chr.11: 119274980-119274980 on Build GRCh38
rs727502913 Chr.11: 119274885-119274885 on Build GRCh38
rs765725680 Chr.11: 119274886-119274886 on Build GRCh38
rs776318077 Chr.11: 119274837-119274837 on Build GRCh38
rs1040022511 Chr.11: 119274813-119274813 on Build GRCh38
rs1244893 Chr.11: 119274821-119274821 on Build GRCh38
rs879784628 Chr.11: 119274817-119274817 on Build GRCh38
rs750348894 Chr.11: 119274999-119274999 on Build GRCh38
rs1055722937 Chr.11: 119274823-119274823 on Build GRCh38
rs767680530 Chr.11: 119274993-119274993 on Build GRCh38
rs778856115 Chr.11: 119274935-119274935 on Build GRCh38
rs752879816 Chr.11: 119274994-119274994 on Build GRCh38

View Full Product Details