Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2408291-2408564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX10
Entrez Gene ID
5192
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
NALD, PBD6A, PBD6B, RNF69
UniGene
Hs.732228
Gene Chromosome Location
Chr.1: 2404802-2412622 on Build GRCh38
Transcripts
NM_002617.3 NM_153818.1 XM_011541573.1 XM_011541576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1340263 c.575C>A p.A192D Chr.1: 2408477-2408477 on Build GRCh38
6744076 c.531A>G p.L177L Chr.1: 2408521-2408521 on Build GRCh38
6744075 c.556G>A p.G186S Chr.1: 2408496-2408496 on Build GRCh38
6744074 c.587C>T p.T196M Chr.1: 2408465-2408465 on Build GRCh38
6356993 c.600+1G>A p.? Chr.1: 2408451-2408451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116354317 Chr.1: 2408469-2408469 on Build GRCh38
rs568936483 Chr.1: 2408352-2408352 on Build GRCh38
rs267608183 Chr.1: 2408451-2408451 on Build GRCh38
rs201488304 Chr.1: 2408465-2408465 on Build GRCh38
rs75377471 Chr.1: 2408497-2408497 on Build GRCh38
rs752730970 Chr.1: 2408478-2408478 on Build GRCh38
rs761202759 Chr.1: 2408443-2408443 on Build GRCh38
rs973491890 Chr.1: 2408372-2408372 on Build GRCh38
rs35373002 Chr.1: 2408385-2408386 on Build GRCh38
rs776004824 Chr.1: 2408433-2408433 on Build GRCh38
rs147848221 Chr.1: 2408496-2408496 on Build GRCh38
rs772790675 Chr.1: 2408539-2408539 on Build GRCh38
rs749637005 Chr.1: 2408510-2408510 on Build GRCh38
rs1026741885 Chr.1: 2408517-2408517 on Build GRCh38
rs766103610 Chr.1: 2408464-2408464 on Build GRCh38
rs944648966 Chr.1: 2408491-2408491 on Build GRCh38
rs780485807 Chr.1: 2408505-2408505 on Build GRCh38
rs747996191 Chr.1: 2408442-2408442 on Build GRCh38
rs532243871 Chr.1: 2408415-2408415 on Build GRCh38
rs773164888 Chr.1: 2408460-2408460 on Build GRCh38
rs116304725 Chr.1: 2408414-2408414 on Build GRCh38
rs376805362 Chr.1: 2408476-2408476 on Build GRCh38
rs757238209 Chr.1: 2408492-2408492 on Build GRCh38
rs546573745 Chr.1: 2408315-2408315 on Build GRCh38
rs549271413 Chr.1: 2408526-2408526 on Build GRCh38
rs769189410 Chr.1: 2408514-2408514 on Build GRCh38
rs772255287 Chr.1: 2408418-2408419 on Build GRCh38
rs766015781 Chr.1: 2408544-2408544 on Build GRCh38
rs184150138 Chr.1: 2408525-2408525 on Build GRCh38
rs375803322 Chr.1: 2408439-2408439 on Build GRCh38
rs760162500 Chr.1: 2408541-2408541 on Build GRCh38
rs774949268 Chr.1: 2408520-2408520 on Build GRCh38
rs533977936 Chr.1: 2408538-2408538 on Build GRCh38
rs201828636 Chr.1: 2408429-2408429 on Build GRCh38
rs763465246 Chr.1: 2408438-2408438 on Build GRCh38
rs753617772 Chr.1: 2408467-2408467 on Build GRCh38
rs756342887 Chr.1: 2408502-2408502 on Build GRCh38
rs1054757339 Chr.1: 2408327-2408327 on Build GRCh38
rs760694123 Chr.1: 2408539-2408559 on Build GRCh38
rs750002192 Chr.1: 2408455-2408455 on Build GRCh38
rs373559391 Chr.1: 2408456-2408456 on Build GRCh38

View Full Product Details