Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33260598-33260867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNAR2
Entrez Gene ID
3455
Gene Name
interferon alpha and beta receptor subunit 2
Gene Aliases
IFN-R, IFN-alpha-REC, IFNABR, IFNARB, IMD45
UniGene
Hs.708195
Gene Chromosome Location
Chr.21: 33229895-33264513 on Build GRCh38
Transcripts
NM_000874.4 NM_001289125.1 NM_001289126.1 NM_001289128.1 NM_207584.2 NM_207585.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207293 c.740T>G p.I247R Chr.21: 33260627-33260627 on Build GRCh38
6267537 c.724G>T p.A242S Chr.21: 33260611-33260611 on Build GRCh38
7283636 c.840+8_840+9insT p.? Chr.21: 33260735-33260736 on Build GRCh38
6207292 c.740T>G p.I247R Chr.21: 33260627-33260627 on Build GRCh38
6267536 c.724G>T p.A242S Chr.21: 33260611-33260611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769304952 Chr.21: 33260639-33260639 on Build GRCh38
rs753261594 Chr.21: 33260765-33260768 on Build GRCh38
rs545235375 Chr.21: 33260788-33260788 on Build GRCh38
rs984234596 Chr.21: 33260649-33260649 on Build GRCh38
rs999804650 Chr.21: 33260822-33260822 on Build GRCh38
rs754280566 Chr.21: 33260746-33260748 on Build GRCh38
rs779941156 Chr.21: 33260626-33260626 on Build GRCh38
rs768442811 Chr.21: 33260634-33260634 on Build GRCh38
rs757314865 Chr.21: 33260685-33260685 on Build GRCh38
rs201848104 Chr.21: 33260693-33260693 on Build GRCh38
rs567371989 Chr.21: 33260655-33260655 on Build GRCh38
rs755289487 Chr.21: 33260749-33260749 on Build GRCh38
rs747004525 Chr.21: 33260627-33260627 on Build GRCh38
rs772694912 Chr.21: 33260769-33260769 on Build GRCh38
rs115301063 Chr.21: 33260699-33260699 on Build GRCh38
rs748049340 Chr.21: 33260759-33260759 on Build GRCh38
rs758917391 Chr.21: 33260778-33260782 on Build GRCh38
rs746775306 Chr.21: 33260728-33260728 on Build GRCh38
rs577147247 Chr.21: 33260793-33260793 on Build GRCh38
rs918622040 Chr.21: 33260690-33260690 on Build GRCh38
rs761211574 Chr.21: 33260661-33260661 on Build GRCh38
rs754323542 Chr.21: 33260669-33260669 on Build GRCh38
rs758683918 Chr.21: 33260709-33260709 on Build GRCh38
rs949178140 Chr.21: 33260737-33260737 on Build GRCh38
rs765919947 Chr.21: 33260725-33260740 on Build GRCh38
rs769199239 Chr.21: 33260764-33260764 on Build GRCh38
rs371848732 Chr.21: 33260647-33260647 on Build GRCh38
rs764488006 Chr.21: 33260668-33260668 on Build GRCh38
rs577735060 Chr.21: 33260752-33260752 on Build GRCh38
rs973165486 Chr.21: 33260663-33260663 on Build GRCh38
rs780031206 Chr.21: 33260712-33260712 on Build GRCh38
rs776250783 Chr.21: 33260635-33260635 on Build GRCh38
rs555431486 Chr.21: 33260637-33260637 on Build GRCh38
rs191901053 Chr.21: 33260810-33260810 on Build GRCh38
rs554352383 Chr.21: 33260801-33260801 on Build GRCh38
rs762532844 Chr.21: 33260650-33260650 on Build GRCh38
rs779544730 Chr.21: 33260753-33260754 on Build GRCh38
rs775827448 Chr.21: 33260659-33260659 on Build GRCh38
rs747375188 Chr.21: 33260742-33260742 on Build GRCh38
rs35152486 Chr.21: 33260744-33260744 on Build GRCh38

View Full Product Details