Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159262437-159262694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCJ
Entrez Gene ID
654502
Gene Name
IQ motif containing J
Gene Aliases
-
UniGene
Hs.570685
Gene Chromosome Location
Chr.3: 159069252-159266307 on Build GRCh38
Transcripts
NM_001042705.2 NM_001042706.2 NM_001197100.1 NM_001197113.1 NM_001197114.1

Gene Symbol
IQCJ-SCHIP1
Entrez Gene ID
100505385
Gene Name
IQCJ-SCHIP1 readthrough
Gene Aliases
-
UniGene
Hs.134665
Gene Chromosome Location
Chr.3: 159069252-159897366 on Build GRCh38
Transcripts
NM_001042705.2 NM_001042706.2 NM_001197100.1 NM_001197113.1 NM_001197114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3005895 c.188G>A p.R63Q Chr.3: 159262580-159262580 on Build GRCh38
7372411 c.199C>A p.L67M Chr.3: 159262591-159262591 on Build GRCh38
3005896 c.188G>A p.R63Q Chr.3: 159262580-159262580 on Build GRCh38
7233617 c.176A>G p.E59G Chr.3: 159262568-159262568 on Build GRCh38
6686873 c.187C>T p.R63W Chr.3: 159262579-159262579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74923751 Chr.3: 159262503-159262503 on Build GRCh38
rs748254612 Chr.3: 159262522-159262522 on Build GRCh38
rs200957112 Chr.3: 159262669-159262669 on Build GRCh38
rs779215402 Chr.3: 159262564-159262564 on Build GRCh38
rs571468794 Chr.3: 159262573-159262573 on Build GRCh38
rs755143969 Chr.3: 159262597-159262597 on Build GRCh38
rs377145005 Chr.3: 159262565-159262565 on Build GRCh38
rs766827460 Chr.3: 159262579-159262579 on Build GRCh38
rs565719628 Chr.3: 159262656-159262656 on Build GRCh38
rs773117966 Chr.3: 159262609-159262610 on Build GRCh38
rs745755758 Chr.3: 159262608-159262608 on Build GRCh38
rs765699106 Chr.3: 159262662-159262662 on Build GRCh38
rs758467568 Chr.3: 159262592-159262592 on Build GRCh38
rs749015086 Chr.3: 159262605-159262605 on Build GRCh38
rs756596599 Chr.3: 159262560-159262560 on Build GRCh38
rs750756440 Chr.3: 159262665-159262665 on Build GRCh38
rs752262085 Chr.3: 159262577-159262577 on Build GRCh38
rs779263033 Chr.3: 159262586-159262586 on Build GRCh38
rs781518251 Chr.3: 159262598-159262598 on Build GRCh38
rs780239968 Chr.3: 159262545-159262545 on Build GRCh38
rs770818723 Chr.3: 159262606-159262606 on Build GRCh38
rs750994629 Chr.3: 159262588-159262588 on Build GRCh38
rs115299774 Chr.3: 159262483-159262483 on Build GRCh38
rs879081010 Chr.3: 159262610-159262610 on Build GRCh38
rs760296981 Chr.3: 159262649-159262649 on Build GRCh38
rs200636747 Chr.3: 159262555-159262555 on Build GRCh38
rs747297020 Chr.3: 159262548-159262548 on Build GRCh38
rs754076299 Chr.3: 159262511-159262511 on Build GRCh38
rs754247661 Chr.3: 159262504-159262504 on Build GRCh38
rs202040350 Chr.3: 159262642-159262642 on Build GRCh38
rs745902630 Chr.3: 159262526-159262526 on Build GRCh38
rs763804161 Chr.3: 159262653-159262653 on Build GRCh38
rs762067284 Chr.3: 159262660-159262660 on Build GRCh38
rs939163676 Chr.3: 159262543-159262543 on Build GRCh38
rs201513869 Chr.3: 159262607-159262607 on Build GRCh38
rs757677393 Chr.3: 159262585-159262585 on Build GRCh38
rs375080915 Chr.3: 159262580-159262580 on Build GRCh38
rs769947972 Chr.3: 159262538-159262538 on Build GRCh38
rs766893360 Chr.3: 159262668-159262668 on Build GRCh38
rs774397281 Chr.3: 159262570-159262570 on Build GRCh38
rs1449007 Chr.3: 159262490-159262490 on Build GRCh38
rs758844693 Chr.3: 159262667-159262667 on Build GRCh38
rs760350127 Chr.3: 159262578-159262578 on Build GRCh38
rs925912769 Chr.3: 159262622-159262622 on Build GRCh38
rs780040850 Chr.3: 159262594-159262594 on Build GRCh38
rs779501859 Chr.3: 159262515-159262515 on Build GRCh38
rs538886885 Chr.3: 159262630-159262630 on Build GRCh38
rs758956389 Chr.3: 159262571-159262571 on Build GRCh38
rs373701672 Chr.3: 159262558-159262558 on Build GRCh38
rs775096379 Chr.3: 159262647-159262647 on Build GRCh38
rs1026776737 Chr.3: 159262602-159262602 on Build GRCh38
rs772239276 Chr.3: 159262528-159262528 on Build GRCh38
rs747105085 Chr.3: 159262595-159262595 on Build GRCh38

View Full Product Details