Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5986947-5987176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PMS2
Entrez Gene ID
5395
Gene Name
PMS1 homolog 2, mismatch repair system component
Gene Aliases
HNPCC4, MLH4, PMS2CL, PMSL2
UniGene
Hs.632637
Gene Chromosome Location
Chr.7: 5970925-6009106 on Build GRCh38
Transcripts
NM_000535.6 NM_001322003.1 NM_001322004.1 NM_001322005.1 NM_001322006.1 NM_001322007.1 NM_001322008.1 NM_001322009.1 NM_001322010.1 NM_001322011.1 NM_001322012.1 NM_001322013.1 NM_001322014.1 NM_001322015.1 NR_136154.1 XM_006715744.3 XM_017012342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6481477 c.1635T>A p.S545S Chr.7: 5987130-5987130 on Build GRCh38
3923870 c.1655A>G p.H552R Chr.7: 5987110-5987110 on Build GRCh38
6912914 c.1697C>A p.P566H Chr.7: 5987068-5987068 on Build GRCh38
1736025 c.1672A>G p.T558A Chr.7: 5987093-5987093 on Build GRCh38
1742257 c.1744G>C p.E582Q Chr.7: 5987021-5987021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs63750278 Chr.7: 5986974-5986974 on Build GRCh38
rs63750477 Chr.7: 5986997-5986997 on Build GRCh38
rs370455197 Chr.7: 5987087-5987087 on Build GRCh38
rs749727182 Chr.7: 5987000-5987000 on Build GRCh38
rs2228006 Chr.7: 5987144-5987144 on Build GRCh38
rs751153838 Chr.7: 5987090-5987090 on Build GRCh38
rs1057515572 Chr.7: 5987033-5987034 on Build GRCh38
rs878854039 Chr.7: 5986969-5986969 on Build GRCh38
rs758018736 Chr.7: 5987045-5987045 on Build GRCh38
rs587778618 Chr.7: 5987078-5987078 on Build GRCh38
rs757989905 Chr.7: 5987086-5987086 on Build GRCh38
rs786203439 Chr.7: 5987133-5987133 on Build GRCh38
rs767724368 Chr.7: 5987069-5987069 on Build GRCh38
rs770625733 Chr.7: 5987050-5987050 on Build GRCh38
rs876659780 Chr.7: 5987097-5987097 on Build GRCh38
rs754542046 Chr.7: 5987037-5987037 on Build GRCh38
rs764749700 Chr.7: 5987079-5987079 on Build GRCh38
rs876658481 Chr.7: 5987083-5987083 on Build GRCh38
rs745836140 Chr.7: 5987101-5987101 on Build GRCh38
rs878854038 Chr.7: 5986975-5986975 on Build GRCh38
rs557906137 Chr.7: 5987131-5987131 on Build GRCh38
rs762100304 Chr.7: 5987005-5987005 on Build GRCh38
rs587781289 Chr.7: 5987143-5987143 on Build GRCh38
rs1057517801 Chr.7: 5987022-5987022 on Build GRCh38
rs786202870 Chr.7: 5987072-5987072 on Build GRCh38
rs786204114 Chr.7: 5987111-5987111 on Build GRCh38
rs63750598 Chr.7: 5987024-5987024 on Build GRCh38
rs138222146 Chr.7: 5987149-5987149 on Build GRCh38
rs786202656 Chr.7: 5987099-5987099 on Build GRCh38
rs63750055 Chr.7: 5987054-5987054 on Build GRCh38
rs786201630 Chr.7: 5987121-5987121 on Build GRCh38
rs876659162 Chr.7: 5987112-5987112 on Build GRCh38
rs587780043 Chr.7: 5987154-5987155 on Build GRCh38
rs753256070 Chr.7: 5987130-5987131 on Build GRCh38
rs876659285 Chr.7: 5987100-5987100 on Build GRCh38
rs779150753 Chr.7: 5987119-5987119 on Build GRCh38
rs773541976 Chr.7: 5987124-5987124 on Build GRCh38
rs750754115 Chr.7: 5987028-5987028 on Build GRCh38
rs764252217 Chr.7: 5986994-5986994 on Build GRCh38
rs267608169 Chr.7: 5987027-5987027 on Build GRCh38
rs142506484 Chr.7: 5987042-5987042 on Build GRCh38
rs587779330 Chr.7: 5987034-5987035 on Build GRCh38
rs876661004 Chr.7: 5987076-5987077 on Build GRCh38
rs63750947 Chr.7: 5987012-5987012 on Build GRCh38
rs267608165 Chr.7: 5986969-5986970 on Build GRCh38
rs112104877 Chr.7: 5987065-5987065 on Build GRCh38
rs63750890 Chr.7: 5987004-5987005 on Build GRCh38
rs899994632 Chr.7: 5986987-5986987 on Build GRCh38
rs1805318 Chr.7: 5986976-5986976 on Build GRCh38
rs63750476 Chr.7: 5986997-5986997 on Build GRCh38
rs864622600 Chr.7: 5987014-5987015 on Build GRCh38
rs111673299 Chr.7: 5987136-5987136 on Build GRCh38
rs786204115 Chr.7: 5987063-5987063 on Build GRCh38
rs869312801 Chr.7: 5987062-5987062 on Build GRCh38
rs876658134 Chr.7: 5987091-5987091 on Build GRCh38
rs781106967 Chr.7: 5987093-5987093 on Build GRCh38
rs876660139 Chr.7: 5987135-5987135 on Build GRCh38
rs374591423 Chr.7: 5987123-5987123 on Build GRCh38
rs63750593 Chr.7: 5986983-5986983 on Build GRCh38
rs747828749 Chr.7: 5987044-5987044 on Build GRCh38
rs63751459 Chr.7: 5987018-5987018 on Build GRCh38
rs63750553 Chr.7: 5987005-5987006 on Build GRCh38
rs876659850 Chr.7: 5987091-5987092 on Build GRCh38
rs752143026 Chr.7: 5987085-5987085 on Build GRCh38
rs776931237 Chr.7: 5987148-5987148 on Build GRCh38
rs63750770 Chr.7: 5987032-5987032 on Build GRCh38
rs745774944 Chr.7: 5987052-5987052 on Build GRCh38
rs766389591 Chr.7: 5986987-5986987 on Build GRCh38
rs754339271 Chr.7: 5987036-5987036 on Build GRCh38
rs863224498 Chr.7: 5987126-5987127 on Build GRCh38
rs774767419 Chr.7: 5986999-5986999 on Build GRCh38
rs63751023 Chr.7: 5987051-5987051 on Build GRCh38
rs876660076 Chr.7: 5987047-5987047 on Build GRCh38
rs762151417 Chr.7: 5987060-5987060 on Build GRCh38
rs63750994 Chr.7: 5986991-5986991 on Build GRCh38
rs113726095 Chr.7: 5987109-5987109 on Build GRCh38
rs115670442 Chr.7: 5987057-5987057 on Build GRCh38
rs114037612 Chr.7: 5987092-5987092 on Build GRCh38
rs587780725 Chr.7: 5987076-5987077 on Build GRCh38
rs63750535 Chr.7: 5987147-5987147 on Build GRCh38
rs63750739 Chr.7: 5987021-5987021 on Build GRCh38
rs63750594 Chr.7: 5986971-5986971 on Build GRCh38
rs63751211 Chr.7: 5987048-5987048 on Build GRCh38
rs551226281 Chr.7: 5987076-5987076 on Build GRCh38
rs863224677 Chr.7: 5987039-5987039 on Build GRCh38
rs63750534 Chr.7: 5987033-5987033 on Build GRCh38
rs63750668 Chr.7: 5987077-5987077 on Build GRCh38

View Full Product Details