Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3298909-3299174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGD7
Entrez Gene ID
91151
Gene Name
tigger transposable element derived 7
Gene Aliases
Sancho
UniGene
Hs.161287
Gene Chromosome Location
Chr.16: 3298808-3305439 on Build GRCh38
Transcripts
NM_033208.3 XM_011522344.1 XM_017022850.1

Gene Symbol
ZNF263
Entrez Gene ID
10127
Gene Name
zinc finger protein 263
Gene Aliases
FPM315, ZKSCAN12, ZSCAN44
UniGene
Hs.611475
Gene Chromosome Location
Chr.16: 3283183-3301401 on Build GRCh38
Transcripts
NM_033208.3 XM_011522344.1 XM_017022850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5722381 c.1621G>A p.G541R Chr.16: 3298994-3298994 on Build GRCh38
970408 c.1512G>A p.L504L Chr.16: 3299103-3299103 on Build GRCh38
1177366 c.1572T>C p.I524I Chr.16: 3299043-3299043 on Build GRCh38
1609256 c.1587delA p.K529fs*>21 Chr.16: 3299028-3299028 on Build GRCh38
6328197 c.1640C>T p.S547F Chr.16: 3298975-3298975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762096054 Chr.16: 3298953-3298953 on Build GRCh38
rs745665015 Chr.16: 3299055-3299055 on Build GRCh38
rs143297362 Chr.16: 3299115-3299115 on Build GRCh38
rs371098080 Chr.16: 3298964-3298964 on Build GRCh38
rs1020656105 Chr.16: 3299017-3299017 on Build GRCh38
rs201590215 Chr.16: 3299022-3299022 on Build GRCh38
rs763345902 Chr.16: 3298993-3298993 on Build GRCh38
rs966881596 Chr.16: 3299024-3299024 on Build GRCh38
rs763572734 Chr.16: 3299137-3299137 on Build GRCh38
rs774764906 Chr.16: 3299124-3299124 on Build GRCh38
rs540573199 Chr.16: 3299148-3299148 on Build GRCh38
rs760069622 Chr.16: 3299132-3299132 on Build GRCh38
rs771286715 Chr.16: 3299121-3299121 on Build GRCh38
rs925411763 Chr.16: 3299111-3299111 on Build GRCh38
rs757804723 Chr.16: 3298935-3298935 on Build GRCh38
rs370541421 Chr.16: 3299021-3299021 on Build GRCh38
rs144459500 Chr.16: 3298996-3298996 on Build GRCh38
rs746925011 Chr.16: 3299079-3299079 on Build GRCh38
rs563938015 Chr.16: 3299025-3299025 on Build GRCh38
rs367867234 Chr.16: 3298997-3298997 on Build GRCh38
rs796348520 Chr.16: 3298968-3298968 on Build GRCh38
rs746484361 Chr.16: 3298930-3298930 on Build GRCh38
rs778748902 Chr.16: 3299043-3299043 on Build GRCh38
rs534783376 Chr.16: 3299127-3299127 on Build GRCh38
rs781307955 Chr.16: 3299090-3299090 on Build GRCh38
rs370768253 Chr.16: 3298932-3298932 on Build GRCh38
rs748437181 Chr.16: 3299096-3299096 on Build GRCh38
rs139056404 Chr.16: 3299013-3299013 on Build GRCh38
rs377637766 Chr.16: 3298950-3298950 on Build GRCh38
rs768657453 Chr.16: 3299084-3299084 on Build GRCh38
rs755611172 Chr.16: 3299002-3299002 on Build GRCh38
rs773512281 Chr.16: 3299104-3299104 on Build GRCh38
rs112964318 Chr.16: 3298985-3298985 on Build GRCh38
rs961596488 Chr.16: 3299052-3299052 on Build GRCh38
rs779874893 Chr.16: 3299073-3299073 on Build GRCh38
rs758127998 Chr.16: 3299064-3299064 on Build GRCh38
rs747069192 Chr.16: 3298937-3298937 on Build GRCh38
rs770146674 Chr.16: 3299097-3299097 on Build GRCh38

View Full Product Details