Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13595406-13595641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT5
Entrez Gene ID
23408
Gene Name
sirtuin 5
Gene Aliases
SIR2L5
UniGene
Hs.567431
Gene Chromosome Location
Chr.6: 13574222-13615158 on Build GRCh38
Transcripts
NM_001193267.2 NM_001242827.1 NM_012241.4 NM_031244.3 XM_005248968.4 XM_011514444.2 XM_017010620.1 XM_017010621.1 XM_017010622.1 XM_017010623.1 XM_017010624.1 XM_017010625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4851686 c.520G>C p.E174Q Chr.6: 13595521-13595521 on Build GRCh38
3858492 c.536C>A p.P179Q Chr.6: 13595537-13595537 on Build GRCh38
3858493 c.536C>A p.P179Q Chr.6: 13595537-13595537 on Build GRCh38
1073641 c.504T>C p.S168S Chr.6: 13595505-13595505 on Build GRCh38
6412048 c.545C>G p.P182R Chr.6: 13595546-13595546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770601877 Chr.6: 13595573-13595573 on Build GRCh38
rs766014704 Chr.6: 13595448-13595448 on Build GRCh38
rs375024007 Chr.6: 13595529-13595529 on Build GRCh38
rs780555346 Chr.6: 13595584-13595584 on Build GRCh38
rs200388975 Chr.6: 13595552-13595552 on Build GRCh38
rs893888985 Chr.6: 13595610-13595610 on Build GRCh38
rs762943307 Chr.6: 13595543-13595543 on Build GRCh38
rs772786827 Chr.6: 13595523-13595523 on Build GRCh38
rs111305646 Chr.6: 13595497-13595497 on Build GRCh38
rs757231999 Chr.6: 13595562-13595562 on Build GRCh38
rs750601949 Chr.6: 13595573-13595573 on Build GRCh38
rs374083957 Chr.6: 13595449-13595449 on Build GRCh38
rs201858712 Chr.6: 13595458-13595458 on Build GRCh38
rs749684655 Chr.6: 13595590-13595590 on Build GRCh38
rs760365829 Chr.6: 13595447-13595447 on Build GRCh38
rs545871951 Chr.6: 13595446-13595446 on Build GRCh38
rs1016909428 Chr.6: 13595495-13595495 on Build GRCh38
rs768709833 Chr.6: 13595597-13595597 on Build GRCh38
rs367921493 Chr.6: 13595579-13595579 on Build GRCh38
rs188775956 Chr.6: 13595515-13595515 on Build GRCh38
rs199972686 Chr.6: 13595526-13595526 on Build GRCh38
rs756503724 Chr.6: 13595508-13595508 on Build GRCh38
rs372612782 Chr.6: 13595539-13595539 on Build GRCh38
rs751655170 Chr.6: 13595477-13595477 on Build GRCh38
rs746112544 Chr.6: 13595499-13595499 on Build GRCh38
rs201959618 Chr.6: 13595494-13595494 on Build GRCh38
rs200137124 Chr.6: 13595461-13595461 on Build GRCh38
rs752590753 Chr.6: 13595462-13595462 on Build GRCh38
rs199854042 Chr.6: 13595512-13595512 on Build GRCh38
rs1001873641 Chr.6: 13595436-13595436 on Build GRCh38
rs746543865 Chr.6: 13595525-13595525 on Build GRCh38
rs772778064 Chr.6: 13595521-13595521 on Build GRCh38
rs758280349 Chr.6: 13595465-13595465 on Build GRCh38
rs115296180 Chr.6: 13595439-13595439 on Build GRCh38
rs759376343 Chr.6: 13595459-13595459 on Build GRCh38
rs779340217 Chr.6: 13595606-13595606 on Build GRCh38
rs767734150 Chr.6: 13595563-13595563 on Build GRCh38
rs200742589 Chr.6: 13595488-13595488 on Build GRCh38
rs201704328 Chr.6: 13595545-13595545 on Build GRCh38
rs145972102 Chr.6: 13595533-13595533 on Build GRCh38
rs755482895 Chr.6: 13595592-13595592 on Build GRCh38
rs757468533 Chr.6: 13595490-13595490 on Build GRCh38
rs148020996 Chr.6: 13595518-13595518 on Build GRCh38
rs764220010 Chr.6: 13595476-13595476 on Build GRCh38
rs982395523 Chr.6: 13595435-13595435 on Build GRCh38

View Full Product Details