Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36306819-36307046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3D21
Entrez Gene ID
79729
Gene Name
SH3 domain containing 21
Gene Aliases
C1orf113
UniGene
Hs.524496
Gene Chromosome Location
Chr.1: 36306348-36329221 on Build GRCh38
Transcripts
NM_001162530.1 XM_011542153.2 XM_017002340.1 XM_017002341.1 XM_017002342.1 XM_017002343.1 XM_017002344.1 XM_017002345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372313 c.167T>A p.I56N Chr.1: 36306846-36306846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542911105 Chr.1: 36306860-36306860 on Build GRCh38
rs772038214 Chr.1: 36306940-36306940 on Build GRCh38
rs933938682 Chr.1: 36307010-36307011 on Build GRCh38
rs1054459617 Chr.1: 36306968-36306968 on Build GRCh38
rs1007685079 Chr.1: 36306864-36306864 on Build GRCh38
rs1033021694 Chr.1: 36306862-36306862 on Build GRCh38
rs915653240 Chr.1: 36306942-36306942 on Build GRCh38
rs527283117 Chr.1: 36306837-36306837 on Build GRCh38
rs971003869 Chr.1: 36306947-36306947 on Build GRCh38
rs780806602 Chr.1: 36306920-36306920 on Build GRCh38
rs376447501 Chr.1: 36306964-36306964 on Build GRCh38
rs950950819 Chr.1: 36306994-36306994 on Build GRCh38
rs562985572 Chr.1: 36306936-36306936 on Build GRCh38
rs903102372 Chr.1: 36306859-36306859 on Build GRCh38
rs966205694 Chr.1: 36306886-36306886 on Build GRCh38
rs769638344 Chr.1: 36306919-36306919 on Build GRCh38
rs549696371 Chr.1: 36306948-36306948 on Build GRCh38
rs1051748087 Chr.1: 36306914-36306914 on Build GRCh38
rs997612703 Chr.1: 36306988-36306988 on Build GRCh38
rs976225246 Chr.1: 36306891-36306891 on Build GRCh38
rs1018183454 Chr.1: 36306877-36306877 on Build GRCh38
rs1017650289 Chr.1: 36306981-36306981 on Build GRCh38
rs774381691 Chr.1: 36306924-36306924 on Build GRCh38
rs982400921 Chr.1: 36306932-36306932 on Build GRCh38
rs371697252 Chr.1: 36306933-36306933 on Build GRCh38
rs1045641864 Chr.1: 36306840-36306840 on Build GRCh38
rs921272243 Chr.1: 36307009-36307009 on Build GRCh38
rs1014578434 Chr.1: 36306838-36306838 on Build GRCh38
rs754958143 Chr.1: 36306903-36306903 on Build GRCh38
rs1045607105 Chr.1: 36306951-36306951 on Build GRCh38
rs561191354 Chr.1: 36306879-36306879 on Build GRCh38
rs934263334 Chr.1: 36306889-36306889 on Build GRCh38
rs1031984583 Chr.1: 36306966-36306966 on Build GRCh38
rs757787363 Chr.1: 36306865-36306865 on Build GRCh38
rs1026469159 Chr.1: 36306993-36306993 on Build GRCh38
rs112999732 Chr.1: 36307004-36307004 on Build GRCh38
rs934591203 Chr.1: 36306967-36306967 on Build GRCh38
rs946771396 Chr.1: 36306872-36306872 on Build GRCh38
rs957669462 Chr.1: 36306863-36306863 on Build GRCh38
rs898267764 Chr.1: 36306928-36306928 on Build GRCh38
rs533265638 Chr.1: 36306987-36306987 on Build GRCh38
rs1007982506 Chr.1: 36306978-36306978 on Build GRCh38
rs779684193 Chr.1: 36306868-36306868 on Build GRCh38
rs968166328 Chr.1: 36306992-36306992 on Build GRCh38
rs138952213 Chr.1: 36306916-36306916 on Build GRCh38
rs764799291 Chr.1: 36306970-36306970 on Build GRCh38
rs1051367608 Chr.1: 36307015-36307015 on Build GRCh38

View Full Product Details