Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57926498-57926749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF418
Entrez Gene ID
147686
Gene Name
zinc finger protein 418
Gene Aliases
-
UniGene
Hs.660728
Gene Chromosome Location
Chr.19: 57921882-57935404 on Build GRCh38
Transcripts
NM_001317027.1 NM_001317028.1 NM_001317029.1 NM_001317030.1 NM_133460.2 XM_017026304.1 XM_017026305.1 XM_017026306.1 XM_017026307.1 XM_017026308.1 XM_017026309.1 XM_017026310.1 XM_017026311.1 XM_017026312.1 XM_017026313.1 XM_017026314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
418748 c.1498C>T p.H500Y Chr.19: 57926683-57926683 on Build GRCh38
4817679 c.1678C>T p.H560Y Chr.19: 57926503-57926503 on Build GRCh38
440525 c.1493G>A p.R498H Chr.19: 57926688-57926688 on Build GRCh38
4835914 c.1469C>T p.S490L Chr.19: 57926712-57926712 on Build GRCh38
193822 c.1580G>A p.R527Q Chr.19: 57926601-57926601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760554720 Chr.19: 57926633-57926633 on Build GRCh38
rs775561982 Chr.19: 57926650-57926651 on Build GRCh38
rs754705254 Chr.19: 57926642-57926642 on Build GRCh38
rs775680787 Chr.19: 57926614-57926614 on Build GRCh38
rs762359439 Chr.19: 57926672-57926673 on Build GRCh38
rs867799450 Chr.19: 57926560-57926560 on Build GRCh38
rs776526938 Chr.19: 57926705-57926705 on Build GRCh38
rs752281408 Chr.19: 57926649-57926649 on Build GRCh38
rs755782877 Chr.19: 57926568-57926568 on Build GRCh38
rs151171553 Chr.19: 57926549-57926549 on Build GRCh38
rs149884051 Chr.19: 57926654-57926654 on Build GRCh38
rs766252059 Chr.19: 57926702-57926702 on Build GRCh38
rs373028676 Chr.19: 57926583-57926583 on Build GRCh38
rs779870015 Chr.19: 57926573-57926573 on Build GRCh38
rs773273464 Chr.19: 57926692-57926692 on Build GRCh38
rs753328465 Chr.19: 57926575-57926575 on Build GRCh38
rs778252629 Chr.19: 57926647-57926647 on Build GRCh38
rs535839531 Chr.19: 57926601-57926601 on Build GRCh38
rs746047456 Chr.19: 57926680-57926680 on Build GRCh38
rs759371951 Chr.19: 57926626-57926627 on Build GRCh38
rs979584949 Chr.19: 57926548-57926548 on Build GRCh38
rs771172922 Chr.19: 57926522-57926522 on Build GRCh38
rs201554779 Chr.19: 57926602-57926602 on Build GRCh38
rs761861989 Chr.19: 57926563-57926563 on Build GRCh38
rs769741818 Chr.19: 57926610-57926610 on Build GRCh38
rs374204768 Chr.19: 57926719-57926719 on Build GRCh38
rs775386760 Chr.19: 57926683-57926683 on Build GRCh38
rs543688252 Chr.19: 57926718-57926718 on Build GRCh38
rs759547468 Chr.19: 57926531-57926531 on Build GRCh38
rs770672131 Chr.19: 57926566-57926567 on Build GRCh38
rs757604706 Chr.19: 57926663-57926663 on Build GRCh38
rs764964252 Chr.19: 57926714-57926714 on Build GRCh38
rs781586376 Chr.19: 57926677-57926677 on Build GRCh38
rs769585723 Chr.19: 57926645-57926646 on Build GRCh38
rs750188839 Chr.19: 57926632-57926632 on Build GRCh38
rs878906163 Chr.19: 57926602-57926604 on Build GRCh38
rs776430205 Chr.19: 57926579-57926580 on Build GRCh38
rs767189157 Chr.19: 57926564-57926564 on Build GRCh38
rs748339790 Chr.19: 57926693-57926693 on Build GRCh38
rs770088774 Chr.19: 57926682-57926682 on Build GRCh38
rs200769352 Chr.19: 57926686-57926686 on Build GRCh38
rs758780950 Chr.19: 57926651-57926651 on Build GRCh38
rs748579604 Chr.19: 57926590-57926590 on Build GRCh38
rs776642342 Chr.19: 57926530-57926530 on Build GRCh38
rs572483773 Chr.19: 57926608-57926608 on Build GRCh38
rs376873884 Chr.19: 57926688-57926688 on Build GRCh38
rs1002283430 Chr.19: 57926669-57926669 on Build GRCh38
rs868117890 Chr.19: 57926551-57926552 on Build GRCh38
rs896570474 Chr.19: 57926672-57926672 on Build GRCh38
rs146628723 Chr.19: 57926655-57926655 on Build GRCh38
rs200332982 Chr.19: 57926595-57926595 on Build GRCh38
rs1034144029 Chr.19: 57926671-57926671 on Build GRCh38
rs371520628 Chr.19: 57926689-57926689 on Build GRCh38
rs765424505 Chr.19: 57926538-57926538 on Build GRCh38
rs201513129 Chr.19: 57926636-57926636 on Build GRCh38
rs200035509 Chr.19: 57926596-57926596 on Build GRCh38
rs767588770 Chr.19: 57926628-57926628 on Build GRCh38
rs1036675571 Chr.19: 57926598-57926598 on Build GRCh38
rs140058497 Chr.19: 57926576-57926576 on Build GRCh38
rs748372712 Chr.19: 57926549-57926557 on Build GRCh38
rs759068633 Chr.19: 57926707-57926707 on Build GRCh38
rs772673747 Chr.19: 57926594-57926594 on Build GRCh38
rs374428351 Chr.19: 57926667-57926667 on Build GRCh38
rs201604700 Chr.19: 57926641-57926641 on Build GRCh38
rs369738449 Chr.19: 57926567-57926567 on Build GRCh38
rs761617650 Chr.19: 57926627-57926627 on Build GRCh38

View Full Product Details