Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67887122-67887347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51B
Entrez Gene ID
5890
Gene Name
RAD51 paralog B
Gene Aliases
R51H2, RAD51L1, REC2
UniGene
Hs.172587
Gene Chromosome Location
Chr.14: 67819779-68683118 on Build GRCh38
Transcripts
NM_001321809.1 NM_001321810.1 NM_001321812.1 NM_001321814.1 NM_001321815.1 NM_001321817.1 NM_001321818.1 NM_001321819.1 NM_001321821.1 NM_002877.5 NM_133509.3 NM_133510.3 XM_011537050.2 XM_011537051.2 XM_017021545.1 XM_017021546.1 XM_017021547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7125355 c.743A>C p.E248A Chr.14: 67887191-67887191 on Build GRCh38
7125358 c.743A>C p.E248A Chr.14: 67887191-67887191 on Build GRCh38
6923094 c.728A>G p.K243R Chr.14: 67887176-67887176 on Build GRCh38
3815169 c.751A>G p.I251V Chr.14: 67887199-67887199 on Build GRCh38
6906627 c.739G>A p.E247K Chr.14: 67887187-67887187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758105904 Chr.14: 67887205-67887205 on Build GRCh38
rs34594234 Chr.14: 67887176-67887176 on Build GRCh38
rs17104744 Chr.14: 67887162-67887162 on Build GRCh38
rs762561329 Chr.14: 67887238-67887238 on Build GRCh38
rs746672778 Chr.14: 67887226-67887226 on Build GRCh38
rs34839595 Chr.14: 67887214-67887215 on Build GRCh38
rs761312484 Chr.14: 67887231-67887231 on Build GRCh38
rs769044305 Chr.14: 67887214-67887214 on Build GRCh38
rs1050559111 Chr.14: 67887267-67887267 on Build GRCh38
rs780243413 Chr.14: 67887203-67887203 on Build GRCh38
rs750320583 Chr.14: 67887315-67887315 on Build GRCh38
rs763766656 Chr.14: 67887244-67887244 on Build GRCh38
rs749503390 Chr.14: 67887208-67887208 on Build GRCh38
rs774477051 Chr.14: 67887303-67887303 on Build GRCh38
rs373016834 Chr.14: 67887217-67887217 on Build GRCh38
rs775115775 Chr.14: 67887236-67887236 on Build GRCh38
rs751395885 Chr.14: 67887156-67887156 on Build GRCh38
rs779712754 Chr.14: 67887217-67887225 on Build GRCh38
rs772651147 Chr.14: 67887219-67887219 on Build GRCh38
rs187955360 Chr.14: 67887183-67887183 on Build GRCh38
rs768145967 Chr.14: 67887245-67887247 on Build GRCh38
rs764641310 Chr.14: 67887233-67887233 on Build GRCh38
rs910009385 Chr.14: 67887288-67887288 on Build GRCh38
rs757055448 Chr.14: 67887253-67887253 on Build GRCh38
rs751304174 Chr.14: 67887248-67887248 on Build GRCh38
rs984172290 Chr.14: 67887283-67887283 on Build GRCh38
rs34175580 Chr.14: 67887222-67887222 on Build GRCh38
rs543824825 Chr.14: 67887273-67887273 on Build GRCh38
rs200290179 Chr.14: 67887218-67887218 on Build GRCh38
rs33929366 Chr.14: 67887196-67887196 on Build GRCh38

View Full Product Details