Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16417542-16417771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPV2
Entrez Gene ID
51393
Gene Name
transient receptor potential cation channel subfamily V member 2
Gene Aliases
VRL, VRL-1, VRL1
UniGene
Hs.279746
Gene Chromosome Location
Chr.17: 16415542-16437003 on Build GRCh38
Transcripts
NM_016113.4 XM_005256676.1 XM_005256677.1 XM_005256678.4 XM_006721541.3 XM_006721543.3 XM_011523922.1 XM_011523923.1 XM_017024730.1 XM_017024731.1 XM_017024732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
975901 c.17G>A p.S6N Chr.17: 16417685-16417685 on Build GRCh38
1128817 c.72G>T p.E24D Chr.17: 16417740-16417740 on Build GRCh38
148200 c.50G>C p.G17A Chr.17: 16417718-16417718 on Build GRCh38
326778 c.70G>C p.E24Q Chr.17: 16417738-16417738 on Build GRCh38
3672359 c.21T>C p.S7S Chr.17: 16417689-16417689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780932240 Chr.17: 16417726-16417726 on Build GRCh38
rs202004299 Chr.17: 16417648-16417648 on Build GRCh38
rs771385099 Chr.17: 16417667-16417667 on Build GRCh38
rs201198733 Chr.17: 16417654-16417654 on Build GRCh38
rs537447669 Chr.17: 16417701-16417701 on Build GRCh38
rs189163490 Chr.17: 16417724-16417724 on Build GRCh38
rs3813768 Chr.17: 16417718-16417718 on Build GRCh38
rs532591531 Chr.17: 16417585-16417585 on Build GRCh38
rs747616843 Chr.17: 16417620-16417620 on Build GRCh38
rs772637396 Chr.17: 16417747-16417747 on Build GRCh38
rs200065409 Chr.17: 16417626-16417626 on Build GRCh38
rs980409404 Chr.17: 16417703-16417703 on Build GRCh38
rs758488359 Chr.17: 16417586-16417586 on Build GRCh38
rs138582403 Chr.17: 16417587-16417587 on Build GRCh38
rs763862235 Chr.17: 16417629-16417629 on Build GRCh38
rs192408615 Chr.17: 16417678-16417678 on Build GRCh38
rs775805678 Chr.17: 16417749-16417749 on Build GRCh38
rs775635605 Chr.17: 16417681-16417681 on Build GRCh38
rs149389070 Chr.17: 16417680-16417680 on Build GRCh38
rs370060704 Chr.17: 16417743-16417743 on Build GRCh38
rs750417395 Chr.17: 16417711-16417711 on Build GRCh38
rs758323970 Chr.17: 16417712-16417712 on Build GRCh38
rs11657598 Chr.17: 16417664-16417664 on Build GRCh38
rs778851143 Chr.17: 16417641-16417641 on Build GRCh38
rs753965667 Chr.17: 16417694-16417694 on Build GRCh38
rs186827127 Chr.17: 16417601-16417601 on Build GRCh38
rs369164734 Chr.17: 16417656-16417656 on Build GRCh38
rs372811311 Chr.17: 16417624-16417624 on Build GRCh38
rs371408651 Chr.17: 16417663-16417663 on Build GRCh38
rs140791443 Chr.17: 16417710-16417710 on Build GRCh38
rs749696148 Chr.17: 16417665-16417665 on Build GRCh38
rs747774378 Chr.17: 16417737-16417737 on Build GRCh38
rs775043213 Chr.17: 16417669-16417669 on Build GRCh38
rs962516249 Chr.17: 16417591-16417591 on Build GRCh38
rs534505924 Chr.17: 16417605-16417605 on Build GRCh38
rs771475328 Chr.17: 16417742-16417742 on Build GRCh38
rs1043632075 Chr.17: 16417707-16417707 on Build GRCh38
rs778067792 Chr.17: 16417637-16417637 on Build GRCh38
rs769494174 Chr.17: 16417653-16417654 on Build GRCh38
rs779784939 Chr.17: 16417713-16417713 on Build GRCh38
rs112440901 Chr.17: 16417683-16417683 on Build GRCh38
rs368892718 Chr.17: 16417582-16417582 on Build GRCh38
rs113850084 Chr.17: 16417647-16417647 on Build GRCh38
rs528539142 Chr.17: 16417633-16417633 on Build GRCh38
rs752978675 Chr.17: 16417584-16417584 on Build GRCh38
rs368492158 Chr.17: 16417632-16417632 on Build GRCh38
rs371881034 Chr.17: 16417644-16417644 on Build GRCh38
rs761634679 Chr.17: 16417700-16417700 on Build GRCh38
rs778953873 Chr.17: 16417581-16417581 on Build GRCh38
rs768356620 Chr.17: 16417657-16417657 on Build GRCh38
rs375556607 Chr.17: 16417746-16417746 on Build GRCh38
rs760606289 Chr.17: 16417692-16417692 on Build GRCh38
rs558249672 Chr.17: 16417628-16417628 on Build GRCh38

View Full Product Details