Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50074145-50074376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
UniGene
Hs.265829
Gene Chromosome Location
Chr.17: 50055968-50090485 on Build GRCh38
Transcripts
NM_002204.3 XM_005257308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4960163 c.1255G>A p.G419R Chr.17: 50074153-50074153 on Build GRCh38
2698768 c.1382G>A p.R461Q Chr.17: 50074280-50074280 on Build GRCh38
2698767 c.1382G>A p.R461Q Chr.17: 50074280-50074280 on Build GRCh38
1384153 c.1252C>A p.H418N Chr.17: 50074150-50074150 on Build GRCh38
6317492 c.1266G>T p.L422L Chr.17: 50074164-50074164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780510840 Chr.17: 50074245-50074245 on Build GRCh38
rs373427028 Chr.17: 50074316-50074316 on Build GRCh38
rs766943378 Chr.17: 50074307-50074307 on Build GRCh38
rs565778216 Chr.17: 50074167-50074167 on Build GRCh38
rs760837392 Chr.17: 50074291-50074291 on Build GRCh38
rs186943165 Chr.17: 50074354-50074354 on Build GRCh38
rs376879376 Chr.17: 50074297-50074297 on Build GRCh38
rs61730084 Chr.17: 50074247-50074247 on Build GRCh38
rs776970761 Chr.17: 50074294-50074294 on Build GRCh38
rs78768954 Chr.17: 50074188-50074188 on Build GRCh38
rs770703080 Chr.17: 50074267-50074267 on Build GRCh38
rs765745707 Chr.17: 50074197-50074197 on Build GRCh38
rs1016251016 Chr.17: 50074211-50074211 on Build GRCh38
rs775640037 Chr.17: 50074180-50074180 on Build GRCh38
rs758949436 Chr.17: 50074233-50074233 on Build GRCh38
rs543346479 Chr.17: 50074296-50074296 on Build GRCh38
rs757664252 Chr.17: 50074193-50074193 on Build GRCh38
rs576406968 Chr.17: 50074345-50074345 on Build GRCh38
rs976168995 Chr.17: 50074269-50074269 on Build GRCh38
rs763321144 Chr.17: 50074300-50074300 on Build GRCh38
rs963624366 Chr.17: 50074268-50074268 on Build GRCh38
rs575130011 Chr.17: 50074184-50074184 on Build GRCh38
rs754213910 Chr.17: 50074189-50074189 on Build GRCh38
rs1004901363 Chr.17: 50074202-50074202 on Build GRCh38
rs755684284 Chr.17: 50074248-50074248 on Build GRCh38
rs144049965 Chr.17: 50074283-50074283 on Build GRCh38
rs777452976 Chr.17: 50074256-50074256 on Build GRCh38
rs556513125 Chr.17: 50074336-50074336 on Build GRCh38
rs554367981 Chr.17: 50074261-50074261 on Build GRCh38
rs750952837 Chr.17: 50074209-50074209 on Build GRCh38
rs375994775 Chr.17: 50074330-50074330 on Build GRCh38
rs766122103 Chr.17: 50074314-50074316 on Build GRCh38
rs966457107 Chr.17: 50074334-50074334 on Build GRCh38
rs772057144 Chr.17: 50074280-50074280 on Build GRCh38
rs768657346 Chr.17: 50074293-50074293 on Build GRCh38
rs151126162 Chr.17: 50074276-50074276 on Build GRCh38
rs368943810 Chr.17: 50074298-50074298 on Build GRCh38
rs752048192 Chr.17: 50074326-50074326 on Build GRCh38

View Full Product Details