Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154584454-154584713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGA
Entrez Gene ID
2243
Gene Name
fibrinogen alpha chain
Gene Aliases
Fib2
UniGene
Hs.351593
Gene Chromosome Location
Chr.4: 154583126-154590766 on Build GRCh38
Transcripts
NM_000508.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934054 c.2226C>G p.F742L Chr.4: 154584499-154584499 on Build GRCh38
5808284 c.2189A>T p.D730V Chr.4: 154584536-154584536 on Build GRCh38
4475917 c.2025C>T p.I675I Chr.4: 154584700-154584700 on Build GRCh38
3946089 c.2209T>C p.Y737H Chr.4: 154584516-154584516 on Build GRCh38
7170347 c.2025C>G p.I675M Chr.4: 154584700-154584700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753284556 Chr.4: 154584669-154584670 on Build GRCh38
rs766941271 Chr.4: 154584581-154584581 on Build GRCh38
rs752055822 Chr.4: 154584671-154584671 on Build GRCh38
rs752141650 Chr.4: 154584583-154584583 on Build GRCh38
rs779930407 Chr.4: 154584644-154584644 on Build GRCh38
rs781434450 Chr.4: 154584503-154584503 on Build GRCh38
rs764795081 Chr.4: 154584575-154584575 on Build GRCh38
rs201769249 Chr.4: 154584498-154584498 on Build GRCh38
rs777521200 Chr.4: 154584476-154584476 on Build GRCh38
rs61731299 Chr.4: 154584540-154584540 on Build GRCh38
rs761425560 Chr.4: 154584552-154584552 on Build GRCh38
rs760416652 Chr.4: 154584536-154584536 on Build GRCh38
rs965539028 Chr.4: 154584517-154584517 on Build GRCh38
rs1024393205 Chr.4: 154584637-154584637 on Build GRCh38
rs761492158 Chr.4: 154584658-154584658 on Build GRCh38
rs982383204 Chr.4: 154584657-154584657 on Build GRCh38
rs368008640 Chr.4: 154584481-154584481 on Build GRCh38
rs376434823 Chr.4: 154584499-154584499 on Build GRCh38
rs781425357 Chr.4: 154584592-154584592 on Build GRCh38
rs373841420 Chr.4: 154584576-154584576 on Build GRCh38
rs745804282 Chr.4: 154584518-154584518 on Build GRCh38
rs369692720 Chr.4: 154584539-154584539 on Build GRCh38
rs754678681 Chr.4: 154584504-154584504 on Build GRCh38
rs904385780 Chr.4: 154584608-154584608 on Build GRCh38
rs185920746 Chr.4: 154584674-154584674 on Build GRCh38
rs999884352 Chr.4: 154584609-154584609 on Build GRCh38
rs1007440681 Chr.4: 154584549-154584549 on Build GRCh38
rs769855583 Chr.4: 154584505-154584505 on Build GRCh38
rs772041960 Chr.4: 154584531-154584531 on Build GRCh38
rs372219583 Chr.4: 154584497-154584497 on Build GRCh38
rs763532423 Chr.4: 154584578-154584578 on Build GRCh38
rs762511080 Chr.4: 154584666-154584666 on Build GRCh38
rs771023837 Chr.4: 154584636-154584636 on Build GRCh38
rs773021324 Chr.4: 154584665-154584665 on Build GRCh38
rs148411201 Chr.4: 154584622-154584622 on Build GRCh38
rs748014488 Chr.4: 154584568-154584568 on Build GRCh38
rs1031411881 Chr.4: 154584619-154584619 on Build GRCh38
rs753047358 Chr.4: 154584681-154584681 on Build GRCh38
rs769380836 Chr.4: 154584659-154584659 on Build GRCh38
rs752951662 Chr.4: 154584601-154584601 on Build GRCh38
rs866001734 Chr.4: 154584676-154584676 on Build GRCh38
rs776352963 Chr.4: 154584650-154584650 on Build GRCh38
rs929349465 Chr.4: 154584492-154584492 on Build GRCh38
rs34985782 Chr.4: 154584678-154584678 on Build GRCh38
rs777775053 Chr.4: 154584508-154584508 on Build GRCh38
rs775246327 Chr.4: 154584534-154584534 on Build GRCh38
rs768481407 Chr.4: 154584649-154584649 on Build GRCh38
rs564725896 Chr.4: 154584621-154584621 on Build GRCh38
rs868002890 Chr.4: 154584683-154584683 on Build GRCh38
rs780806928 Chr.4: 154584523-154584523 on Build GRCh38
rs755329084 Chr.4: 154584590-154584590 on Build GRCh38
rs753629473 Chr.4: 154584498-154584498 on Build GRCh38
rs763664594 Chr.4: 154584662-154584662 on Build GRCh38

View Full Product Details