Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25773228-25773495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A1
Entrez Gene ID
6568
Gene Name
solute carrier family 17 member 1
Gene Aliases
NAPI-1, NPT-1, NPT1
UniGene
Hs.205816
Gene Chromosome Location
Chr.6: 25723744-25832108 on Build GRCh38
Transcripts
XM_017011201.1 NM_001286121.1 NM_005495.2 XM_011514211.1 XM_011514213.2 XM_011514214.2 XM_011514215.2 XM_011514217.2 XM_011514218.2 XM_011514219.2 XM_017010150.1 XM_017010151.1

Gene Symbol
SLC17A4
Entrez Gene ID
10050
Gene Name
solute carrier family 17 member 4
Gene Aliases
KAIA2138
UniGene
Hs.282931
Gene Chromosome Location
Chr.6: 25754673-25781175 on Build GRCh38
Transcripts
XM_017011201.1 NM_001286121.1 NM_005495.2 XM_011514211.1 XM_011514213.2 XM_011514214.2 XM_011514215.2 XM_011514217.2 XM_011514218.2 XM_011514219.2 XM_017010150.1 XM_017010151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596046 c.743C>T p.P248L Chr.6: 25773311-25773311 on Build GRCh38
6779888 c.800A>C p.Y267S Chr.6: 25773368-25773368 on Build GRCh38
1265722 c.745C>T p.L249F Chr.6: 25773313-25773313 on Build GRCh38
1754676 c.799T>A p.Y267N Chr.6: 25773367-25773367 on Build GRCh38
7286715 c.731C>T p.P244L Chr.6: 25773299-25773299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745477612 Chr.6: 25773356-25773356 on Build GRCh38
rs866570216 Chr.6: 25773275-25773275 on Build GRCh38
rs549918283 Chr.6: 25773383-25773383 on Build GRCh38
rs1039146744 Chr.6: 25773438-25773438 on Build GRCh38
rs780130824 Chr.6: 25773274-25773274 on Build GRCh38
rs757104412 Chr.6: 25773256-25773256 on Build GRCh38
rs989493713 Chr.6: 25773264-25773264 on Build GRCh38
rs943545693 Chr.6: 25773417-25773417 on Build GRCh38
rs996979005 Chr.6: 25773398-25773398 on Build GRCh38
rs775248986 Chr.6: 25773369-25773369 on Build GRCh38
rs767281369 Chr.6: 25773258-25773258 on Build GRCh38
rs767283000 Chr.6: 25773399-25773399 on Build GRCh38
rs772417110 Chr.6: 25773334-25773334 on Build GRCh38
rs966715806 Chr.6: 25773371-25773371 on Build GRCh38
rs778973307 Chr.6: 25773467-25773467 on Build GRCh38
rs756302339 Chr.6: 25773266-25773266 on Build GRCh38
rs200697427 Chr.6: 25773367-25773367 on Build GRCh38
rs761845040 Chr.6: 25773381-25773381 on Build GRCh38
rs755086812 Chr.6: 25773287-25773287 on Build GRCh38
rs762529750 Chr.6: 25773373-25773373 on Build GRCh38
rs763754920 Chr.6: 25773374-25773374 on Build GRCh38
rs754104302 Chr.6: 25773434-25773434 on Build GRCh38
rs750204498 Chr.6: 25773409-25773409 on Build GRCh38
rs899874919 Chr.6: 25773269-25773269 on Build GRCh38
rs755050187 Chr.6: 25773439-25773439 on Build GRCh38
rs192246809 Chr.6: 25773377-25773377 on Build GRCh38
rs201472019 Chr.6: 25773277-25773277 on Build GRCh38
rs773465799 Chr.6: 25773354-25773354 on Build GRCh38
rs545026310 Chr.6: 25773320-25773320 on Build GRCh38
rs116471288 Chr.6: 25773263-25773263 on Build GRCh38
rs760908659 Chr.6: 25773413-25773413 on Build GRCh38
rs563340494 Chr.6: 25773425-25773425 on Build GRCh38
rs370664049 Chr.6: 25773324-25773324 on Build GRCh38

View Full Product Details