Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54806034-54806273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIR2DL4
Entrez Gene ID
3805
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4
Gene Aliases
CD158D, G9P, KIR, KIR-103AS, KIR-2DL4, KIR103, KIR103AS
UniGene
Hs.661219
Gene Chromosome Location
Chr.19: 54803612-54814517 on Build GRCh38
Transcripts
NM_001080770.1 NM_001080772.1 NM_002255.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296303 c.626C>T p.P209L Chr.19: 54806215-54806215 on Build GRCh38
1396372 c.480delG p.E162fs*83 Chr.19: 54806069-54806069 on Build GRCh38
217178 c.497T>G p.L166R Chr.19: 54806086-54806086 on Build GRCh38
5505992 c.458A>G p.D153G Chr.19: 54806047-54806047 on Build GRCh38
5349384 c.491T>G p.L164R Chr.19: 54806086-54806086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049277022 Chr.19: 54806198-54806198 on Build GRCh38
rs1048721503 Chr.19: 54806099-54806099 on Build GRCh38
rs796093143 Chr.19: 54806074-54806074 on Build GRCh38
rs577091862 Chr.19: 54806126-54806126 on Build GRCh38
rs751766410 Chr.19: 54806191-54806191 on Build GRCh38
rs796666015 Chr.19: 54806075-54806075 on Build GRCh38
rs954023147 Chr.19: 54806129-54806129 on Build GRCh38
rs996706860 Chr.19: 54806113-54806113 on Build GRCh38
rs540514355 Chr.19: 54806073-54806073 on Build GRCh38
rs1028119455 Chr.19: 54806114-54806114 on Build GRCh38
rs886411212 Chr.19: 54806068-54806068 on Build GRCh38
rs796888355 Chr.19: 54806112-54806112 on Build GRCh38
rs1022240407 Chr.19: 54806136-54806136 on Build GRCh38
rs1051456 Chr.19: 54806214-54806214 on Build GRCh38
rs878977590 Chr.19: 54806201-54806201 on Build GRCh38
rs749007396 Chr.19: 54806085-54806086 on Build GRCh38
rs927654415 Chr.19: 54806150-54806150 on Build GRCh38
rs374413343 Chr.19: 54806069-54806069 on Build GRCh38
rs546044168 Chr.19: 54806130-54806130 on Build GRCh38
rs553817989 Chr.19: 54806095-54806095 on Build GRCh38
rs373028201 Chr.19: 54806230-54806230 on Build GRCh38
rs1040375417 Chr.19: 54806216-54806216 on Build GRCh38
rs1001230224 Chr.19: 54806171-54806171 on Build GRCh38
rs977740939 Chr.19: 54806142-54806142 on Build GRCh38
rs903729815 Chr.19: 54806170-54806170 on Build GRCh38
rs956068523 Chr.19: 54806188-54806188 on Build GRCh38
rs562937136 Chr.19: 54806178-54806178 on Build GRCh38
rs749648141 Chr.19: 54806061-54806061 on Build GRCh38
rs901846164 Chr.19: 54806238-54806238 on Build GRCh38
rs771691398 Chr.19: 54806065-54806065 on Build GRCh38
rs112694450 Chr.19: 54806086-54806086 on Build GRCh38
rs530171318 Chr.19: 54806215-54806215 on Build GRCh38
rs878920557 Chr.19: 54806204-54806204 on Build GRCh38
rs781227427 Chr.19: 54806240-54806240 on Build GRCh38
rs200435373 Chr.19: 54806109-54806109 on Build GRCh38
rs772895115 Chr.19: 54806177-54806177 on Build GRCh38
rs762659354 Chr.19: 54806157-54806157 on Build GRCh38
rs765986548 Chr.19: 54806076-54806076 on Build GRCh38
rs752494082 Chr.19: 54806139-54806139 on Build GRCh38
rs1049307899 Chr.19: 54806064-54806064 on Build GRCh38
rs916719545 Chr.19: 54806160-54806160 on Build GRCh38
rs879223877 Chr.19: 54806156-54806156 on Build GRCh38
rs901911372 Chr.19: 54806106-54806106 on Build GRCh38

View Full Product Details