Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35009941-35010214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGXT2
Entrez Gene ID
64902
Gene Name
alanine--glyoxylate aminotransferase 2
Gene Aliases
AGT2, DAIBAT
UniGene
Hs.34494
Gene Chromosome Location
Chr.5: 34998101-35048188 on Build GRCh38
Transcripts
NM_001306173.1 NM_031900.3 XM_005248337.2 XM_005248338.2 XM_017009748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
592453 c.1295G>A p.R432Q Chr.5: 35010043-35010043 on Build GRCh38
7424166 c.1286G>C p.G429A Chr.5: 35010052-35010052 on Build GRCh38
5589590 c.1267G>A p.D423N Chr.5: 35010071-35010071 on Build GRCh38
6596550 c.1270G>T p.E424* Chr.5: 35010068-35010068 on Build GRCh38
5934936 c.1231G>A p.G411R Chr.5: 35010107-35010107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758630572 Chr.5: 35010048-35010048 on Build GRCh38
rs752461972 Chr.5: 35009970-35009970 on Build GRCh38
rs745927049 Chr.5: 35010090-35010090 on Build GRCh38
rs760390165 Chr.5: 35009976-35009976 on Build GRCh38
rs776375099 Chr.5: 35010042-35010042 on Build GRCh38
rs781719128 Chr.5: 35010009-35010009 on Build GRCh38
rs759563019 Chr.5: 35010025-35010025 on Build GRCh38
rs776264048 Chr.5: 35010106-35010106 on Build GRCh38
rs768483818 Chr.5: 35010185-35010185 on Build GRCh38
rs142368612 Chr.5: 35010047-35010047 on Build GRCh38
rs1013635681 Chr.5: 35010018-35010018 on Build GRCh38
rs771996715 Chr.5: 35010027-35010027 on Build GRCh38
rs761319044 Chr.5: 35010043-35010043 on Build GRCh38
rs146075538 Chr.5: 35010012-35010012 on Build GRCh38
rs755414251 Chr.5: 35010006-35010006 on Build GRCh38
rs771065822 Chr.5: 35010022-35010022 on Build GRCh38
rs772084689 Chr.5: 35010091-35010091 on Build GRCh38
rs370870851 Chr.5: 35010019-35010019 on Build GRCh38
rs1047952870 Chr.5: 35009972-35009972 on Build GRCh38
rs752126134 Chr.5: 35010005-35010005 on Build GRCh38
rs753433403 Chr.5: 35009990-35009990 on Build GRCh38
rs151278208 Chr.5: 35010055-35010055 on Build GRCh38
rs775414339 Chr.5: 35010094-35010094 on Build GRCh38
rs368570551 Chr.5: 35010035-35010035 on Build GRCh38
rs763102408 Chr.5: 35009973-35009974 on Build GRCh38
rs566706169 Chr.5: 35010096-35010096 on Build GRCh38
rs150037067 Chr.5: 35010149-35010149 on Build GRCh38
rs764792216 Chr.5: 35010113-35010113 on Build GRCh38
rs1006208255 Chr.5: 35009996-35009996 on Build GRCh38
rs376158578 Chr.5: 35010073-35010073 on Build GRCh38
rs200965634 Chr.5: 35010174-35010174 on Build GRCh38
rs749869447 Chr.5: 35010050-35010050 on Build GRCh38
rs756793628 Chr.5: 35009992-35009992 on Build GRCh38
rs895227113 Chr.5: 35010016-35010016 on Build GRCh38
rs866461623 Chr.5: 35010044-35010044 on Build GRCh38
rs780057421 Chr.5: 35010181-35010181 on Build GRCh38
rs756667430 Chr.5: 35010144-35010144 on Build GRCh38
rs373871490 Chr.5: 35010023-35010023 on Build GRCh38
rs553649751 Chr.5: 35010074-35010074 on Build GRCh38
rs768459704 Chr.5: 35010098-35010098 on Build GRCh38
rs766316919 Chr.5: 35010120-35010121 on Build GRCh38
rs772835644 Chr.5: 35010120-35010120 on Build GRCh38
rs1003634950 Chr.5: 35010080-35010080 on Build GRCh38
rs139344660 Chr.5: 35010067-35010067 on Build GRCh38
rs199631717 Chr.5: 35010069-35010069 on Build GRCh38
rs199953023 Chr.5: 35010056-35010056 on Build GRCh38
rs746039390 Chr.5: 35010175-35010175 on Build GRCh38
rs971189650 Chr.5: 35010037-35010037 on Build GRCh38
rs758502169 Chr.5: 35010176-35010176 on Build GRCh38
rs757561316 Chr.5: 35010150-35010150 on Build GRCh38
rs368638991 Chr.5: 35010125-35010125 on Build GRCh38
rs761563619 Chr.5: 35010110-35010110 on Build GRCh38
rs775324605 Chr.5: 35010030-35010030 on Build GRCh38
rs200532818 Chr.5: 35010128-35010128 on Build GRCh38
rs548763349 Chr.5: 35010010-35010010 on Build GRCh38
rs199863290 Chr.5: 35010071-35010071 on Build GRCh38
rs975770624 Chr.5: 35010143-35010143 on Build GRCh38
rs767349408 Chr.5: 35009969-35009969 on Build GRCh38
rs768546156 Chr.5: 35010187-35010187 on Build GRCh38
rs466067 Chr.5: 35010033-35010033 on Build GRCh38
rs375641300 Chr.5: 35009985-35009985 on Build GRCh38
rs753196170 Chr.5: 35010066-35010066 on Build GRCh38
rs749432935 Chr.5: 35010021-35010021 on Build GRCh38
rs778244122 Chr.5: 35010000-35010000 on Build GRCh38

View Full Product Details