Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62786301-62786550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF6L
Entrez Gene ID
10629
Gene Name
TATA-box binding protein associated factor 6 like
Gene Aliases
PAF65A
UniGene
Hs.714400
Gene Chromosome Location
Chr.11: 62771303-62787342 on Build GRCh38
Transcripts
NM_006473.3 XM_005273714.1 XM_017017100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4854469 c.1068C>G p.H356Q Chr.11: 62786367-62786367 on Build GRCh38
3791717 c.1074C>T p.V358V Chr.11: 62786373-62786373 on Build GRCh38
4034892 c.1121C>G p.A374G Chr.11: 62786548-62786548 on Build GRCh38
929818 c.1019T>C p.V340A Chr.11: 62786318-62786318 on Build GRCh38
5798586 c.1097T>C p.V366A Chr.11: 62786524-62786524 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777636663 Chr.11: 62786474-62786474 on Build GRCh38
rs917436655 Chr.11: 62786493-62786493 on Build GRCh38
rs759034810 Chr.11: 62786424-62786424 on Build GRCh38
rs767946917 Chr.11: 62786413-62786413 on Build GRCh38
rs774533909 Chr.11: 62786403-62786403 on Build GRCh38
rs890636739 Chr.11: 62786500-62786500 on Build GRCh38
rs779960954 Chr.11: 62786529-62786529 on Build GRCh38
rs377546066 Chr.11: 62786396-62786396 on Build GRCh38
rs746963714 Chr.11: 62786355-62786355 on Build GRCh38
rs747952166 Chr.11: 62786322-62786322 on Build GRCh38
rs140375142 Chr.11: 62786352-62786352 on Build GRCh38
rs757328404 Chr.11: 62786505-62786505 on Build GRCh38
rs757938663 Chr.11: 62786473-62786473 on Build GRCh38
rs370454121 Chr.11: 62786430-62786431 on Build GRCh38
rs143657600 Chr.11: 62786455-62786455 on Build GRCh38
rs776852452 Chr.11: 62786367-62786367 on Build GRCh38
rs955812235 Chr.11: 62786467-62786467 on Build GRCh38
rs781258697 Chr.11: 62786510-62786510 on Build GRCh38
rs753414777 Chr.11: 62786423-62786423 on Build GRCh38
rs932061279 Chr.11: 62786490-62786490 on Build GRCh38
rs373387773 Chr.11: 62786394-62786394 on Build GRCh38
rs374737675 Chr.11: 62786331-62786331 on Build GRCh38
rs12514 Chr.11: 62786359-62786359 on Build GRCh38
rs528645704 Chr.11: 62786489-62786489 on Build GRCh38
rs201909306 Chr.11: 62786386-62786386 on Build GRCh38
rs369063031 Chr.11: 62786391-62786391 on Build GRCh38
rs777391386 Chr.11: 62786346-62786346 on Build GRCh38
rs769890556 Chr.11: 62786526-62786526 on Build GRCh38
rs373550641 Chr.11: 62786470-62786470 on Build GRCh38
rs968416362 Chr.11: 62786430-62786430 on Build GRCh38
rs893929357 Chr.11: 62786484-62786484 on Build GRCh38
rs764847438 Chr.11: 62786468-62786468 on Build GRCh38
rs761915976 Chr.11: 62786494-62786494 on Build GRCh38
rs1031290876 Chr.11: 62786465-62786465 on Build GRCh38
rs764913619 Chr.11: 62786510-62786513 on Build GRCh38
rs371168609 Chr.11: 62786406-62786406 on Build GRCh38
rs978364530 Chr.11: 62786440-62786440 on Build GRCh38
rs770933367 Chr.11: 62786363-62786363 on Build GRCh38
rs759162420 Chr.11: 62786492-62786495 on Build GRCh38
rs754174702 Chr.11: 62786464-62786464 on Build GRCh38

View Full Product Details