Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57096735-57097006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT6
Entrez Gene ID
6778
Gene Name
signal transducer and activator of transcription 6
Gene Aliases
D12S1644, IL-4-STAT, STAT6B, STAT6C
UniGene
Hs.524518
Gene Chromosome Location
Chr.12: 57095404-57111433 on Build GRCh38
Transcripts
NM_001178078.1 NM_001178079.1 NM_001178080.1 NM_001178081.1 NM_003153.4 NR_033659.1 XM_006719575.2 XM_011538703.2 XM_011538704.2 XM_011538705.2 XM_011538706.2 XM_011538707.2 XM_011538708.2 XM_011538709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6791722 c.2357T>G p.I786S Chr.12: 57096759-57096759 on Build GRCh38
3463651 c.2368A>T p.I790L Chr.12: 57096748-57096748 on Build GRCh38
6137755 c.2258C>T p.A753V Chr.12: 57096946-57096946 on Build GRCh38
3398920 c.2315G>A p.C772Y Chr.12: 57096889-57096889 on Build GRCh38
1988304 c.2281C>T p.L761F Chr.12: 57096923-57096923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373268442 Chr.12: 57096967-57096967 on Build GRCh38
rs201768332 Chr.12: 57096914-57096914 on Build GRCh38
rs754668222 Chr.12: 57096976-57096976 on Build GRCh38
rs960406056 Chr.12: 57096950-57096950 on Build GRCh38
rs369234436 Chr.12: 57096937-57096937 on Build GRCh38
rs1018001291 Chr.12: 57096958-57096958 on Build GRCh38
rs1017208191 Chr.12: 57096898-57096898 on Build GRCh38
rs180715438 Chr.12: 57096904-57096904 on Build GRCh38
rs576330506 Chr.12: 57096828-57096828 on Build GRCh38
rs763425373 Chr.12: 57096841-57096841 on Build GRCh38
rs769007397 Chr.12: 57096819-57096819 on Build GRCh38
rs757093061 Chr.12: 57096768-57096768 on Build GRCh38
rs749736022 Chr.12: 57096938-57096938 on Build GRCh38
rs753111770 Chr.12: 57096878-57096878 on Build GRCh38
rs777099800 Chr.12: 57096823-57096823 on Build GRCh38
rs780219389 Chr.12: 57096900-57096900 on Build GRCh38
rs774984579 Chr.12: 57096855-57096855 on Build GRCh38
rs751722933 Chr.12: 57096902-57096902 on Build GRCh38
rs775228499 Chr.12: 57096891-57096892 on Build GRCh38
rs781440827 Chr.12: 57096916-57096916 on Build GRCh38
rs747173993 Chr.12: 57096809-57096809 on Build GRCh38
rs750363199 Chr.12: 57096882-57096882 on Build GRCh38
rs760279462 Chr.12: 57096857-57096857 on Build GRCh38
rs761639300 Chr.12: 57096870-57096870 on Build GRCh38
rs760154963 Chr.12: 57096953-57096953 on Build GRCh38
rs765026200 Chr.12: 57096983-57096983 on Build GRCh38
rs138011454 Chr.12: 57096945-57096945 on Build GRCh38
rs145346629 Chr.12: 57096868-57096868 on Build GRCh38
rs763758520 Chr.12: 57096867-57096867 on Build GRCh38
rs149082783 Chr.12: 57096966-57096966 on Build GRCh38
rs761431628 Chr.12: 57096982-57096982 on Build GRCh38
rs866002159 Chr.12: 57096946-57096946 on Build GRCh38
rs200876370 Chr.12: 57096949-57096949 on Build GRCh38
rs375600000 Chr.12: 57096845-57096845 on Build GRCh38
rs758425219 Chr.12: 57096893-57096893 on Build GRCh38
rs372642833 Chr.12: 57096799-57096799 on Build GRCh38
rs766952769 Chr.12: 57096847-57096847 on Build GRCh38
rs780340575 Chr.12: 57096807-57096807 on Build GRCh38
rs184794768 Chr.12: 57096800-57096800 on Build GRCh38
rs760080733 Chr.12: 57096895-57096896 on Build GRCh38
rs778888759 Chr.12: 57096792-57096792 on Build GRCh38
rs748570794 Chr.12: 57096824-57096824 on Build GRCh38
rs748381508 Chr.12: 57096925-57096925 on Build GRCh38
rs770178770 Chr.12: 57096936-57096936 on Build GRCh38
rs755206351 Chr.12: 57096915-57096915 on Build GRCh38

View Full Product Details