Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212791819-212792078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSL1
Entrez Gene ID
25936
Gene Name
NSL1, MIS12 kinetochore complex component
Gene Aliases
C1orf48, DC8, MIS14
UniGene
Hs.497692
Gene Chromosome Location
Chr.1: 212726153-212798900 on Build GRCh38
Transcripts
XM_017000968.1 XM_017000969.1 NM_001042552.2 NM_001042553.2 NM_001146169.1 NM_001146170.1 NM_001146171.1 XM_011509202.2 XM_017000328.1 XM_017000329.1 XM_017000330.1 XM_017000331.1

Gene Symbol
TATDN3
Entrez Gene ID
128387
Gene Name
TatD DNase domain containing 3
Gene Aliases
-
UniGene
Hs.530538
Gene Chromosome Location
Chr.1: 212791828-212816825 on Build GRCh38
Transcripts
XM_017000968.1 XM_017000969.1 NM_001042552.2 NM_001042553.2 NM_001146169.1 NM_001146170.1 NM_001146171.1 XM_011509202.2 XM_017000328.1 XM_017000329.1 XM_017000330.1 XM_017000331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6496659 c.24G>C p.L8F Chr.1: 212791945-212791945 on Build GRCh38
425259 c.21C>G p.G7G Chr.1: 212791942-212791942 on Build GRCh38
5174624 c.10G>A p.A4T Chr.1: 212791931-212791931 on Build GRCh38
6367721 c.50C>A p.A17D Chr.1: 212791971-212791971 on Build GRCh38
678699 c.55G>C p.D19H Chr.1: 212791976-212791976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377084448 Chr.1: 212791969-212791969 on Build GRCh38
rs753199404 Chr.1: 212791886-212791886 on Build GRCh38
rs200532843 Chr.1: 212792002-212792002 on Build GRCh38
rs748135480 Chr.1: 212791949-212791949 on Build GRCh38
rs533667077 Chr.1: 212791980-212791980 on Build GRCh38
rs377530037 Chr.1: 212792018-212792018 on Build GRCh38
rs752149740 Chr.1: 212791922-212791922 on Build GRCh38
rs980644855 Chr.1: 212791850-212791850 on Build GRCh38
rs900989635 Chr.1: 212791905-212791905 on Build GRCh38
rs61736925 Chr.1: 212791930-212791930 on Build GRCh38
rs768495915 Chr.1: 212791904-212791904 on Build GRCh38
rs1037225640 Chr.1: 212791866-212791866 on Build GRCh38
rs757477847 Chr.1: 212791893-212791893 on Build GRCh38
rs752592724 Chr.1: 212792031-212792031 on Build GRCh38
rs57451249 Chr.1: 212791897-212791897 on Build GRCh38
rs918834878 Chr.1: 212792030-212792030 on Build GRCh38
rs749178337 Chr.1: 212791902-212791902 on Build GRCh38
rs760871693 Chr.1: 212792009-212792009 on Build GRCh38
rs542317831 Chr.1: 212791951-212791951 on Build GRCh38
rs750145439 Chr.1: 212791876-212791876 on Build GRCh38
rs754748482 Chr.1: 212792024-212792024 on Build GRCh38
rs1010776617 Chr.1: 212791894-212791894 on Build GRCh38
rs375418818 Chr.1: 212791889-212791889 on Build GRCh38
rs577834282 Chr.1: 212792050-212792050 on Build GRCh38
rs774334959 Chr.1: 212791909-212791909 on Build GRCh38
rs1048839553 Chr.1: 212792044-212792044 on Build GRCh38
rs942755192 Chr.1: 212792051-212792051 on Build GRCh38
rs767724016 Chr.1: 212791926-212791926 on Build GRCh38
rs772741669 Chr.1: 212791962-212791962 on Build GRCh38
rs754133081 Chr.1: 212792023-212792023 on Build GRCh38
rs778799691 Chr.1: 212791877-212791877 on Build GRCh38
rs772831638 Chr.1: 212791914-212791914 on Build GRCh38
rs185741445 Chr.1: 212791988-212791988 on Build GRCh38
rs756290328 Chr.1: 212791933-212791933 on Build GRCh38
rs3738799 Chr.1: 212791895-212791895 on Build GRCh38
rs760268927 Chr.1: 212791915-212791915 on Build GRCh38
rs146856249 Chr.1: 212791950-212791950 on Build GRCh38
rs774376959 Chr.1: 212791853-212791853 on Build GRCh38
rs115784129 Chr.1: 212791920-212791920 on Build GRCh38
rs372927766 Chr.1: 212791875-212791875 on Build GRCh38
rs61736926 Chr.1: 212791942-212791942 on Build GRCh38
rs938911411 Chr.1: 212791864-212791864 on Build GRCh38
rs765598422 Chr.1: 212791917-212791917 on Build GRCh38
rs779844019 Chr.1: 212791935-212791935 on Build GRCh38
rs180739415 Chr.1: 212791885-212791885 on Build GRCh38
rs745828342 Chr.1: 212791938-212791939 on Build GRCh38
rs200093236 Chr.1: 212791945-212791945 on Build GRCh38
rs751675806 Chr.1: 212791892-212791892 on Build GRCh38
rs755872731 Chr.1: 212791991-212791991 on Build GRCh38
rs371080805 Chr.1: 212792019-212792019 on Build GRCh38
rs771612725 Chr.1: 212791913-212791913 on Build GRCh38
rs947834929 Chr.1: 212791840-212791840 on Build GRCh38
rs188376617 Chr.1: 212791874-212791874 on Build GRCh38
rs749027271 Chr.1: 212791936-212791936 on Build GRCh38
rs373601307 Chr.1: 212792010-212792010 on Build GRCh38
rs750477887 Chr.1: 212792007-212792007 on Build GRCh38
rs778703681 Chr.1: 212792029-212792029 on Build GRCh38
rs931585281 Chr.1: 212792042-212792042 on Build GRCh38
rs989514049 Chr.1: 212791931-212791931 on Build GRCh38
rs763195697 Chr.1: 212791984-212791984 on Build GRCh38
rs906225564 Chr.1: 212791855-212791855 on Build GRCh38
rs994163615 Chr.1: 212791839-212791839 on Build GRCh38
rs757387845 Chr.1: 212791923-212791923 on Build GRCh38
rs775722225 Chr.1: 212791918-212791918 on Build GRCh38
rs561364755 Chr.1: 212791865-212791865 on Build GRCh38
rs369482556 Chr.1: 212791990-212791990 on Build GRCh38
rs201732884 Chr.1: 212792003-212792003 on Build GRCh38
rs759554345 Chr.1: 212791888-212791888 on Build GRCh38
rs759864779 Chr.1: 212791879-212791879 on Build GRCh38

View Full Product Details